Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37861043-37861400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF10
Entrez Gene ID
79269
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
WDR32
UniGene
Hs.118394
Gene Chromosome Location
Chr.9: 37800554-37867669 on Build GRCh38
Transcripts
NM_001286810.1 NM_024345.4 XM_005251577.3 XM_005251578.3 XM_017015128.1 XM_017015129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109082 c.1564C>T p.R522C Chr.9: 37861392-37861392 on Build GRCh38
4676311 c.1395T>C p.I465I Chr.9: 37861223-37861223 on Build GRCh38
4676310 c.1379G>A p.R460Q Chr.9: 37861207-37861207 on Build GRCh38
1109081 c.1560G>T p.V520V Chr.9: 37861388-37861388 on Build GRCh38
1196147 c.1555C>T p.R519W Chr.9: 37861383-37861383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71494680 Chr.9: 37861263-37861263 on Build GRCh38
rs780960408 Chr.9: 37861134-37861134 on Build GRCh38
rs778016658 Chr.9: 37861361-37861361 on Build GRCh38
rs758068545 Chr.9: 37861282-37861282 on Build GRCh38
rs756696717 Chr.9: 37861100-37861100 on Build GRCh38
rs374875555 Chr.9: 37861116-37861116 on Build GRCh38
rs764892910 Chr.9: 37861366-37861366 on Build GRCh38
rs1011950008 Chr.9: 37861082-37861082 on Build GRCh38
rs759784118 Chr.9: 37861191-37861191 on Build GRCh38
rs749335106 Chr.9: 37861364-37861364 on Build GRCh38
rs748310902 Chr.9: 37861135-37861135 on Build GRCh38
rs778682229 Chr.9: 37861102-37861102 on Build GRCh38
rs201769665 Chr.9: 37861320-37861320 on Build GRCh38
rs754708460 Chr.9: 37861337-37861337 on Build GRCh38
rs771315533 Chr.9: 37861368-37861368 on Build GRCh38
rs774883992 Chr.9: 37861189-37861189 on Build GRCh38
rs763274317 Chr.9: 37861180-37861180 on Build GRCh38
rs768671135 Chr.9: 37861118-37861118 on Build GRCh38
rs907754896 Chr.9: 37861111-37861111 on Build GRCh38
rs765976905 Chr.9: 37861307-37861307 on Build GRCh38
rs761451489 Chr.9: 37861236-37861236 on Build GRCh38
rs751396066 Chr.9: 37861331-37861331 on Build GRCh38
rs773639107 Chr.9: 37861152-37861152 on Build GRCh38
rs568381039 Chr.9: 37861165-37861165 on Build GRCh38
rs202210611 Chr.9: 37861093-37861093 on Build GRCh38
rs750070041 Chr.9: 37861103-37861103 on Build GRCh38
rs948125986 Chr.9: 37861179-37861179 on Build GRCh38
rs749877265 Chr.9: 37861277-37861277 on Build GRCh38
rs200328443 Chr.9: 37861333-37861333 on Build GRCh38
rs113211375 Chr.9: 37861206-37861206 on Build GRCh38
rs368448137 Chr.9: 37861376-37861376 on Build GRCh38
rs748034198 Chr.9: 37861341-37861341 on Build GRCh38
rs151036318 Chr.9: 37861346-37861346 on Build GRCh38
rs375659508 Chr.9: 37861231-37861231 on Build GRCh38
rs1023283930 Chr.9: 37861083-37861083 on Build GRCh38
rs753128985 Chr.9: 37861234-37861234 on Build GRCh38
rs371412241 Chr.9: 37861184-37861184 on Build GRCh38
rs199538284 Chr.9: 37861274-37861274 on Build GRCh38
rs767868394 Chr.9: 37861095-37861095 on Build GRCh38
rs781249730 Chr.9: 37861338-37861338 on Build GRCh38
rs757976748 Chr.9: 37861114-37861114 on Build GRCh38
rs753424111 Chr.9: 37861096-37861096 on Build GRCh38
rs368436088 Chr.9: 37861140-37861140 on Build GRCh38

View Full Product Details