Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114431689-114432285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
UniGene
Hs.438236
Gene Chromosome Location
Chr.10: 114431110-114821643 on Build GRCh38
Transcripts
NM_001003407.1 NM_001003408.1 NM_001322882.1 NM_001322883.1 NM_001322884.1 NM_001322885.1 NM_001322886.1 NM_001322887.1 NM_001322888.1 NM_001322889.1 NM_001322890.1 NM_001322891.1 NM_001322892.1 NM_001322893.1 NM_001322894.1 NM_001322895.1 NM_001322896.1 NM_001322897.1 NM_001322898.1 NM_001322899.1 NM_001322900.1 NM_002313.5 NM_006720.3 XM_005269824.2 XM_005269826.1 XM_005269827.4 XM_006717837.1 XM_006717846.3 XM_006717847.1 XM_011539801.1 XM_011539802.1 XM_011539803.1 XM_011539805.1 XM_017016242.1 XM_017016243.1 XM_017016244.1 XM_017016245.1 XM_017016246.1 XM_017016247.1 XM_017016248.1 XM_017016249.1 XM_017016250.1 XM_017016251.1 XM_017016252.1 XM_017016253.1 XM_017016254.1 XM_017016255.1 XM_017016256.1 XM_017016258.1 XM_017016259.1

SNP Detail

SNP ID Chromosome Location
rs988475656 Chr.10: 114431753-114431753 on Build GRCh38
rs879723427 Chr.10: 114432228-114432228 on Build GRCh38
rs377273709 Chr.10: 114432037-114432037 on Build GRCh38
rs988526372 Chr.10: 114432046-114432046 on Build GRCh38
rs540657505 Chr.10: 114432096-114432096 on Build GRCh38
rs947969599 Chr.10: 114431880-114431880 on Build GRCh38
rs138269965 Chr.10: 114431904-114431904 on Build GRCh38
rs1056442668 Chr.10: 114432206-114432206 on Build GRCh38
rs554923947 Chr.10: 114431732-114431732 on Build GRCh38
rs1000026487 Chr.10: 114432022-114432022 on Build GRCh38
rs773879308 Chr.10: 114431822-114431822 on Build GRCh38
rs865990185 Chr.10: 114432102-114432102 on Build GRCh38
rs144127339 Chr.10: 114432217-114432217 on Build GRCh38
rs927584059 Chr.10: 114431906-114431906 on Build GRCh38
rs951260514 Chr.10: 114431751-114431751 on Build GRCh38
rs111614372 Chr.10: 114431930-114431930 on Build GRCh38
rs182582772 Chr.10: 114432020-114432020 on Build GRCh38
rs908010343 Chr.10: 114432180-114432180 on Build GRCh38
rs930807709 Chr.10: 114431823-114431823 on Build GRCh38
rs74160407 Chr.10: 114432239-114432239 on Build GRCh38
rs1049723477 Chr.10: 114431868-114431868 on Build GRCh38
rs1039259644 Chr.10: 114431817-114431817 on Build GRCh38
rs7916374 Chr.10: 114432040-114432040 on Build GRCh38
rs1000484320 Chr.10: 114432240-114432240 on Build GRCh38
rs564537370 Chr.10: 114432082-114432082 on Build GRCh38
rs573374748 Chr.10: 114432258-114432258 on Build GRCh38
rs904811261 Chr.10: 114432001-114432001 on Build GRCh38
rs925681186 Chr.10: 114431745-114431745 on Build GRCh38
rs1045508350 Chr.10: 114432237-114432237 on Build GRCh38
rs562109185 Chr.10: 114432193-114432193 on Build GRCh38
rs944362645 Chr.10: 114431809-114431809 on Build GRCh38
rs556096536 Chr.10: 114431929-114431929 on Build GRCh38
rs74582830 Chr.10: 114431992-114431993 on Build GRCh38
rs892204875 Chr.10: 114431959-114431959 on Build GRCh38
rs909825491 Chr.10: 114431869-114431869 on Build GRCh38
rs762166949 Chr.10: 114432119-114432119 on Build GRCh38
rs965718865 Chr.10: 114432065-114432065 on Build GRCh38
rs185927241 Chr.10: 114432092-114432092 on Build GRCh38
rs930924493 Chr.10: 114432098-114432098 on Build GRCh38
rs983905660 Chr.10: 114431816-114431816 on Build GRCh38
rs896592157 Chr.10: 114432212-114432212 on Build GRCh38
rs1010434852 Chr.10: 114431986-114431986 on Build GRCh38
rs5788065 Chr.10: 114431985-114431985 on Build GRCh38
rs1043245881 Chr.10: 114431989-114431989 on Build GRCh38
rs796679000 Chr.10: 114431993-114431993 on Build GRCh38
rs7917021 Chr.10: 114432200-114432200 on Build GRCh38
rs369895331 Chr.10: 114432055-114432055 on Build GRCh38
rs886517324 Chr.10: 114431899-114431899 on Build GRCh38
rs544487021 Chr.10: 114431918-114431918 on Build GRCh38
rs761153222 Chr.10: 114431919-114431919 on Build GRCh38
rs983957872 Chr.10: 114432115-114432115 on Build GRCh38
rs911523287 Chr.10: 114431781-114431781 on Build GRCh38
rs1018278440 Chr.10: 114432029-114432029 on Build GRCh38

View Full Product Details