Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52802254-52802851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

SNP Detail

SNP ID Chromosome Location
rs915631638 Chr.5: 52802373-52802373 on Build GRCh38
rs981567383 Chr.5: 52802717-52802717 on Build GRCh38
rs940535462 Chr.5: 52802299-52802299 on Build GRCh38
rs983086102 Chr.5: 52802757-52802757 on Build GRCh38
rs528765075 Chr.5: 52802810-52802810 on Build GRCh38
rs577211718 Chr.5: 52802362-52802362 on Build GRCh38
rs745760302 Chr.5: 52802518-52802518 on Build GRCh38
rs6870317 Chr.5: 52802428-52802428 on Build GRCh38
rs928403847 Chr.5: 52802737-52802737 on Build GRCh38
rs780701396 Chr.5: 52802537-52802537 on Build GRCh38
rs570614505 Chr.5: 52802593-52802593 on Build GRCh38
rs755993473 Chr.5: 52802667-52802667 on Build GRCh38
rs1036615687 Chr.5: 52802587-52802587 on Build GRCh38
rs909124201 Chr.5: 52802788-52802788 on Build GRCh38
rs769539020 Chr.5: 52802819-52802819 on Build GRCh38
rs1028545919 Chr.5: 52802764-52802764 on Build GRCh38
rs1055500785 Chr.5: 52802327-52802327 on Build GRCh38
rs1019823305 Chr.5: 52802425-52802425 on Build GRCh38
rs1011259605 Chr.5: 52802642-52802642 on Build GRCh38
rs747479120 Chr.5: 52802491-52802491 on Build GRCh38
rs977700312 Chr.5: 52802302-52802302 on Build GRCh38
rs369037479 Chr.5: 52802675-52802675 on Build GRCh38
rs188458182 Chr.5: 52802680-52802680 on Build GRCh38
rs894224981 Chr.5: 52802353-52802353 on Build GRCh38
rs1042802525 Chr.5: 52802666-52802666 on Build GRCh38
rs948510808 Chr.5: 52802381-52802381 on Build GRCh38
rs566206855 Chr.5: 52802649-52802649 on Build GRCh38
rs1023595397 Chr.5: 52802688-52802688 on Build GRCh38
rs894174066 Chr.5: 52802617-52802617 on Build GRCh38
rs7356703 Chr.5: 52802596-52802596 on Build GRCh38
rs921289526 Chr.5: 52802499-52802499 on Build GRCh38
rs866268597 Chr.5: 52802697-52802697 on Build GRCh38
rs1047611785 Chr.5: 52802496-52802496 on Build GRCh38
rs183263830 Chr.5: 52802516-52802516 on Build GRCh38
rs770376356 Chr.5: 52802531-52802531 on Build GRCh38
rs75391450 Chr.5: 52802817-52802817 on Build GRCh38
rs559395464 Chr.5: 52802360-52802360 on Build GRCh38
rs575426873 Chr.5: 52802745-52802745 on Build GRCh38
rs535537073 Chr.5: 52802725-52802725 on Build GRCh38
rs952785734 Chr.5: 52802513-52802513 on Build GRCh38
rs867150548 Chr.5: 52802825-52802825 on Build GRCh38
rs780997062 Chr.5: 52802412-52802412 on Build GRCh38
rs536161716 Chr.5: 52802493-52802493 on Build GRCh38
rs941839151 Chr.5: 52802522-52802522 on Build GRCh38
rs984141248 Chr.5: 52802515-52802515 on Build GRCh38
rs950348587 Chr.5: 52802754-52802754 on Build GRCh38
rs147586254 Chr.5: 52802413-52802413 on Build GRCh38
rs1031891348 Chr.5: 52802656-52802656 on Build GRCh38
rs1002510881 Chr.5: 52802317-52802317 on Build GRCh38
rs62358443 Chr.5: 52802542-52802542 on Build GRCh38
rs780675618 Chr.5: 52802590-52802591 on Build GRCh38
rs191748324 Chr.5: 52802729-52802729 on Build GRCh38
rs552437855 Chr.5: 52802568-52802568 on Build GRCh38
rs907118428 Chr.5: 52802316-52802317 on Build GRCh38
rs1031943851 Chr.5: 52802325-52802325 on Build GRCh38
rs569182369 Chr.5: 52802434-52802434 on Build GRCh38
rs981111257 Chr.5: 52802429-52802429 on Build GRCh38
rs891102819 Chr.5: 52802601-52802601 on Build GRCh38
rs191064521 Chr.5: 52802345-52802345 on Build GRCh38
rs957618785 Chr.5: 52802668-52802668 on Build GRCh38
rs549619537 Chr.5: 52802610-52802610 on Build GRCh38

View Full Product Details