Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142942731-142943309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KEL
Entrez Gene ID
3792
Gene Name
Kell blood group, metallo-endopeptidase
Gene Aliases
CD238, ECE3
UniGene
Hs.368588
Gene Chromosome Location
Chr.7: 142941114-142962416 on Build GRCh38
Transcripts
NM_000420.2 XM_005249993.2 XM_005249994.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508358 c.1806T>C p.H602H Chr.7: 142943010-142943010 on Build GRCh38
6109355 c.1743C>T p.H581H Chr.7: 142943304-142943304 on Build GRCh38
5467048 c.1771+6T>G p.? Chr.7: 142943270-142943270 on Build GRCh38
1448798 c.1794C>A p.A598A Chr.7: 142943022-142943022 on Build GRCh38
2151305 c.1914C>T p.D638D Chr.7: 142942902-142942902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747237668 Chr.7: 142942836-142942836 on Build GRCh38
rs1054734931 Chr.7: 142942884-142942884 on Build GRCh38
rs942761578 Chr.7: 142942802-142942802 on Build GRCh38
rs759242093 Chr.7: 142943054-142943054 on Build GRCh38
rs41309237 Chr.7: 142943007-142943007 on Build GRCh38
rs772178431 Chr.7: 142943013-142943013 on Build GRCh38
rs974248310 Chr.7: 142942994-142942994 on Build GRCh38
rs8176040 Chr.7: 142942759-142942759 on Build GRCh38
rs923741387 Chr.7: 142943119-142943119 on Build GRCh38
rs758961140 Chr.7: 142942978-142942978 on Build GRCh38
rs771122793 Chr.7: 142942899-142942899 on Build GRCh38
rs866749534 Chr.7: 142943036-142943036 on Build GRCh38
rs763976752 Chr.7: 142942860-142942860 on Build GRCh38
rs960927472 Chr.7: 142943123-142943123 on Build GRCh38
rs750172557 Chr.7: 142942874-142942874 on Build GRCh38
rs913078541 Chr.7: 142943110-142943110 on Build GRCh38
rs866567552 Chr.7: 142942960-142942960 on Build GRCh38
rs975850212 Chr.7: 142943056-142943056 on Build GRCh38
rs61729048 Chr.7: 142943284-142943284 on Build GRCh38
rs372615948 Chr.7: 142942839-142942839 on Build GRCh38
rs757662766 Chr.7: 142943093-142943093 on Build GRCh38
rs370914535 Chr.7: 142943212-142943212 on Build GRCh38
rs767499938 Chr.7: 142942883-142942883 on Build GRCh38
rs147342331 Chr.7: 142943238-142943238 on Build GRCh38
rs776223205 Chr.7: 142943047-142943047 on Build GRCh38
rs369564583 Chr.7: 142943080-142943080 on Build GRCh38
rs1022273157 Chr.7: 142943289-142943289 on Build GRCh38
rs755134816 Chr.7: 142943258-142943258 on Build GRCh38
rs374902111 Chr.7: 142942966-142942966 on Build GRCh38
rs61729043 Chr.7: 142942948-142942948 on Build GRCh38
rs898240420 Chr.7: 142943252-142943252 on Build GRCh38
rs776632496 Chr.7: 142942847-142942847 on Build GRCh38
rs138511569 Chr.7: 142943280-142943280 on Build GRCh38
rs755893574 Chr.7: 142942880-142942880 on Build GRCh38
rs544863534 Chr.7: 142942799-142942799 on Build GRCh38
rs762416377 Chr.7: 142943073-142943073 on Build GRCh38
rs138693131 Chr.7: 142942796-142942796 on Build GRCh38
rs1055890106 Chr.7: 142943223-142943223 on Build GRCh38
rs752126191 Chr.7: 142943089-142943089 on Build GRCh38
rs1056547720 Chr.7: 142942766-142942766 on Build GRCh38
rs916471611 Chr.7: 142943153-142943153 on Build GRCh38
rs753552670 Chr.7: 142942949-142942949 on Build GRCh38
rs773211677 Chr.7: 142942944-142942944 on Build GRCh38
rs8176038 Chr.7: 142943026-142943026 on Build GRCh38
rs549232567 Chr.7: 142943203-142943203 on Build GRCh38
rs869025768 Chr.7: 142943275-142943275 on Build GRCh38
rs939217070 Chr.7: 142942760-142942760 on Build GRCh38
rs185432321 Chr.7: 142943000-142943000 on Build GRCh38
rs995695928 Chr.7: 142943282-142943282 on Build GRCh38
rs760490521 Chr.7: 142942947-142942947 on Build GRCh38
rs775667424 Chr.7: 142942911-142942911 on Build GRCh38
rs111834310 Chr.7: 142943070-142943070 on Build GRCh38
rs200383000 Chr.7: 142942954-142942954 on Build GRCh38
rs142691451 Chr.7: 142942813-142942813 on Build GRCh38
rs749431493 Chr.7: 142943256-142943256 on Build GRCh38
rs552971293 Chr.7: 142943281-142943281 on Build GRCh38
rs372478760 Chr.7: 142943085-142943085 on Build GRCh38
rs367661434 Chr.7: 142942902-142942902 on Build GRCh38
rs759072657 Chr.7: 142942951-142942951 on Build GRCh38
rs371547298 Chr.7: 142942959-142942959 on Build GRCh38
rs750691476 Chr.7: 142943239-142943239 on Build GRCh38
rs945907123 Chr.7: 142943149-142943149 on Build GRCh38
rs925374641 Chr.7: 142943062-142943062 on Build GRCh38
rs773526421 Chr.7: 142942831-142942831 on Build GRCh38
rs552745361 Chr.7: 142942778-142942778 on Build GRCh38
rs375875934 Chr.7: 142942867-142942867 on Build GRCh38
rs749378396 Chr.7: 142943004-142943004 on Build GRCh38
rs369331299 Chr.7: 142942996-142942996 on Build GRCh38
rs766053821 Chr.7: 142942881-142942881 on Build GRCh38
rs773342756 Chr.7: 142942818-142942818 on Build GRCh38
rs573337898 Chr.7: 142942758-142942758 on Build GRCh38
rs1025731174 Chr.7: 142943283-142943283 on Build GRCh38
rs747360606 Chr.7: 142942897-142942897 on Build GRCh38
rs751958640 Chr.7: 142942979-142942979 on Build GRCh38
rs759655406 Chr.7: 142942853-142942853 on Build GRCh38
rs140258059 Chr.7: 142942893-142942893 on Build GRCh38
rs996196122 Chr.7: 142943277-142943277 on Build GRCh38
rs758007153 Chr.7: 142942954-142942954 on Build GRCh38
rs375062353 Chr.7: 142943025-142943025 on Build GRCh38
rs780395823 Chr.7: 142943255-142943255 on Build GRCh38
rs768779190 Chr.7: 142943157-142943157 on Build GRCh38
rs757674085 Chr.7: 142943272-142943272 on Build GRCh38
rs748685628 Chr.7: 142942892-142942892 on Build GRCh38
rs929692521 Chr.7: 142943199-142943199 on Build GRCh38
rs745981653 Chr.7: 142942901-142942901 on Build GRCh38
rs368328555 Chr.7: 142942828-142942828 on Build GRCh38
rs537335736 Chr.7: 142943197-142943197 on Build GRCh38
rs572463647 Chr.7: 142943046-142943046 on Build GRCh38
rs977918956 Chr.7: 142942755-142942755 on Build GRCh38
rs8176037 Chr.7: 142943190-142943190 on Build GRCh38
rs1009459527 Chr.7: 142943218-142943218 on Build GRCh38
rs202117440 Chr.7: 142943069-142943069 on Build GRCh38
rs542976466 Chr.7: 142942896-142942896 on Build GRCh38
rs8176039 Chr.7: 142942917-142942917 on Build GRCh38
rs772125360 Chr.7: 142942942-142942942 on Build GRCh38
rs781442954 Chr.7: 142942900-142942900 on Build GRCh38
rs116257811 Chr.7: 142942870-142942870 on Build GRCh38
rs781359849 Chr.7: 142942992-142942992 on Build GRCh38
rs752390429 Chr.7: 142942892-142942893 on Build GRCh38
rs149066842 Chr.7: 142943024-142943024 on Build GRCh38
rs774405908 Chr.7: 142942866-142942866 on Build GRCh38
rs780251767 Chr.7: 142942822-142942823 on Build GRCh38
rs756428775 Chr.7: 142943247-142943247 on Build GRCh38
rs147851584 Chr.7: 142942882-142942882 on Build GRCh38

View Full Product Details