Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9965270-9965578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL1
Entrez Gene ID
29841
Gene Name
grainyhead like transcription factor 1
Gene Aliases
LBP32, MGR, NH32, TFCP2L2
UniGene
Hs.418493
Gene Chromosome Location
Chr.2: 9951663-10002284 on Build GRCh38
Transcripts
NM_198182.2 XM_005246159.3 XM_006711882.2 XM_006711884.3 XM_011510343.2 XM_017003900.1 XM_017003901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208867 c.499T>C p.Y167H Chr.2: 9965337-9965337 on Build GRCh38
3564293 c.513C>T p.S171S Chr.2: 9965351-9965351 on Build GRCh38
4688458 c.1071C>T p.N357N Chr.2: 9965342-9965342 on Build GRCh38
3564292 c.1080C>T p.S360S Chr.2: 9965351-9965351 on Build GRCh38
4688461 c.1072G>A p.A358T Chr.2: 9965343-9965343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771855601 Chr.2: 9965317-9965317 on Build GRCh38
rs77303006 Chr.2: 9965469-9965469 on Build GRCh38
rs2303919 Chr.2: 9965530-9965530 on Build GRCh38
rs373058096 Chr.2: 9965405-9965405 on Build GRCh38
rs367582209 Chr.2: 9965324-9965324 on Build GRCh38
rs1028725200 Chr.2: 9965468-9965469 on Build GRCh38
rs148615174 Chr.2: 9965345-9965345 on Build GRCh38
rs41264203 Chr.2: 9965406-9965406 on Build GRCh38
rs11884250 Chr.2: 9965515-9965515 on Build GRCh38
rs915194552 Chr.2: 9965540-9965540 on Build GRCh38
rs990887083 Chr.2: 9965548-9965548 on Build GRCh38
rs771093470 Chr.2: 9965389-9965389 on Build GRCh38
rs749544331 Chr.2: 9965381-9965381 on Build GRCh38
rs79082586 Chr.2: 9965468-9965468 on Build GRCh38
rs772933149 Chr.2: 9965369-9965369 on Build GRCh38
rs758017922 Chr.2: 9965333-9965333 on Build GRCh38
rs1027927143 Chr.2: 9965496-9965496 on Build GRCh38
rs145297770 Chr.2: 9965308-9965308 on Build GRCh38
rs774256858 Chr.2: 9965306-9965306 on Build GRCh38
rs77941294 Chr.2: 9965455-9965455 on Build GRCh38
rs556635581 Chr.2: 9965312-9965312 on Build GRCh38
rs369326255 Chr.2: 9965404-9965404 on Build GRCh38
rs774504745 Chr.2: 9965391-9965391 on Build GRCh38
rs529271384 Chr.2: 9965418-9965418 on Build GRCh38
rs756163592 Chr.2: 9965455-9965456 on Build GRCh38
rs995142919 Chr.2: 9965492-9965492 on Build GRCh38
rs185240007 Chr.2: 9965401-9965401 on Build GRCh38
rs147415325 Chr.2: 9965301-9965301 on Build GRCh38
rs115772224 Chr.2: 9965357-9965357 on Build GRCh38
rs1003685627 Chr.2: 9965297-9965297 on Build GRCh38
rs111854908 Chr.2: 9965456-9965456 on Build GRCh38
rs140008573 Chr.2: 9965291-9965291 on Build GRCh38
rs758808349 Chr.2: 9965546-9965548 on Build GRCh38
rs955515633 Chr.2: 9965495-9965495 on Build GRCh38
rs753673674 Chr.2: 9965313-9965313 on Build GRCh38
rs950958002 Chr.2: 9965500-9965500 on Build GRCh38
rs758142886 Chr.2: 9965411-9965411 on Build GRCh38
rs988066604 Chr.2: 9965518-9965518 on Build GRCh38
rs781482693 Chr.2: 9965352-9965352 on Build GRCh38
rs750082521 Chr.2: 9965326-9965326 on Build GRCh38
rs758948219 Chr.2: 9965407-9965407 on Build GRCh38
rs151143088 Chr.2: 9965363-9965363 on Build GRCh38
rs879165409 Chr.2: 9965443-9965443 on Build GRCh38
rs746451999 Chr.2: 9965342-9965342 on Build GRCh38
rs759743031 Chr.2: 9965508-9965508 on Build GRCh38
rs779642667 Chr.2: 9965336-9965336 on Build GRCh38
rs371814208 Chr.2: 9965370-9965370 on Build GRCh38
rs1005469329 Chr.2: 9965506-9965506 on Build GRCh38

View Full Product Details