Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3069272-3069813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL32
Entrez Gene ID
9235
Gene Name
interleukin 32
Gene Aliases
IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
UniGene
Hs.943
Gene Chromosome Location
Chr.16: 3065312-3069819 on Build GRCh38
Transcripts
NM_001012631.1 NM_001012632.1 NM_001012633.1 NM_001012634.1 NM_001012635.1 NM_001012636.1 NM_001012718.1 NM_001308078.1 NM_004221.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144339 c.521A>T p.E174V Chr.16: 3069309-3069309 on Build GRCh38
435163 c.498C>T p.Y166Y Chr.16: 3069286-3069286 on Build GRCh38
87703 c.509_510insG p.D172fs*12 Chr.16: 3069297-3069298 on Build GRCh38
6079037 c.523G>T p.E175* Chr.16: 3069311-3069311 on Build GRCh38
6079038 c.505C>A p.P169T Chr.16: 3069293-3069293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748787639 Chr.16: 3069740-3069740 on Build GRCh38
rs189696299 Chr.16: 3069441-3069441 on Build GRCh38
rs998542892 Chr.16: 3069496-3069496 on Build GRCh38
rs143941291 Chr.16: 3069338-3069338 on Build GRCh38
rs769043924 Chr.16: 3069350-3069350 on Build GRCh38
rs929341485 Chr.16: 3069453-3069453 on Build GRCh38
rs71818662 Chr.16: 3069296-3069297 on Build GRCh38
rs556921671 Chr.16: 3069542-3069542 on Build GRCh38
rs903106288 Chr.16: 3069664-3069664 on Build GRCh38
rs766011549 Chr.16: 3069320-3069320 on Build GRCh38
rs758992017 Chr.16: 3069407-3069407 on Build GRCh38
rs902822521 Chr.16: 3069494-3069494 on Build GRCh38
rs938019564 Chr.16: 3069775-3069775 on Build GRCh38
rs763134593 Chr.16: 3069379-3069379 on Build GRCh38
rs1032687170 Chr.16: 3069504-3069504 on Build GRCh38
rs887450498 Chr.16: 3069644-3069644 on Build GRCh38
rs765849632 Chr.16: 3069362-3069362 on Build GRCh38
rs150738098 Chr.16: 3069312-3069312 on Build GRCh38
rs374074230 Chr.16: 3069382-3069382 on Build GRCh38
rs780936859 Chr.16: 3069330-3069330 on Build GRCh38
rs375758526 Chr.16: 3069465-3069465 on Build GRCh38
rs1007250848 Chr.16: 3069698-3069698 on Build GRCh38
rs990881484 Chr.16: 3069777-3069777 on Build GRCh38
rs12449264 Chr.16: 3069489-3069489 on Build GRCh38
rs377536439 Chr.16: 3069308-3069308 on Build GRCh38
rs976984785 Chr.16: 3069485-3069485 on Build GRCh38
rs961833554 Chr.16: 3069472-3069472 on Build GRCh38
rs529819738 Chr.16: 3069375-3069375 on Build GRCh38
rs544334042 Chr.16: 3069772-3069772 on Build GRCh38
rs925960676 Chr.16: 3069535-3069535 on Build GRCh38
rs775205321 Chr.16: 3069300-3069301 on Build GRCh38
rs1017718500 Chr.16: 3069537-3069537 on Build GRCh38
rs1001597122 Chr.16: 3069454-3069454 on Build GRCh38
rs950178190 Chr.16: 3069789-3069789 on Build GRCh38
rs932945788 Chr.16: 3069495-3069495 on Build GRCh38
rs561075523 Chr.16: 3069682-3069682 on Build GRCh38
rs528434073 Chr.16: 3069440-3069440 on Build GRCh38
rs749912248 Chr.16: 3069314-3069314 on Build GRCh38
rs191704156 Chr.16: 3069554-3069554 on Build GRCh38
rs770464943 Chr.16: 3069298-3069299 on Build GRCh38
rs748660588 Chr.16: 3069396-3069396 on Build GRCh38
rs985429330 Chr.16: 3069529-3069529 on Build GRCh38
rs894296981 Chr.16: 3069618-3069618 on Build GRCh38
rs369007437 Chr.16: 3069348-3069348 on Build GRCh38
rs140102091 Chr.16: 3069325-3069325 on Build GRCh38
rs746055082 Chr.16: 3069345-3069345 on Build GRCh38
rs532050114 Chr.16: 3069778-3069778 on Build GRCh38
rs759220727 Chr.16: 3069369-3069369 on Build GRCh38
rs147297738 Chr.16: 3069343-3069343 on Build GRCh38
rs185335398 Chr.16: 3069374-3069374 on Build GRCh38
rs563290217 Chr.16: 3069773-3069773 on Build GRCh38
rs547462668 Chr.16: 3069346-3069346 on Build GRCh38
rs183072816 Chr.16: 3069463-3069463 on Build GRCh38
rs966265488 Chr.16: 3069540-3069540 on Build GRCh38
rs748058182 Chr.16: 3069296-3069296 on Build GRCh38
rs772017218 Chr.16: 3069299-3069300 on Build GRCh38
rs574564850 Chr.16: 3069464-3069464 on Build GRCh38
rs772552901 Chr.16: 3069307-3069307 on Build GRCh38
rs111432878 Chr.16: 3069471-3069471 on Build GRCh38
rs779682227 Chr.16: 3069708-3069708 on Build GRCh38
rs780352930 Chr.16: 3069390-3069390 on Build GRCh38
rs762036065 Chr.16: 3069768-3069768 on Build GRCh38
rs949906241 Chr.16: 3069579-3069579 on Build GRCh38
rs539295865 Chr.16: 3069410-3069410 on Build GRCh38
rs919530656 Chr.16: 3069557-3069557 on Build GRCh38
rs181077045 Chr.16: 3069319-3069319 on Build GRCh38
rs71391801 Chr.16: 3069386-3069387 on Build GRCh38
rs372935622 Chr.16: 3069400-3069400 on Build GRCh38
rs541184024 Chr.16: 3069603-3069603 on Build GRCh38
rs2981599 Chr.16: 3069303-3069303 on Build GRCh38
rs929265110 Chr.16: 3069560-3069560 on Build GRCh38
rs79583387 Chr.16: 3069383-3069383 on Build GRCh38
rs749342733 Chr.16: 3069299-3069299 on Build GRCh38
rs779364490 Chr.16: 3069780-3069780 on Build GRCh38
rs749333838 Chr.16: 3069340-3069340 on Build GRCh38
rs1046030093 Chr.16: 3069646-3069646 on Build GRCh38
rs398100042 Chr.16: 3069303-3069304 on Build GRCh38
rs985712519 Chr.16: 3069752-3069752 on Build GRCh38
rs951828852 Chr.16: 3069301-3069301 on Build GRCh38
rs755531826 Chr.16: 3069757-3069757 on Build GRCh38
rs1014122664 Chr.16: 3069431-3069431 on Build GRCh38
rs112304177 Chr.16: 3069569-3069569 on Build GRCh38
rs781656938 Chr.16: 3069295-3069296 on Build GRCh38
rs949935270 Chr.16: 3069641-3069641 on Build GRCh38
rs183602551 Chr.16: 3069562-3069562 on Build GRCh38
rs762454460 Chr.16: 3069316-3069316 on Build GRCh38
rs751197524 Chr.16: 3069323-3069323 on Build GRCh38
rs374547717 Chr.16: 3069391-3069391 on Build GRCh38
rs1014623442 Chr.16: 3069696-3069696 on Build GRCh38
rs1029843272 Chr.16: 3069724-3069724 on Build GRCh38
rs767112161 Chr.16: 3069371-3069371 on Build GRCh38
rs925741339 Chr.16: 3069377-3069377 on Build GRCh38
rs150104859 Chr.16: 3069592-3069592 on Build GRCh38
rs371381564 Chr.16: 3069389-3069389 on Build GRCh38
rs913295079 Chr.16: 3069784-3069784 on Build GRCh38
rs985076942 Chr.16: 3069772-3069772 on Build GRCh38
rs1053185799 Chr.16: 3069556-3069556 on Build GRCh38
rs764612413 Chr.16: 3069313-3069313 on Build GRCh38
rs577198356 Chr.16: 3069553-3069553 on Build GRCh38
rs141220342 Chr.16: 3069302-3069302 on Build GRCh38
rs1052917189 Chr.16: 3069412-3069412 on Build GRCh38
rs752453644 Chr.16: 3069331-3069331 on Build GRCh38
rs1004536871 Chr.16: 3069680-3069680 on Build GRCh38
rs967102447 Chr.16: 3069363-3069363 on Build GRCh38
rs534864975 Chr.16: 3069458-3069458 on Build GRCh38
rs145823986 Chr.16: 3069710-3069710 on Build GRCh38
rs760045825 Chr.16: 3069304-3069304 on Build GRCh38
rs1001480181 Chr.16: 3069602-3069602 on Build GRCh38
rs748964193 Chr.16: 3069297-3069298 on Build GRCh38
rs559031069 Chr.16: 3069430-3069430 on Build GRCh38
rs551467687 Chr.16: 3069475-3069475 on Build GRCh38
rs371524589 Chr.16: 3069473-3069473 on Build GRCh38
rs377647974 Chr.16: 3069384-3069384 on Build GRCh38
rs755655795 Chr.16: 3069598-3069598 on Build GRCh38
rs187915770 Chr.16: 3069742-3069742 on Build GRCh38
rs773877730 Chr.16: 3069399-3069399 on Build GRCh38
rs762425007 Chr.16: 3069360-3069360 on Build GRCh38
rs777050227 Chr.16: 3069392-3069392 on Build GRCh38
rs905806019 Chr.16: 3069449-3069449 on Build GRCh38
rs368836091 Chr.16: 3069366-3069366 on Build GRCh38
rs760465682 Chr.16: 3069301-3069302 on Build GRCh38
rs781160848 Chr.16: 3069293-3069293 on Build GRCh38
rs919238538 Chr.16: 3069452-3069453 on Build GRCh38
rs773468767 Chr.16: 3069320-3069320 on Build GRCh38
rs941134897 Chr.16: 3069470-3069470 on Build GRCh38
rs531600758 Chr.16: 3069297-3069297 on Build GRCh38

View Full Product Details