Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151259444-151260043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GM2A
Entrez Gene ID
2760
Gene Name
GM2 ganglioside activator
Gene Aliases
GM2-AP, SAP-3
UniGene
Hs.483873
Gene Chromosome Location
Chr.5: 151253052-151270394 on Build GRCh38
Transcripts
NM_000405.4 NM_001167607.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218830 c.114delC p.C39fs*9 Chr.5: 151259787-151259787 on Build GRCh38
1064531 c.220G>A p.V74I Chr.5: 151259893-151259893 on Build GRCh38
3244693 c.122A>T p.E41V Chr.5: 151259795-151259795 on Build GRCh38
1241216 c.137C>T p.A46V Chr.5: 151259810-151259810 on Build GRCh38
5426808 c.175A>G p.I59V Chr.5: 151259848-151259848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761268810 Chr.5: 151259810-151259810 on Build GRCh38
rs773683744 Chr.5: 151259815-151259815 on Build GRCh38
rs16838 Chr.5: 151259601-151259601 on Build GRCh38
rs375938028 Chr.5: 151259720-151259720 on Build GRCh38
rs182903268 Chr.5: 151259528-151259528 on Build GRCh38
rs993224658 Chr.5: 151259634-151259634 on Build GRCh38
rs752936822 Chr.5: 151259944-151259944 on Build GRCh38
rs147511920 Chr.5: 151259811-151259811 on Build GRCh38
rs975289769 Chr.5: 151259787-151259787 on Build GRCh38
rs142082513 Chr.5: 151259976-151259976 on Build GRCh38
rs757366495 Chr.5: 151259957-151259957 on Build GRCh38
rs1054356748 Chr.5: 151259639-151259639 on Build GRCh38
rs998154333 Chr.5: 151259982-151259982 on Build GRCh38
rs1026306619 Chr.5: 151259684-151259684 on Build GRCh38
rs763407122 Chr.5: 151259822-151259822 on Build GRCh38
rs1002884421 Chr.5: 151259673-151259673 on Build GRCh38
rs104893897 Chr.5: 151259833-151259833 on Build GRCh38
rs557498468 Chr.5: 151259596-151259596 on Build GRCh38
rs369106753 Chr.5: 151259740-151259740 on Build GRCh38
rs376412265 Chr.5: 151259484-151259484 on Build GRCh38
rs893006434 Chr.5: 151259640-151259640 on Build GRCh38
rs730882196 Chr.5: 151259837-151259837 on Build GRCh38
rs153476 Chr.5: 151259663-151259663 on Build GRCh38
rs752989938 Chr.5: 151259847-151259847 on Build GRCh38
rs770025057 Chr.5: 151259759-151259759 on Build GRCh38
rs141199254 Chr.5: 151259855-151259855 on Build GRCh38
rs1030500560 Chr.5: 151259997-151259997 on Build GRCh38
rs751772881 Chr.5: 151259737-151259737 on Build GRCh38
rs375909851 Chr.5: 151259806-151259806 on Build GRCh38
rs149636307 Chr.5: 151259650-151259650 on Build GRCh38
rs780328889 Chr.5: 151259736-151259736 on Build GRCh38
rs776214439 Chr.5: 151259877-151259877 on Build GRCh38
rs369748999 Chr.5: 151259951-151259951 on Build GRCh38
rs371296403 Chr.5: 151259763-151259763 on Build GRCh38
rs766738446 Chr.5: 151259925-151259925 on Build GRCh38
rs756108911 Chr.5: 151259718-151259720 on Build GRCh38
rs201633382 Chr.5: 151259903-151259903 on Build GRCh38
rs779819860 Chr.5: 151259961-151259961 on Build GRCh38
rs773722449 Chr.5: 151259510-151259510 on Build GRCh38
rs1054345682 Chr.5: 151259521-151259521 on Build GRCh38
rs1039483615 Chr.5: 151259520-151259520 on Build GRCh38
rs768523926 Chr.5: 151259868-151259868 on Build GRCh38
rs564494401 Chr.5: 151259932-151259932 on Build GRCh38
rs775218532 Chr.5: 151259773-151259773 on Build GRCh38
rs1051159610 Chr.5: 151259619-151259619 on Build GRCh38
rs902845100 Chr.5: 151259633-151259633 on Build GRCh38
rs1047548185 Chr.5: 151259921-151259921 on Build GRCh38
rs745799277 Chr.5: 151259850-151259850 on Build GRCh38
rs377641017 Chr.5: 151259950-151259950 on Build GRCh38
rs758710005 Chr.5: 151259735-151259735 on Build GRCh38
rs571763910 Chr.5: 151259490-151259490 on Build GRCh38
rs767781109 Chr.5: 151259846-151259846 on Build GRCh38
rs153478 Chr.5: 151259878-151259878 on Build GRCh38
rs760694909 Chr.5: 151260015-151260015 on Build GRCh38
rs779969671 Chr.5: 151259783-151259784 on Build GRCh38
rs759769913 Chr.5: 151259937-151259937 on Build GRCh38
rs920983022 Chr.5: 151259548-151259548 on Build GRCh38
rs774751651 Chr.5: 151259685-151259685 on Build GRCh38
rs153477 Chr.5: 151259848-151259848 on Build GRCh38
rs746984959 Chr.5: 151259965-151259965 on Build GRCh38
rs965313477 Chr.5: 151259686-151259686 on Build GRCh38
rs185826297 Chr.5: 151259638-151259638 on Build GRCh38
rs769261253 Chr.5: 151259879-151259879 on Build GRCh38
rs755382727 Chr.5: 151259845-151259845 on Build GRCh38
rs745850680 Chr.5: 151259765-151259765 on Build GRCh38
rs201829650 Chr.5: 151259851-151259851 on Build GRCh38
rs766759666 Chr.5: 151259826-151259826 on Build GRCh38
rs756473323 Chr.5: 151259947-151259947 on Build GRCh38
rs781318081 Chr.5: 151259746-151259746 on Build GRCh38
rs978077074 Chr.5: 151259494-151259494 on Build GRCh38
rs772774529 Chr.5: 151259895-151259895 on Build GRCh38
rs574730395 Chr.5: 151259702-151259702 on Build GRCh38
rs139369020 Chr.5: 151259768-151259768 on Build GRCh38
rs778041179 Chr.5: 151259849-151259849 on Build GRCh38
rs376211344 Chr.5: 151259747-151259747 on Build GRCh38
rs113271740 Chr.5: 151259799-151259799 on Build GRCh38
rs948618135 Chr.5: 151259556-151259556 on Build GRCh38
rs749309977 Chr.5: 151259906-151259908 on Build GRCh38
rs760365843 Chr.5: 151259783-151259783 on Build GRCh38
rs750956020 Chr.5: 151259726-151259726 on Build GRCh38
rs148610682 Chr.5: 151259941-151259941 on Build GRCh38
rs763502453 Chr.5: 151259899-151259899 on Build GRCh38
rs1023068951 Chr.5: 151259655-151259655 on Build GRCh38
rs761513426 Chr.5: 151259852-151259852 on Build GRCh38
rs878905080 Chr.5: 151259693-151259693 on Build GRCh38
rs893036133 Chr.5: 151259876-151259876 on Build GRCh38
rs774838727 Chr.5: 151259611-151259611 on Build GRCh38
rs745791765 Chr.5: 151259713-151259713 on Build GRCh38

View Full Product Details