Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54650592-54651190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
UniGene
Hs.473133
Gene Chromosome Location
Chr.20: 54475472-54651171 on Build GRCh38
Transcripts
NM_018431.4 NM_177959.2 XM_011528904.1

SNP Detail

SNP ID Chromosome Location
rs982880351 Chr.20: 54650958-54650958 on Build GRCh38
rs984408417 Chr.20: 54650750-54650750 on Build GRCh38
rs569425845 Chr.20: 54651046-54651046 on Build GRCh38
rs866061319 Chr.20: 54651125-54651125 on Build GRCh38
rs949147516 Chr.20: 54650646-54650646 on Build GRCh38
rs984155297 Chr.20: 54651058-54651058 on Build GRCh38
rs1034502638 Chr.20: 54651015-54651015 on Build GRCh38
rs1026469291 Chr.20: 54651121-54651121 on Build GRCh38
rs747944022 Chr.20: 54650697-54650697 on Build GRCh38
rs1052100242 Chr.20: 54650865-54650865 on Build GRCh38
rs747025404 Chr.20: 54650869-54650869 on Build GRCh38
rs917690108 Chr.20: 54650645-54650645 on Build GRCh38
rs371667416 Chr.20: 54650901-54650901 on Build GRCh38
rs2841 Chr.20: 54651054-54651054 on Build GRCh38
rs909800269 Chr.20: 54650771-54650771 on Build GRCh38
rs533982941 Chr.20: 54651122-54651122 on Build GRCh38
rs946406743 Chr.20: 54650909-54650909 on Build GRCh38
rs1053521118 Chr.20: 54651045-54651045 on Build GRCh38
rs1018222638 Chr.20: 54650846-54650846 on Build GRCh38
rs960142531 Chr.20: 54650709-54650709 on Build GRCh38
rs1021886596 Chr.20: 54650683-54650683 on Build GRCh38
rs1001644275 Chr.20: 54650990-54650990 on Build GRCh38
rs777754462 Chr.20: 54650749-54650749 on Build GRCh38
rs114025120 Chr.20: 54650746-54650746 on Build GRCh38
rs936475016 Chr.20: 54650724-54650724 on Build GRCh38
rs1039117079 Chr.20: 54651095-54651095 on Build GRCh38
rs1007639388 Chr.20: 54650824-54650824 on Build GRCh38
rs183194169 Chr.20: 54650832-54650832 on Build GRCh38
rs926451004 Chr.20: 54650661-54650661 on Build GRCh38
rs1001675593 Chr.20: 54650691-54650691 on Build GRCh38
rs895900972 Chr.20: 54651019-54651019 on Build GRCh38
rs1007936960 Chr.20: 54651094-54651094 on Build GRCh38
rs1010939832 Chr.20: 54650673-54650673 on Build GRCh38
rs1016850894 Chr.20: 54651074-54651074 on Build GRCh38
rs764782317 Chr.20: 54651088-54651088 on Build GRCh38
rs150414359 Chr.20: 54650806-54650806 on Build GRCh38
rs904660916 Chr.20: 54650689-54650689 on Build GRCh38
rs950935073 Chr.20: 54650899-54650899 on Build GRCh38
rs771015717 Chr.20: 54650939-54650939 on Build GRCh38
rs995034918 Chr.20: 54651101-54651101 on Build GRCh38
rs994964906 Chr.20: 54650864-54650864 on Build GRCh38
rs761687987 Chr.20: 54651084-54651084 on Build GRCh38
rs1026582334 Chr.20: 54650884-54650884 on Build GRCh38
rs560989605 Chr.20: 54650847-54650847 on Build GRCh38
rs555481837 Chr.20: 54650656-54650656 on Build GRCh38
rs890558795 Chr.20: 54650815-54650815 on Build GRCh38
rs112799548 Chr.20: 54650986-54650986 on Build GRCh38
rs573057467 Chr.20: 54650664-54650664 on Build GRCh38
rs545334495 Chr.20: 54650684-54650684 on Build GRCh38
rs4809970 Chr.20: 54650747-54650747 on Build GRCh38
rs201024891 Chr.20: 54650989-54650989 on Build GRCh38
rs78439370 Chr.20: 54650778-54650778 on Build GRCh38
rs1043678180 Chr.20: 54650919-54650919 on Build GRCh38
rs2842 Chr.20: 54651043-54651043 on Build GRCh38
rs1034368312 Chr.20: 54650699-54650699 on Build GRCh38
rs1014502485 Chr.20: 54651157-54651157 on Build GRCh38
rs950985465 Chr.20: 54651146-54651146 on Build GRCh38
rs15899 Chr.20: 54651088-54651088 on Build GRCh38
rs529772477 Chr.20: 54650928-54650928 on Build GRCh38
rs758320131 Chr.20: 54650748-54650748 on Build GRCh38
rs2843 Chr.20: 54651096-54651096 on Build GRCh38
rs2840 Chr.20: 54651037-54651037 on Build GRCh38
rs776671747 Chr.20: 54651020-54651020 on Build GRCh38

View Full Product Details