Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42206853-42207375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG1
Entrez Gene ID
9619
Gene Name
ATP binding cassette subfamily G member 1
Gene Aliases
ABC8, WHITE1
UniGene
Hs.124649
Gene Chromosome Location
Chr.21: 42199689-42304389 on Build GRCh38
Transcripts
NM_207627.1 NM_207628.1

SNP Detail

SNP ID Chromosome Location
rs748776557 Chr.21: 42207071-42207071 on Build GRCh38
rs113709987 Chr.21: 42207348-42207348 on Build GRCh38
rs201191364 Chr.21: 42206906-42206908 on Build GRCh38
rs1033007809 Chr.21: 42206955-42206955 on Build GRCh38
rs963073887 Chr.21: 42207210-42207210 on Build GRCh38
rs796463048 Chr.21: 42206887-42206887 on Build GRCh38
rs571403143 Chr.21: 42207171-42207171 on Build GRCh38
rs1049580808 Chr.21: 42207141-42207141 on Build GRCh38
rs1036750944 Chr.21: 42207037-42207037 on Build GRCh38
rs13051040 Chr.21: 42206968-42206968 on Build GRCh38
rs13051166 Chr.21: 42207100-42207100 on Build GRCh38
rs980627055 Chr.21: 42207308-42207308 on Build GRCh38
rs577728825 Chr.21: 42206909-42206909 on Build GRCh38
rs927470457 Chr.21: 42207316-42207316 on Build GRCh38
rs556929128 Chr.21: 42206995-42206995 on Build GRCh38
rs13047412 Chr.21: 42207002-42207002 on Build GRCh38
rs1032915920 Chr.21: 42207175-42207175 on Build GRCh38
rs542613655 Chr.21: 42207266-42207266 on Build GRCh38
rs221949 Chr.21: 42207205-42207205 on Build GRCh38
rs777476015 Chr.21: 42207033-42207033 on Build GRCh38
rs1025204623 Chr.21: 42207114-42207114 on Build GRCh38
rs956070184 Chr.21: 42206956-42206956 on Build GRCh38
rs909125231 Chr.21: 42207191-42207191 on Build GRCh38
rs575242759 Chr.21: 42207038-42207038 on Build GRCh38
rs556192303 Chr.21: 42207313-42207313 on Build GRCh38
rs537743070 Chr.21: 42207094-42207094 on Build GRCh38
rs757037541 Chr.21: 42207077-42207077 on Build GRCh38
rs61623715 Chr.21: 42206874-42206874 on Build GRCh38
rs745464540 Chr.21: 42207253-42207253 on Build GRCh38
rs568811093 Chr.21: 42207039-42207039 on Build GRCh38
rs58412126 Chr.21: 42206887-42206888 on Build GRCh38
rs930036121 Chr.21: 42207288-42207288 on Build GRCh38
rs914706838 Chr.21: 42206888-42206888 on Build GRCh38
rs377501232 Chr.21: 42206888-42206888 on Build GRCh38
rs754815504 Chr.21: 42206957-42206957 on Build GRCh38
rs13047540 Chr.21: 42207134-42207134 on Build GRCh38
rs930170384 Chr.21: 42206981-42206981 on Build GRCh38
rs956562390 Chr.21: 42206892-42206892 on Build GRCh38
rs80157302 Chr.21: 42206994-42206994 on Build GRCh38
rs115417708 Chr.21: 42206963-42206963 on Build GRCh38
rs544866944 Chr.21: 42206885-42206885 on Build GRCh38
rs897244204 Chr.21: 42206980-42206980 on Build GRCh38
rs182325611 Chr.21: 42206969-42206969 on Build GRCh38
rs115580707 Chr.21: 42207132-42207132 on Build GRCh38
rs889653825 Chr.21: 42207142-42207142 on Build GRCh38
rs930900526 Chr.21: 42206997-42206997 on Build GRCh38
rs917869477 Chr.21: 42206950-42206950 on Build GRCh38
rs573616458 Chr.21: 42207217-42207217 on Build GRCh38
rs1024305180 Chr.21: 42207228-42207228 on Build GRCh38
rs533721298 Chr.21: 42207197-42207197 on Build GRCh38
rs77603571 Chr.21: 42206991-42206991 on Build GRCh38
rs952334195 Chr.21: 42207340-42207340 on Build GRCh38
rs909528476 Chr.21: 42207008-42207008 on Build GRCh38
rs1000105698 Chr.21: 42207143-42207143 on Build GRCh38
rs13048493 Chr.21: 42207099-42207099 on Build GRCh38
rs1030746881 Chr.21: 42206889-42206889 on Build GRCh38

View Full Product Details