Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219238527-219238993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L
Entrez Gene ID
79411
Gene Name
galactosidase beta 1 like
Gene Aliases
-
UniGene
Hs.181173
Gene Chromosome Location
Chr.2: 219236598-219245429 on Build GRCh38
Transcripts
NM_001286423.1 NM_001286427.1 NM_024506.4 XM_011511813.2 XM_017004895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016654 c.1178G>A p.R393H Chr.2: 219238544-219238544 on Build GRCh38
3972036 c.1174C>T p.P392S Chr.2: 219238548-219238548 on Build GRCh38
3044728 c.1144C>T p.H382Y Chr.2: 219238578-219238578 on Build GRCh38
6466983 c.1120C>T p.P374S Chr.2: 219238722-219238722 on Build GRCh38
4001512 c.1073T>C p.V358A Chr.2: 219238769-219238769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139033988 Chr.2: 219238560-219238560 on Build GRCh38
rs750109380 Chr.2: 219238743-219238743 on Build GRCh38
rs371896647 Chr.2: 219238631-219238631 on Build GRCh38
rs777489339 Chr.2: 219238611-219238611 on Build GRCh38
rs976242830 Chr.2: 219238705-219238705 on Build GRCh38
rs755686523 Chr.2: 219238632-219238632 on Build GRCh38
rs754331961 Chr.2: 219238590-219238591 on Build GRCh38
rs751400969 Chr.2: 219238720-219238720 on Build GRCh38
rs565599084 Chr.2: 219238800-219238800 on Build GRCh38
rs764063773 Chr.2: 219238712-219238712 on Build GRCh38
rs953425075 Chr.2: 219238757-219238757 on Build GRCh38
rs368910955 Chr.2: 219238607-219238607 on Build GRCh38
rs146847433 Chr.2: 219238671-219238689 on Build GRCh38
rs969289899 Chr.2: 219238953-219238953 on Build GRCh38
rs372592862 Chr.2: 219238549-219238549 on Build GRCh38
rs73079103 Chr.2: 219238847-219238847 on Build GRCh38
rs776168293 Chr.2: 219238547-219238547 on Build GRCh38
rs1050736767 Chr.2: 219238691-219238691 on Build GRCh38
rs748563532 Chr.2: 219238789-219238789 on Build GRCh38
rs754419106 Chr.2: 219238670-219238670 on Build GRCh38
rs904972297 Chr.2: 219238853-219238853 on Build GRCh38
rs1029113316 Chr.2: 219238744-219238744 on Build GRCh38
rs147127165 Chr.2: 219238915-219238915 on Build GRCh38
rs762947124 Chr.2: 219238707-219238707 on Build GRCh38
rs892100880 Chr.2: 219238642-219238642 on Build GRCh38
rs547440047 Chr.2: 219238838-219238838 on Build GRCh38
rs752639686 Chr.2: 219238604-219238604 on Build GRCh38
rs760766368 Chr.2: 219238598-219238598 on Build GRCh38
rs564988390 Chr.2: 219238761-219238761 on Build GRCh38
rs760821224 Chr.2: 219238702-219238702 on Build GRCh38
rs779499678 Chr.2: 219238775-219238775 on Build GRCh38
rs779982477 Chr.2: 219238679-219238683 on Build GRCh38
rs756740528 Chr.2: 219238600-219238608 on Build GRCh38
rs774764307 Chr.2: 219238564-219238564 on Build GRCh38
rs935113527 Chr.2: 219238652-219238652 on Build GRCh38
rs766463871 Chr.2: 219238704-219238704 on Build GRCh38
rs1044863378 Chr.2: 219238849-219238849 on Build GRCh38
rs545199045 Chr.2: 219238656-219238656 on Build GRCh38
rs76908853 Chr.2: 219238769-219238769 on Build GRCh38
rs141653050 Chr.2: 219238593-219238593 on Build GRCh38
rs544839674 Chr.2: 219238965-219238965 on Build GRCh38
rs201561729 Chr.2: 219238583-219238583 on Build GRCh38
rs201287356 Chr.2: 219238827-219238827 on Build GRCh38
rs200348120 Chr.2: 219238689-219238689 on Build GRCh38
rs150309380 Chr.2: 219238745-219238745 on Build GRCh38
rs751367803 Chr.2: 219238618-219238618 on Build GRCh38
rs771140471 Chr.2: 219238806-219238806 on Build GRCh38
rs137908162 Chr.2: 219238748-219238748 on Build GRCh38
rs780748547 Chr.2: 219238839-219238839 on Build GRCh38
rs202221491 Chr.2: 219238634-219238634 on Build GRCh38
rs767146478 Chr.2: 219238736-219238736 on Build GRCh38
rs762005816 Chr.2: 219238570-219238570 on Build GRCh38
rs768449041 Chr.2: 219238760-219238760 on Build GRCh38
rs749867884 Chr.2: 219238635-219238635 on Build GRCh38
rs992403658 Chr.2: 219238908-219238908 on Build GRCh38
rs772491778 Chr.2: 219238795-219238795 on Build GRCh38
rs993626551 Chr.2: 219238833-219238833 on Build GRCh38
rs745446517 Chr.2: 219238660-219238661 on Build GRCh38
rs780916120 Chr.2: 219238627-219238627 on Build GRCh38
rs756906872 Chr.2: 219238626-219238626 on Build GRCh38
rs766406496 Chr.2: 219238600-219238600 on Build GRCh38
rs748525589 Chr.2: 219238675-219238675 on Build GRCh38
rs778578574 Chr.2: 219238661-219238661 on Build GRCh38
rs1053916809 Chr.2: 219238922-219238922 on Build GRCh38
rs747293237 Chr.2: 219238801-219238801 on Build GRCh38
rs757104639 Chr.2: 219238725-219238725 on Build GRCh38
rs765968843 Chr.2: 219238584-219238584 on Build GRCh38
rs75234434 Chr.2: 219238555-219238555 on Build GRCh38
rs550287163 Chr.2: 219238930-219238930 on Build GRCh38
rs752671672 Chr.2: 219238852-219238852 on Build GRCh38
rs773394642 Chr.2: 219238796-219238796 on Build GRCh38
rs746872066 Chr.2: 219238745-219238747 on Build GRCh38

View Full Product Details