Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127786522-127786978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK9
Entrez Gene ID
1025
Gene Name
cyclin dependent kinase 9
Gene Aliases
C-2k, CDC2L4, CTK1, PITALRE, TAK
UniGene
Hs.150423
Gene Chromosome Location
Chr.9: 127786026-127790782 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5998495 c.525+6T>C p.? Chr.9: 127786788-127786788 on Build GRCh38
5998494 c.174+6T>C p.? Chr.9: 127786788-127786788 on Build GRCh38
6485853 c.172G>A p.G58R Chr.9: 127786780-127786780 on Build GRCh38
6485854 c.523G>A p.G175R Chr.9: 127786780-127786780 on Build GRCh38
4152073 c.129G>C p.Q43H Chr.9: 127786737-127786737 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372145564 Chr.9: 127786814-127786814 on Build GRCh38
rs755047732 Chr.9: 127786824-127786824 on Build GRCh38
rs1044636695 Chr.9: 127786604-127786604 on Build GRCh38
rs779029217 Chr.9: 127786828-127786828 on Build GRCh38
rs538521254 Chr.9: 127786621-127786621 on Build GRCh38
rs542635366 Chr.9: 127786696-127786696 on Build GRCh38
rs775287060 Chr.9: 127786669-127786669 on Build GRCh38
rs761040673 Chr.9: 127786757-127786757 on Build GRCh38
rs773818931 Chr.9: 127786653-127786653 on Build GRCh38
rs62587186 Chr.9: 127786547-127786547 on Build GRCh38
rs775950469 Chr.9: 127786949-127786949 on Build GRCh38
rs993835778 Chr.9: 127786642-127786642 on Build GRCh38
rs376772241 Chr.9: 127786626-127786626 on Build GRCh38
rs576021592 Chr.9: 127786601-127786601 on Build GRCh38
rs201862836 Chr.9: 127786693-127786693 on Build GRCh38
rs771561004 Chr.9: 127786755-127786755 on Build GRCh38
rs1053172655 Chr.9: 127786585-127786585 on Build GRCh38
rs867815204 Chr.9: 127786641-127786641 on Build GRCh38
rs961901776 Chr.9: 127786546-127786546 on Build GRCh38
rs774013467 Chr.9: 127786782-127786782 on Build GRCh38
rs1007081493 Chr.9: 127786909-127786909 on Build GRCh38
rs1040334979 Chr.9: 127786574-127786574 on Build GRCh38
rs931906921 Chr.9: 127786899-127786899 on Build GRCh38
rs3217734 Chr.9: 127786798-127786798 on Build GRCh38
rs375099649 Chr.9: 127786656-127786656 on Build GRCh38
rs928744740 Chr.9: 127786592-127786592 on Build GRCh38
rs769375453 Chr.9: 127786663-127786663 on Build GRCh38
rs771202984 Chr.9: 127786657-127786657 on Build GRCh38
rs911576995 Chr.9: 127786921-127786921 on Build GRCh38
rs996150464 Chr.9: 127786687-127786687 on Build GRCh38
rs528961894 Chr.9: 127786927-127786927 on Build GRCh38
rs572591176 Chr.9: 127786624-127786624 on Build GRCh38
rs914635422 Chr.9: 127786724-127786724 on Build GRCh38
rs377063441 Chr.9: 127786854-127786854 on Build GRCh38
rs1044221315 Chr.9: 127786796-127786796 on Build GRCh38
rs546674123 Chr.9: 127786545-127786545 on Build GRCh38
rs748495865 Chr.9: 127786716-127786716 on Build GRCh38
rs371774606 Chr.9: 127786650-127786650 on Build GRCh38
rs550439902 Chr.9: 127786857-127786857 on Build GRCh38
rs369043963 Chr.9: 127786911-127786911 on Build GRCh38
rs546438470 Chr.9: 127786889-127786889 on Build GRCh38
rs759922327 Chr.9: 127786891-127786891 on Build GRCh38
rs897081474 Chr.9: 127786844-127786844 on Build GRCh38
rs369152308 Chr.9: 127786810-127786810 on Build GRCh38
rs541621403 Chr.9: 127786634-127786634 on Build GRCh38
rs776932315 Chr.9: 127786777-127786778 on Build GRCh38
rs772437157 Chr.9: 127786725-127786725 on Build GRCh38
rs183267477 Chr.9: 127786954-127786954 on Build GRCh38
rs140701825 Chr.9: 127786576-127786576 on Build GRCh38
rs779167622 Chr.9: 127786715-127786715 on Build GRCh38
rs780038803 Chr.9: 127786699-127786699 on Build GRCh38
rs780602892 Chr.9: 127786713-127786713 on Build GRCh38
rs960085621 Chr.9: 127786813-127786813 on Build GRCh38
rs200589213 Chr.9: 127786674-127786674 on Build GRCh38
rs761535526 Chr.9: 127786790-127786790 on Build GRCh38
rs533145001 Chr.9: 127786868-127786868 on Build GRCh38
rs755386803 Chr.9: 127786838-127786838 on Build GRCh38
rs564175592 Chr.9: 127786862-127786862 on Build GRCh38
rs139206714 Chr.9: 127786779-127786779 on Build GRCh38
rs902229048 Chr.9: 127786642-127786642 on Build GRCh38
rs759142793 Chr.9: 127786657-127786657 on Build GRCh38
rs753912033 Chr.9: 127786815-127786815 on Build GRCh38
rs373517826 Chr.9: 127786553-127786553 on Build GRCh38
rs1036142850 Chr.9: 127786946-127786946 on Build GRCh38
rs767241242 Chr.9: 127786793-127786793 on Build GRCh38
rs538585577 Chr.9: 127786959-127786959 on Build GRCh38
rs368523668 Chr.9: 127786670-127786670 on Build GRCh38
rs753944760 Chr.9: 127786701-127786701 on Build GRCh38
rs149950953 Chr.9: 127786780-127786780 on Build GRCh38
rs980177761 Chr.9: 127786580-127786580 on Build GRCh38
rs905857521 Chr.9: 127786730-127786730 on Build GRCh38
rs979083261 Chr.9: 127786877-127786877 on Build GRCh38
rs1020521376 Chr.9: 127786817-127786817 on Build GRCh38
rs943636724 Chr.9: 127786572-127786572 on Build GRCh38
rs778441690 Chr.9: 127786731-127786731 on Build GRCh38
rs760668207 Chr.9: 127786812-127786812 on Build GRCh38
rs558093842 Chr.9: 127786622-127786622 on Build GRCh38
rs993804631 Chr.9: 127786858-127786858 on Build GRCh38
rs747453571 Chr.9: 127786733-127786733 on Build GRCh38
rs151087657 Chr.9: 127786931-127786931 on Build GRCh38
rs200127201 Chr.9: 127786678-127786678 on Build GRCh38
rs756194891 Chr.9: 127786697-127786697 on Build GRCh38
rs988782815 Chr.9: 127786717-127786717 on Build GRCh38

View Full Product Details