Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27153594-27154161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA-AS3
Entrez Gene ID
100133311
Gene Name
HOXA cluster antisense RNA 3
Gene Aliases
HOXA6as
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27140364-27155928 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

Gene Symbol
HOXA7
Entrez Gene ID
3204
Gene Name
homeobox A7
Gene Aliases
ANTP, HOX1, HOX1.1, HOX1A
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27153716-27156677 on Build GRCh38
Transcripts
NR_038831.1 NR_038832.1 NM_006896.3

SNP Detail

SNP ID Chromosome Location
rs373259624 Chr.7: 27153846-27153846 on Build GRCh38
rs981090569 Chr.7: 27153939-27153939 on Build GRCh38
rs937294170 Chr.7: 27153784-27153784 on Build GRCh38
rs1041051144 Chr.7: 27154066-27154066 on Build GRCh38
rs966873932 Chr.7: 27153620-27153620 on Build GRCh38
rs1018546431 Chr.7: 27153981-27153981 on Build GRCh38
rs1029869054 Chr.7: 27153961-27153961 on Build GRCh38
rs181591017 Chr.7: 27153956-27153956 on Build GRCh38
rs751726644 Chr.7: 27153972-27153972 on Build GRCh38
rs946719766 Chr.7: 27154062-27154062 on Build GRCh38
rs999684286 Chr.7: 27153905-27153905 on Build GRCh38
rs941028871 Chr.7: 27153705-27153705 on Build GRCh38
rs757398915 Chr.7: 27154023-27154023 on Build GRCh38
rs899866326 Chr.7: 27154078-27154078 on Build GRCh38
rs41304930 Chr.7: 27153932-27153932 on Build GRCh38
rs554291478 Chr.7: 27153747-27153747 on Build GRCh38
rs1051186332 Chr.7: 27154091-27154091 on Build GRCh38
rs540758394 Chr.7: 27154074-27154074 on Build GRCh38
rs1057512 Chr.7: 27153984-27153984 on Build GRCh38
rs534304376 Chr.7: 27153733-27153733 on Build GRCh38
rs920135972 Chr.7: 27153661-27153661 on Build GRCh38
rs552711590 Chr.7: 27154009-27154009 on Build GRCh38
rs931503542 Chr.7: 27154011-27154011 on Build GRCh38
rs116895484 Chr.7: 27153697-27153697 on Build GRCh38
rs1040288225 Chr.7: 27153761-27153761 on Build GRCh38
rs1022355504 Chr.7: 27153847-27153847 on Build GRCh38
rs1011701878 Chr.7: 27153873-27153873 on Build GRCh38
rs1011420691 Chr.7: 27154110-27154110 on Build GRCh38
rs151012022 Chr.7: 27154036-27154036 on Build GRCh38
rs185652202 Chr.7: 27153630-27153630 on Build GRCh38
rs772760261 Chr.7: 27153876-27153876 on Build GRCh38
rs536241937 Chr.7: 27153835-27153835 on Build GRCh38
rs560644548 Chr.7: 27154080-27154080 on Build GRCh38
rs891267719 Chr.7: 27154093-27154093 on Build GRCh38
rs1047558466 Chr.7: 27154016-27154016 on Build GRCh38
rs996089984 Chr.7: 27154085-27154085 on Build GRCh38
rs556482394 Chr.7: 27153935-27153935 on Build GRCh38
rs375261420 Chr.7: 27153648-27153651 on Build GRCh38
rs957264309 Chr.7: 27153861-27153861 on Build GRCh38
rs118016485 Chr.7: 27154113-27154113 on Build GRCh38
rs974166802 Chr.7: 27153982-27153982 on Build GRCh38
rs187649677 Chr.7: 27154027-27154027 on Build GRCh38
rs1055870758 Chr.7: 27153646-27153646 on Build GRCh38
rs920087113 Chr.7: 27153989-27153989 on Build GRCh38
rs1050365389 Chr.7: 27154090-27154090 on Build GRCh38
rs1008241500 Chr.7: 27153746-27153746 on Build GRCh38
rs774706408 Chr.7: 27153653-27153653 on Build GRCh38
rs958934190 Chr.7: 27153980-27153980 on Build GRCh38
rs17500925 Chr.7: 27153981-27153981 on Build GRCh38
rs764616299 Chr.7: 27153810-27153810 on Build GRCh38
rs190750155 Chr.7: 27153794-27153794 on Build GRCh38
rs955604338 Chr.7: 27153979-27153979 on Build GRCh38
rs17500918 Chr.7: 27153859-27153859 on Build GRCh38
rs1047185036 Chr.7: 27153732-27153732 on Build GRCh38
rs895112554 Chr.7: 27153803-27153803 on Build GRCh38
rs550176855 Chr.7: 27153980-27153981 on Build GRCh38
rs1020429771 Chr.7: 27154111-27154111 on Build GRCh38
rs929744209 Chr.7: 27153990-27153990 on Build GRCh38
rs985114571 Chr.7: 27153660-27153660 on Build GRCh38
rs902618746 Chr.7: 27153852-27153852 on Build GRCh38
rs966986612 Chr.7: 27153889-27153889 on Build GRCh38
rs928903524 Chr.7: 27153625-27153625 on Build GRCh38
rs777765835 Chr.7: 27153839-27153839 on Build GRCh38
rs775860691 Chr.7: 27153767-27153767 on Build GRCh38
rs529731632 Chr.7: 27154095-27154095 on Build GRCh38
rs556629939 Chr.7: 27154098-27154098 on Build GRCh38
rs918346625 Chr.7: 27153718-27153721 on Build GRCh38
rs973816400 Chr.7: 27153715-27153715 on Build GRCh38
rs764389620 Chr.7: 27153879-27153879 on Build GRCh38
rs75300473 Chr.7: 27153662-27153662 on Build GRCh38

View Full Product Details