Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32334283-32334625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf75
Entrez Gene ID
64149
Gene Name
chromosome 17 open reading frame 75
Gene Aliases
NJMU-R1, SRI2
UniGene
Hs.655257
Gene Chromosome Location
Chr.17: 32328441-32350047 on Build GRCh38
Transcripts
NM_022344.3 XM_005258022.3 XM_017024940.1 XM_017024941.1

SNP Detail

SNP ID Chromosome Location
rs767243043 Chr.17: 32334570-32334570 on Build GRCh38
rs779766504 Chr.17: 32334592-32334592 on Build GRCh38
rs1034318460 Chr.17: 32334495-32334495 on Build GRCh38
rs887049211 Chr.17: 32334545-32334545 on Build GRCh38
rs570158260 Chr.17: 32334347-32334347 on Build GRCh38
rs778826076 Chr.17: 32334433-32334433 on Build GRCh38
rs190434891 Chr.17: 32334503-32334503 on Build GRCh38
rs764395826 Chr.17: 32334543-32334543 on Build GRCh38
rs761554687 Chr.17: 32334462-32334462 on Build GRCh38
rs535535364 Chr.17: 32334369-32334369 on Build GRCh38
rs371405177 Chr.17: 32334436-32334436 on Build GRCh38
rs1000211226 Chr.17: 32334483-32334483 on Build GRCh38
rs758683474 Chr.17: 32334428-32334428 on Build GRCh38
rs1127742 Chr.17: 32334591-32334591 on Build GRCh38
rs778502688 Chr.17: 32334604-32334604 on Build GRCh38
rs774065743 Chr.17: 32334458-32334458 on Build GRCh38
rs758555466 Chr.17: 32334530-32334530 on Build GRCh38
rs368181633 Chr.17: 32334505-32334505 on Build GRCh38
rs987042932 Chr.17: 32334403-32334403 on Build GRCh38
rs768262036 Chr.17: 32334457-32334457 on Build GRCh38
rs930961705 Chr.17: 32334398-32334398 on Build GRCh38
rs558393012 Chr.17: 32334492-32334492 on Build GRCh38
rs752776532 Chr.17: 32334528-32334528 on Build GRCh38
rs182533138 Chr.17: 32334548-32334548 on Build GRCh38
rs904750368 Chr.17: 32334478-32334478 on Build GRCh38
rs766693751 Chr.17: 32334500-32334500 on Build GRCh38
rs749239338 Chr.17: 32334596-32334596 on Build GRCh38
rs773392379 Chr.17: 32334482-32334482 on Build GRCh38
rs1010936466 Chr.17: 32334456-32334456 on Build GRCh38
rs907196978 Chr.17: 32334488-32334488 on Build GRCh38
rs185717321 Chr.17: 32334465-32334465 on Build GRCh38
rs1016666650 Chr.17: 32334319-32334319 on Build GRCh38
rs548602501 Chr.17: 32334399-32334399 on Build GRCh38
rs765295011 Chr.17: 32334506-32334506 on Build GRCh38
rs769565360 Chr.17: 32334446-32334446 on Build GRCh38
rs942418461 Chr.17: 32334429-32334429 on Build GRCh38
rs1017151874 Chr.17: 32334562-32334562 on Build GRCh38
rs749227675 Chr.17: 32334454-32334454 on Build GRCh38
rs975196083 Chr.17: 32334331-32334331 on Build GRCh38
rs773503250 Chr.17: 32334427-32334427 on Build GRCh38
rs924884030 Chr.17: 32334417-32334417 on Build GRCh38
rs182529057 Chr.17: 32334307-32334307 on Build GRCh38
rs151127049 Chr.17: 32334447-32334447 on Build GRCh38
rs1056298082 Chr.17: 32334527-32334527 on Build GRCh38
rs550028075 Chr.17: 32334326-32334326 on Build GRCh38
rs918276180 Chr.17: 32334393-32334393 on Build GRCh38
rs147109840 Chr.17: 32334400-32334400 on Build GRCh38
rs565301058 Chr.17: 32334512-32334512 on Build GRCh38

View Full Product Details