Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 65960580-65961128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSB
Entrez Gene ID
2990
Gene Name
glucuronidase beta
Gene Aliases
BG, MPS7
UniGene
Hs.255230
Gene Chromosome Location
Chr.7: 65960684-65982314 on Build GRCh38
Transcripts
NM_000181.3 NM_001284290.1 NM_001293104.1 NM_001293105.1 NR_120531.1 XM_005250297.3 XM_011516113.1 XM_011516114.1 XM_017012091.1 XM_017012092.1 XM_017012093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1635371 c.1892A>C p.N631T Chr.7: 65960961-65960961 on Build GRCh38
6110761 c.1806G>C p.L602L Chr.7: 65961047-65961047 on Build GRCh38
6859061 c.1790-5delT p.? Chr.7: 65961068-65961068 on Build GRCh38
1091311 c.1894G>T p.E632* Chr.7: 65960959-65960959 on Build GRCh38
6856602 c.1790-3_1790-2insT p.? Chr.7: 65961065-65961066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777613366 Chr.7: 65960977-65960977 on Build GRCh38
rs1023997983 Chr.7: 65960801-65960801 on Build GRCh38
rs1003656203 Chr.7: 65960821-65960821 on Build GRCh38
rs908000956 Chr.7: 65960603-65960603 on Build GRCh38
rs953646431 Chr.7: 65961088-65961088 on Build GRCh38
rs35302712 Chr.7: 65961008-65961008 on Build GRCh38
rs780999350 Chr.7: 65960961-65960961 on Build GRCh38
rs751075889 Chr.7: 65960952-65960952 on Build GRCh38
rs772478693 Chr.7: 65960908-65960908 on Build GRCh38
rs893612668 Chr.7: 65960767-65960767 on Build GRCh38
rs755492085 Chr.7: 65961096-65961096 on Build GRCh38
rs769514976 Chr.7: 65960885-65960885 on Build GRCh38
rs750022485 Chr.7: 65961068-65961068 on Build GRCh38
rs992206868 Chr.7: 65960610-65960610 on Build GRCh38
rs371981054 Chr.7: 65961067-65961067 on Build GRCh38
rs751840849 Chr.7: 65960986-65960986 on Build GRCh38
rs140236850 Chr.7: 65960736-65960736 on Build GRCh38
rs779485917 Chr.7: 65960872-65960872 on Build GRCh38
rs1011311679 Chr.7: 65960772-65960772 on Build GRCh38
rs922193838 Chr.7: 65960658-65960658 on Build GRCh38
rs557221338 Chr.7: 65960901-65960901 on Build GRCh38
rs753655611 Chr.7: 65961086-65961086 on Build GRCh38
rs145910162 Chr.7: 65960947-65960947 on Build GRCh38
rs766019399 Chr.7: 65960951-65960951 on Build GRCh38
rs113552467 Chr.7: 65960659-65960659 on Build GRCh38
rs765983374 Chr.7: 65961057-65961057 on Build GRCh38
rs752576315 Chr.7: 65960962-65960962 on Build GRCh38
rs754474825 Chr.7: 65960955-65960955 on Build GRCh38
rs903992911 Chr.7: 65960782-65960782 on Build GRCh38
rs756497692 Chr.7: 65960851-65960851 on Build GRCh38
rs779147312 Chr.7: 65960996-65960996 on Build GRCh38
rs767165241 Chr.7: 65961068-65961069 on Build GRCh38
rs536903750 Chr.7: 65960803-65960803 on Build GRCh38
rs567259021 Chr.7: 65960810-65960810 on Build GRCh38
rs752200387 Chr.7: 65961068-65961068 on Build GRCh38
rs951115772 Chr.7: 65960850-65960850 on Build GRCh38
rs751073317 Chr.7: 65960874-65960874 on Build GRCh38
rs960705802 Chr.7: 65960809-65960809 on Build GRCh38
rs374209414 Chr.7: 65961092-65961092 on Build GRCh38
rs756087154 Chr.7: 65961074-65961074 on Build GRCh38
rs138519677 Chr.7: 65960703-65960703 on Build GRCh38
rs763997663 Chr.7: 65961081-65961081 on Build GRCh38
rs372550726 Chr.7: 65960979-65960979 on Build GRCh38
rs767115386 Chr.7: 65961062-65961062 on Build GRCh38
rs754908402 Chr.7: 65960876-65960876 on Build GRCh38
rs548050859 Chr.7: 65960661-65960661 on Build GRCh38
rs942275302 Chr.7: 65960634-65960634 on Build GRCh38
rs757993425 Chr.7: 65960860-65960860 on Build GRCh38
rs201204553 Chr.7: 65960893-65960893 on Build GRCh38
rs121918172 Chr.7: 65960997-65960997 on Build GRCh38
rs753596986 Chr.7: 65960828-65960828 on Build GRCh38
rs764493374 Chr.7: 65960924-65960924 on Build GRCh38
rs1043928320 Chr.7: 65960781-65960781 on Build GRCh38
rs775873162 Chr.7: 65961003-65961003 on Build GRCh38
rs762757920 Chr.7: 65960946-65960946 on Build GRCh38
rs879010457 Chr.7: 65961067-65961067 on Build GRCh38
rs1010682367 Chr.7: 65960796-65960796 on Build GRCh38
rs761319717 Chr.7: 65961008-65961008 on Build GRCh38
rs935464108 Chr.7: 65960978-65960979 on Build GRCh38
rs960968321 Chr.7: 65960606-65960606 on Build GRCh38
rs552724524 Chr.7: 65961056-65961056 on Build GRCh38
rs776024156 Chr.7: 65960911-65960911 on Build GRCh38
rs62470933 Chr.7: 65960863-65960863 on Build GRCh38
rs774336516 Chr.7: 65960906-65960906 on Build GRCh38
rs150311243 Chr.7: 65960877-65960877 on Build GRCh38
rs756035101 Chr.7: 65960966-65960966 on Build GRCh38
rs769662185 Chr.7: 65960878-65960879 on Build GRCh38
rs121918184 Chr.7: 65960972-65960972 on Build GRCh38
rs190201470 Chr.7: 65960689-65960689 on Build GRCh38
rs909063611 Chr.7: 65960861-65960861 on Build GRCh38
rs1051767192 Chr.7: 65960666-65960666 on Build GRCh38
rs77293332 Chr.7: 65960725-65960725 on Build GRCh38
rs372007777 Chr.7: 65960911-65960912 on Build GRCh38
rs773791166 Chr.7: 65961065-65961066 on Build GRCh38
rs777173084 Chr.7: 65961031-65961031 on Build GRCh38
rs758984878 Chr.7: 65961060-65961060 on Build GRCh38
rs764071830 Chr.7: 65961064-65961065 on Build GRCh38
rs779006886 Chr.7: 65961099-65961099 on Build GRCh38
rs370579504 Chr.7: 65960629-65960630 on Build GRCh38
rs973621021 Chr.7: 65960637-65960637 on Build GRCh38
rs747928978 Chr.7: 65960884-65960884 on Build GRCh38
rs121918176 Chr.7: 65961022-65961022 on Build GRCh38
rs754238750 Chr.7: 65960928-65960928 on Build GRCh38
rs772122474 Chr.7: 65961000-65961000 on Build GRCh38
rs756495234 Chr.7: 65961067-65961068 on Build GRCh38
rs762241590 Chr.7: 65961051-65961051 on Build GRCh38
rs762261099 Chr.7: 65960725-65960725 on Build GRCh38
rs1023590215 Chr.7: 65960818-65960818 on Build GRCh38
rs762840002 Chr.7: 65960948-65960948 on Build GRCh38
rs765012426 Chr.7: 65960856-65960856 on Build GRCh38
rs751161248 Chr.7: 65961059-65961059 on Build GRCh38
rs754204979 Chr.7: 65961068-65961072 on Build GRCh38
rs770237165 Chr.7: 65960934-65960935 on Build GRCh38
rs568655640 Chr.7: 65960749-65960749 on Build GRCh38
rs532147376 Chr.7: 65960662-65960662 on Build GRCh38
rs9530 Chr.7: 65960907-65960907 on Build GRCh38
rs753051726 Chr.7: 65960847-65960847 on Build GRCh38
rs372605666 Chr.7: 65960842-65960842 on Build GRCh38
rs771138602 Chr.7: 65960899-65960899 on Build GRCh38

View Full Product Details