Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134148347-134148939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAM3
Entrez Gene ID
83700
Gene Name
junctional adhesion molecule 3
Gene Aliases
JAM-2, JAM-3, JAM-C, JAMC
UniGene
Hs.150718
Gene Chromosome Location
Chr.11: 134068925-134151757 on Build GRCh38
Transcripts
NM_001205329.1 NM_032801.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925144 c.900A>G p.V300V Chr.11: 134148599-134148599 on Build GRCh38
3967422 c.1026C>T p.D342D Chr.11: 134148812-134148812 on Build GRCh38
6315414 c.895G>T p.A299S Chr.11: 134148594-134148594 on Build GRCh38
7136243 c.938G>A p.R313K Chr.11: 134148637-134148637 on Build GRCh38
3445493 c.866G>A p.G289E Chr.11: 134148565-134148565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778582774 Chr.11: 134148527-134148527 on Build GRCh38
rs375062995 Chr.11: 134148724-134148724 on Build GRCh38
rs772533647 Chr.11: 134148640-134148640 on Build GRCh38
rs937867019 Chr.11: 134148918-134148918 on Build GRCh38
rs142616737 Chr.11: 134148753-134148753 on Build GRCh38
rs889654202 Chr.11: 134148908-134148908 on Build GRCh38
rs761535214 Chr.11: 134148585-134148585 on Build GRCh38
rs745941185 Chr.11: 134148719-134148719 on Build GRCh38
rs773938871 Chr.11: 134148759-134148759 on Build GRCh38
rs758416723 Chr.11: 134148723-134148723 on Build GRCh38
rs539233593 Chr.11: 134148853-134148853 on Build GRCh38
rs1032560374 Chr.11: 134148483-134148483 on Build GRCh38
rs760188908 Chr.11: 134148623-134148625 on Build GRCh38
rs199852260 Chr.11: 134148530-134148530 on Build GRCh38
rs768837349 Chr.11: 134148652-134148652 on Build GRCh38
rs754945069 Chr.11: 134148690-134148690 on Build GRCh38
rs759125261 Chr.11: 134148817-134148817 on Build GRCh38
rs779947297 Chr.11: 134148540-134148540 on Build GRCh38
rs773321334 Chr.11: 134148503-134148503 on Build GRCh38
rs781052938 Chr.11: 134148617-134148617 on Build GRCh38
rs780188202 Chr.11: 134148731-134148731 on Build GRCh38
rs766900201 Chr.11: 134148597-134148598 on Build GRCh38
rs568101956 Chr.11: 134148754-134148754 on Build GRCh38
rs776037736 Chr.11: 134148647-134148647 on Build GRCh38
rs1013543995 Chr.11: 134148394-134148394 on Build GRCh38
rs908976926 Chr.11: 134148381-134148381 on Build GRCh38
rs763700386 Chr.11: 134148517-134148517 on Build GRCh38
rs769917401 Chr.11: 134148503-134148506 on Build GRCh38
rs1024944538 Chr.11: 134148739-134148739 on Build GRCh38
rs550090586 Chr.11: 134148650-134148650 on Build GRCh38
rs763250381 Chr.11: 134148572-134148573 on Build GRCh38
rs764804184 Chr.11: 134148594-134148594 on Build GRCh38
rs187264208 Chr.11: 134148390-134148390 on Build GRCh38
rs756778310 Chr.11: 134148526-134148526 on Build GRCh38
rs1034747170 Chr.11: 134148407-134148407 on Build GRCh38
rs771169059 Chr.11: 134148630-134148630 on Build GRCh38
rs200909561 Chr.11: 134148756-134148756 on Build GRCh38
rs755967947 Chr.11: 134148773-134148773 on Build GRCh38
rs544142279 Chr.11: 134148451-134148451 on Build GRCh38
rs145370974 Chr.11: 134148819-134148819 on Build GRCh38
rs763891551 Chr.11: 134148777-134148777 on Build GRCh38
rs752559365 Chr.11: 134148693-134148693 on Build GRCh38
rs777922773 Chr.11: 134148553-134148553 on Build GRCh38
rs573885020 Chr.11: 134148393-134148393 on Build GRCh38
rs769568279 Chr.11: 134148751-134148751 on Build GRCh38
rs779145565 Chr.11: 134148712-134148712 on Build GRCh38
rs772820653 Chr.11: 134148672-134148672 on Build GRCh38
rs371851115 Chr.11: 134148757-134148757 on Build GRCh38
rs761327267 Chr.11: 134148663-134148663 on Build GRCh38
rs939936706 Chr.11: 134148857-134148857 on Build GRCh38
rs958764347 Chr.11: 134148411-134148411 on Build GRCh38
rs770027732 Chr.11: 134148551-134148551 on Build GRCh38
rs774418995 Chr.11: 134148564-134148564 on Build GRCh38
rs372726462 Chr.11: 134148686-134148686 on Build GRCh38
rs770857042 Chr.11: 134148563-134148563 on Build GRCh38
rs977595486 Chr.11: 134148499-134148499 on Build GRCh38
rs610382 Chr.11: 134148761-134148761 on Build GRCh38
rs758019736 Chr.11: 134148826-134148826 on Build GRCh38
rs550482165 Chr.11: 134148805-134148805 on Build GRCh38
rs760983670 Chr.11: 134148578-134148578 on Build GRCh38
rs775553888 Chr.11: 134148828-134148828 on Build GRCh38
rs751943176 Chr.11: 134148614-134148616 on Build GRCh38
rs201187917 Chr.11: 134148574-134148574 on Build GRCh38
rs753300699 Chr.11: 134148726-134148728 on Build GRCh38
rs146181188 Chr.11: 134148559-134148559 on Build GRCh38
rs781436965 Chr.11: 134148895-134148895 on Build GRCh38
rs779948713 Chr.11: 134148787-134148787 on Build GRCh38
rs150992119 Chr.11: 134148812-134148812 on Build GRCh38
rs997541897 Chr.11: 134148743-134148743 on Build GRCh38
rs374187365 Chr.11: 134148579-134148579 on Build GRCh38
rs772586545 Chr.11: 134148571-134148571 on Build GRCh38
rs610829 Chr.11: 134148850-134148850 on Build GRCh38
rs150029724 Chr.11: 134148810-134148810 on Build GRCh38
rs753387122 Chr.11: 134148521-134148521 on Build GRCh38
rs1038157586 Chr.11: 134148651-134148651 on Build GRCh38
rs765181788 Chr.11: 134148754-134148756 on Build GRCh38
rs752401690 Chr.11: 134148767-134148767 on Build GRCh38
rs762621149 Chr.11: 134148682-134148682 on Build GRCh38
rs749976280 Chr.11: 134148596-134148596 on Build GRCh38
rs917528291 Chr.11: 134148495-134148495 on Build GRCh38
rs139455299 Chr.11: 134148747-134148747 on Build GRCh38
rs760612817 Chr.11: 134148830-134148830 on Build GRCh38
rs183001343 Chr.11: 134148878-134148878 on Build GRCh38
rs139421684 Chr.11: 134148804-134148804 on Build GRCh38
rs369063893 Chr.11: 134148785-134148785 on Build GRCh38
rs753333888 Chr.11: 134148439-134148439 on Build GRCh38
rs966017130 Chr.11: 134148528-134148528 on Build GRCh38
rs746796674 Chr.11: 134148865-134148867 on Build GRCh38
rs963437173 Chr.11: 134148386-134148386 on Build GRCh38
rs532937108 Chr.11: 134148746-134148746 on Build GRCh38
rs765035029 Chr.11: 134148664-134148664 on Build GRCh38
rs779861276 Chr.11: 134148855-134148856 on Build GRCh38
rs1042724834 Chr.11: 134148619-134148619 on Build GRCh38
rs894445488 Chr.11: 134148701-134148701 on Build GRCh38
rs773448111 Chr.11: 134148508-134148509 on Build GRCh38
rs748589121 Chr.11: 134148749-134148749 on Build GRCh38
rs898306543 Chr.11: 134148917-134148917 on Build GRCh38
rs751589803 Chr.11: 134148868-134148868 on Build GRCh38
rs933683740 Chr.11: 134148575-134148575 on Build GRCh38
rs746851076 Chr.11: 134148541-134148541 on Build GRCh38
rs767538532 Chr.11: 134148766-134148766 on Build GRCh38
rs754681546 Chr.11: 134148488-134148488 on Build GRCh38
rs1042304520 Chr.11: 134148399-134148399 on Build GRCh38
rs767172958 Chr.11: 134148691-134148691 on Build GRCh38
rs146383325 Chr.11: 134148797-134148797 on Build GRCh38
rs758388533 Chr.11: 134148597-134148597 on Build GRCh38
rs201770276 Chr.11: 134148605-134148605 on Build GRCh38
rs759991628 Chr.11: 134148566-134148566 on Build GRCh38
rs753087518 Chr.11: 134148512-134148512 on Build GRCh38
rs745993203 Chr.11: 134148635-134148635 on Build GRCh38
rs143992931 Chr.11: 134148659-134148659 on Build GRCh38
rs575958709 Chr.11: 134148503-134148504 on Build GRCh38
rs760323072 Chr.11: 134148513-134148513 on Build GRCh38
rs778912033 Chr.11: 134148632-134148632 on Build GRCh38
rs751392634 Chr.11: 134148688-134148688 on Build GRCh38
rs949126262 Chr.11: 134148813-134148815 on Build GRCh38
rs1000690502 Chr.11: 134148453-134148453 on Build GRCh38
rs555979256 Chr.11: 134148508-134148508 on Build GRCh38
rs764829016 Chr.11: 134148795-134148797 on Build GRCh38
rs202004850 Chr.11: 134148855-134148855 on Build GRCh38
rs201643656 Chr.11: 134148509-134148509 on Build GRCh38
rs754785863 Chr.11: 134148613-134148613 on Build GRCh38
rs758441523 Chr.11: 134148536-134148536 on Build GRCh38
rs761662253 Chr.11: 134148665-134148665 on Build GRCh38
rs761896383 Chr.11: 134148374-134148374 on Build GRCh38
rs139043096 Chr.11: 134148607-134148607 on Build GRCh38
rs768122983 Chr.11: 134148725-134148725 on Build GRCh38
rs1049314876 Chr.11: 134148375-134148375 on Build GRCh38
rs750040081 Chr.11: 134148810-134148811 on Build GRCh38
rs753845978 Chr.11: 134148439-134148439 on Build GRCh38
rs747435076 Chr.11: 134148732-134148732 on Build GRCh38
rs748905480 Chr.11: 134148813-134148813 on Build GRCh38
rs775806924 Chr.11: 134148576-134148576 on Build GRCh38
rs368709127 Chr.11: 134148626-134148626 on Build GRCh38
rs553013113 Chr.11: 134148823-134148823 on Build GRCh38
rs967500313 Chr.11: 134148498-134148498 on Build GRCh38
rs1001165065 Chr.11: 134148643-134148643 on Build GRCh38
rs750578435 Chr.11: 134148784-134148784 on Build GRCh38
rs757448452 Chr.11: 134148705-134148705 on Build GRCh38
rs926144804 Chr.11: 134148506-134148506 on Build GRCh38
rs1038280973 Chr.11: 134148891-134148891 on Build GRCh38
rs917627662 Chr.11: 134148609-134148609 on Build GRCh38
rs376974612 Chr.11: 134148815-134148815 on Build GRCh38

View Full Product Details