Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190055536-190056005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf88
Entrez Gene ID
84281
Gene Name
chromosome 2 open reading frame 88
Gene Aliases
smAKAP
UniGene
Hs.389311
Gene Chromosome Location
Chr.2: 189879479-190203484 on Build GRCh38
Transcripts
XM_005246905.1 XM_011511982.1 XM_011511986.2 NM_005259.2

Gene Symbol
MSTN
Entrez Gene ID
2660
Gene Name
myostatin
Gene Aliases
GDF8, MSLHP
UniGene
Hs.41565
Gene Chromosome Location
Chr.2: 190055700-190062729 on Build GRCh38
Transcripts
XM_005246905.1 XM_011511982.1 XM_011511986.2 NM_005259.2

SNP Detail

SNP ID Chromosome Location
rs181396471 Chr.2: 190055653-190055653 on Build GRCh38
rs893500966 Chr.2: 190055774-190055774 on Build GRCh38
rs953442537 Chr.2: 190055637-190055637 on Build GRCh38
rs144167726 Chr.2: 190055808-190055808 on Build GRCh38
rs1033766515 Chr.2: 190055597-190055597 on Build GRCh38
rs752240875 Chr.2: 190055767-190055767 on Build GRCh38
rs902712256 Chr.2: 190055902-190055902 on Build GRCh38
rs906442543 Chr.2: 190055830-190055830 on Build GRCh38
rs370998672 Chr.2: 190055903-190055903 on Build GRCh38
rs781576390 Chr.2: 190055867-190055867 on Build GRCh38
rs550928430 Chr.2: 190055594-190055594 on Build GRCh38
rs1021053891 Chr.2: 190055619-190055619 on Build GRCh38
rs12105165 Chr.2: 190055744-190055744 on Build GRCh38
rs1046346416 Chr.2: 190055821-190055821 on Build GRCh38
rs911049509 Chr.2: 190055671-190055671 on Build GRCh38
rs1055015744 Chr.2: 190055857-190055857 on Build GRCh38
rs908399248 Chr.2: 190055785-190055785 on Build GRCh38
rs537925465 Chr.2: 190055588-190055588 on Build GRCh38
rs965111337 Chr.2: 190055681-190055681 on Build GRCh38
rs975444933 Chr.2: 190055728-190055728 on Build GRCh38
rs893908868 Chr.2: 190055858-190055858 on Build GRCh38
rs938031175 Chr.2: 190055844-190055844 on Build GRCh38
rs983973977 Chr.2: 190055784-190055784 on Build GRCh38
rs1011039186 Chr.2: 190055601-190055601 on Build GRCh38
rs1019607408 Chr.2: 190055680-190055680 on Build GRCh38
rs975088588 Chr.2: 190055965-190055965 on Build GRCh38
rs906283449 Chr.2: 190055814-190055814 on Build GRCh38
rs377503267 Chr.2: 190055896-190055896 on Build GRCh38
rs1009477368 Chr.2: 190055904-190055904 on Build GRCh38
rs987766641 Chr.2: 190055670-190055670 on Build GRCh38
rs965294989 Chr.2: 190055944-190055944 on Build GRCh38
rs906563355 Chr.2: 190055580-190055580 on Build GRCh38
rs920633210 Chr.2: 190055615-190055615 on Build GRCh38
rs923832212 Chr.2: 190055710-190055710 on Build GRCh38
rs3791781 Chr.2: 190055660-190055660 on Build GRCh38
rs1015551459 Chr.2: 190055590-190055590 on Build GRCh38
rs935276590 Chr.2: 190055730-190055730 on Build GRCh38
rs898056802 Chr.2: 190055912-190055912 on Build GRCh38
rs189939536 Chr.2: 190055600-190055600 on Build GRCh38
rs3791782 Chr.2: 190055926-190055926 on Build GRCh38

View Full Product Details