Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 101868580-101869076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
UniGene
Hs.508616
Gene Chromosome Location
Chr.13: 101720855-102402428 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321937.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813341 c.381G>A p.M127I Chr.13: 101868752-101868752 on Build GRCh38
6663505 c.356T>A p.V119D Chr.13: 101868792-101868792 on Build GRCh38
369889 c.363C>A p.I121I Chr.13: 101868785-101868785 on Build GRCh38
5342270 c.392G>T p.G131V Chr.13: 101868741-101868741 on Build GRCh38
3813340 c.396G>A p.M132I Chr.13: 101868752-101868752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145714821 Chr.13: 101868965-101868965 on Build GRCh38
rs756734702 Chr.13: 101868937-101868937 on Build GRCh38
rs768508227 Chr.13: 101868699-101868699 on Build GRCh38
rs765928609 Chr.13: 101868842-101868842 on Build GRCh38
rs571128663 Chr.13: 101868677-101868677 on Build GRCh38
rs757596950 Chr.13: 101868752-101868752 on Build GRCh38
rs753360142 Chr.13: 101868967-101868967 on Build GRCh38
rs542910520 Chr.13: 101869007-101869007 on Build GRCh38
rs545603436 Chr.13: 101868927-101868927 on Build GRCh38
rs777310638 Chr.13: 101868952-101868952 on Build GRCh38
rs913833788 Chr.13: 101868671-101868671 on Build GRCh38
rs549573953 Chr.13: 101868665-101868665 on Build GRCh38
rs547320003 Chr.13: 101868693-101868693 on Build GRCh38
rs764478111 Chr.13: 101868854-101868854 on Build GRCh38
rs7992613 Chr.13: 101868958-101868958 on Build GRCh38
rs772880836 Chr.13: 101868826-101868826 on Build GRCh38
rs763584306 Chr.13: 101868850-101868850 on Build GRCh38
rs200882097 Chr.13: 101868899-101868902 on Build GRCh38
rs756297243 Chr.13: 101868775-101868775 on Build GRCh38
rs752022905 Chr.13: 101868859-101868859 on Build GRCh38
rs746040831 Chr.13: 101868764-101868764 on Build GRCh38
rs753208217 Chr.13: 101868750-101868750 on Build GRCh38
rs538352361 Chr.13: 101868899-101868899 on Build GRCh38
rs767124603 Chr.13: 101868704-101868704 on Build GRCh38
rs373547111 Chr.13: 101868716-101868716 on Build GRCh38
rs979375434 Chr.13: 101869042-101869042 on Build GRCh38
rs753254332 Chr.13: 101868714-101868714 on Build GRCh38
rs1004402869 Chr.13: 101868728-101868728 on Build GRCh38
rs531445068 Chr.13: 101868627-101868627 on Build GRCh38
rs779936498 Chr.13: 101868781-101868781 on Build GRCh38
rs556354477 Chr.13: 101868919-101868919 on Build GRCh38
rs952395143 Chr.13: 101868761-101868761 on Build GRCh38
rs369630983 Chr.13: 101868702-101868702 on Build GRCh38
rs41281646 Chr.13: 101868749-101868749 on Build GRCh38
rs370656178 Chr.13: 101868732-101868732 on Build GRCh38
rs370967683 Chr.13: 101868769-101868769 on Build GRCh38
rs934873674 Chr.13: 101868976-101868976 on Build GRCh38
rs750681546 Chr.13: 101868873-101868873 on Build GRCh38
rs749966703 Chr.13: 101869011-101869011 on Build GRCh38
rs771804837 Chr.13: 101868821-101868821 on Build GRCh38
rs954074692 Chr.13: 101868844-101868844 on Build GRCh38
rs201487248 Chr.13: 101868684-101868684 on Build GRCh38
rs1045887528 Chr.13: 101868939-101868939 on Build GRCh38
rs749302947 Chr.13: 101868785-101868785 on Build GRCh38
rs990942466 Chr.13: 101868799-101868799 on Build GRCh38
rs768469422 Chr.13: 101868789-101868789 on Build GRCh38
rs762761721 Chr.13: 101868694-101868694 on Build GRCh38
rs186978667 Chr.13: 101868701-101868701 on Build GRCh38
rs775993763 Chr.13: 101868847-101868847 on Build GRCh38
rs542655718 Chr.13: 101868860-101868860 on Build GRCh38
rs916328650 Chr.13: 101868909-101868909 on Build GRCh38
rs1053376626 Chr.13: 101868988-101868988 on Build GRCh38
rs760198072 Chr.13: 101868712-101868712 on Build GRCh38
rs578232125 Chr.13: 101868920-101868920 on Build GRCh38
rs7327756 Chr.13: 101868949-101868949 on Build GRCh38
rs944581259 Chr.13: 101868611-101868611 on Build GRCh38
rs372609706 Chr.13: 101868635-101868635 on Build GRCh38
rs747914647 Chr.13: 101868797-101868797 on Build GRCh38
rs34073982 Chr.13: 101868760-101868760 on Build GRCh38
rs765835014 Chr.13: 101868713-101868713 on Build GRCh38
rs926512251 Chr.13: 101869043-101869043 on Build GRCh38
rs1012044010 Chr.13: 101868604-101868604 on Build GRCh38
rs774237770 Chr.13: 101868796-101868796 on Build GRCh38
rs200021210 Chr.13: 101868837-101868837 on Build GRCh38
rs767955575 Chr.13: 101868865-101868865 on Build GRCh38

View Full Product Details