Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76035777-76036352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001302123.1 NM_005409.4

Gene Symbol
CXCL11
Entrez Gene ID
6373
Gene Name
C-X-C motif chemokine ligand 11
Gene Aliases
H174, I-TAC, IP-9, IP9, SCYB11, SCYB9B, b-R1
UniGene
Hs.632592
Gene Chromosome Location
Chr.4: 76033682-76036197 on Build GRCh38
Transcripts
NM_001130017.2 NM_001302123.1 NM_005409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545529 c.8T>A p.V3E Chr.4: 76035980-76035980 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000137559 Chr.4: 76036210-76036210 on Build GRCh38
rs776540552 Chr.4: 76035939-76035939 on Build GRCh38
rs201865900 Chr.4: 76035961-76035961 on Build GRCh38
rs377638073 Chr.4: 76036057-76036057 on Build GRCh38
rs1036801647 Chr.4: 76036160-76036160 on Build GRCh38
rs938083924 Chr.4: 76036190-76036190 on Build GRCh38
rs980347071 Chr.4: 76036091-76036091 on Build GRCh38
rs891814664 Chr.4: 76036304-76036304 on Build GRCh38
rs747293649 Chr.4: 76036035-76036035 on Build GRCh38
rs6819597 Chr.4: 76036018-76036018 on Build GRCh38
rs763263552 Chr.4: 76035965-76035965 on Build GRCh38
rs926658384 Chr.4: 76035929-76035929 on Build GRCh38
rs763670498 Chr.4: 76035895-76035895 on Build GRCh38
rs750416577 Chr.4: 76035901-76035901 on Build GRCh38
rs765701525 Chr.4: 76035883-76035883 on Build GRCh38
rs780049725 Chr.4: 76035920-76035920 on Build GRCh38
rs6817952 Chr.4: 76035890-76035890 on Build GRCh38
rs972301249 Chr.4: 76036127-76036127 on Build GRCh38
rs747499993 Chr.4: 76036119-76036119 on Build GRCh38
rs1048136573 Chr.4: 76035822-76035822 on Build GRCh38
rs933820569 Chr.4: 76036170-76036170 on Build GRCh38
rs1024819970 Chr.4: 76036327-76036327 on Build GRCh38
rs10021313 Chr.4: 76036090-76036090 on Build GRCh38
rs755947087 Chr.4: 76036015-76036016 on Build GRCh38
rs1019006971 Chr.4: 76036094-76036094 on Build GRCh38
rs1049144437 Chr.4: 76035925-76035925 on Build GRCh38
rs746446876 Chr.4: 76035940-76035940 on Build GRCh38
rs776023611 Chr.4: 76036098-76036098 on Build GRCh38
rs1032995708 Chr.4: 76036247-76036247 on Build GRCh38
rs374075849 Chr.4: 76035900-76035900 on Build GRCh38
rs755999293 Chr.4: 76035919-76035919 on Build GRCh38
rs140658168 Chr.4: 76036079-76036079 on Build GRCh38
rs749029946 Chr.4: 76035927-76035927 on Build GRCh38
rs778827835 Chr.4: 76036033-76036033 on Build GRCh38
rs1041665056 Chr.4: 76036326-76036326 on Build GRCh38
rs966683655 Chr.4: 76035875-76035875 on Build GRCh38
rs751105525 Chr.4: 76035897-76035897 on Build GRCh38
rs1010247046 Chr.4: 76036325-76036325 on Build GRCh38
rs201011439 Chr.4: 76036012-76036012 on Build GRCh38
rs775837584 Chr.4: 76035959-76035959 on Build GRCh38
rs753717714 Chr.4: 76036026-76036026 on Build GRCh38
rs888239542 Chr.4: 76035886-76035886 on Build GRCh38
rs767119436 Chr.4: 76035993-76035993 on Build GRCh38
rs767775467 Chr.4: 76035993-76035993 on Build GRCh38
rs573293891 Chr.4: 76036182-76036182 on Build GRCh38
rs771090412 Chr.4: 76035937-76035937 on Build GRCh38
rs939776923 Chr.4: 76036157-76036157 on Build GRCh38
rs778100475 Chr.4: 76035932-76035932 on Build GRCh38
rs377419643 Chr.4: 76036198-76036198 on Build GRCh38
rs1024402096 Chr.4: 76036084-76036084 on Build GRCh38
rs956150522 Chr.4: 76036076-76036076 on Build GRCh38
rs770143382 Chr.4: 76035948-76035948 on Build GRCh38
rs115261822 Chr.4: 76035898-76035898 on Build GRCh38
rs965715898 Chr.4: 76036124-76036124 on Build GRCh38
rs903033399 Chr.4: 76036056-76036056 on Build GRCh38
rs1041633998 Chr.4: 76036047-76036047 on Build GRCh38
rs986160914 Chr.4: 76035850-76035850 on Build GRCh38
rs945331541 Chr.4: 76036301-76036301 on Build GRCh38
rs1056861895 Chr.4: 76035994-76035994 on Build GRCh38
rs747081305 Chr.4: 76035934-76035934 on Build GRCh38
rs763762627 Chr.4: 76035974-76035974 on Build GRCh38
rs142872466 Chr.4: 76036188-76036188 on Build GRCh38
rs774042923 Chr.4: 76035987-76035987 on Build GRCh38
rs74885947 Chr.4: 76036143-76036143 on Build GRCh38
rs1033048986 Chr.4: 76036066-76036066 on Build GRCh38
rs983823179 Chr.4: 76036144-76036144 on Build GRCh38
rs900969372 Chr.4: 76036167-76036167 on Build GRCh38
rs138292834 Chr.4: 76036155-76036155 on Build GRCh38
rs752489277 Chr.4: 76036002-76036002 on Build GRCh38
rs371284323 Chr.4: 76036029-76036029 on Build GRCh38
rs1028666045 Chr.4: 76035942-76035942 on Build GRCh38
rs761343612 Chr.4: 76035989-76035989 on Build GRCh38
rs749825078 Chr.4: 76036001-76036001 on Build GRCh38
rs917930711 Chr.4: 76036260-76036260 on Build GRCh38
rs745486810 Chr.4: 76036177-76036177 on Build GRCh38
rs912180922 Chr.4: 76035869-76035869 on Build GRCh38
rs756087891 Chr.4: 76036013-76036013 on Build GRCh38

View Full Product Details