Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 20770139-20770731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOCAD
Entrez Gene ID
54914
Gene Name
focadhesin
Gene Aliases
KIAA1797
UniGene
Hs.136247
Gene Chromosome Location
Chr.9: 20658309-20995955 on Build GRCh38
Transcripts
NM_017794.4 XM_005251494.3 XM_011517945.2 XM_017014852.1 XM_017014853.1 XM_017014854.1 XM_017014855.1 XM_017014856.1 XM_017014857.1 XM_017014858.1 XM_017014859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746966 c.851G>A p.G284D Chr.9: 20770183-20770183 on Build GRCh38
3433063 c.892G>T p.E298* Chr.9: 20770224-20770224 on Build GRCh38
4773099 c.853G>T p.E285* Chr.9: 20770185-20770185 on Build GRCh38
5743324 c.823G>A p.V275I Chr.9: 20770155-20770155 on Build GRCh38
3091720 c.904G>A p.E302K Chr.9: 20770236-20770236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770893289 Chr.9: 20770176-20770176 on Build GRCh38
rs961205922 Chr.9: 20770683-20770683 on Build GRCh38
rs147542760 Chr.9: 20770291-20770291 on Build GRCh38
rs775010386 Chr.9: 20770186-20770186 on Build GRCh38
rs559621945 Chr.9: 20770587-20770587 on Build GRCh38
rs986630285 Chr.9: 20770246-20770246 on Build GRCh38
rs761092805 Chr.9: 20770200-20770200 on Build GRCh38
rs771290178 Chr.9: 20770446-20770446 on Build GRCh38
rs375930818 Chr.9: 20770589-20770589 on Build GRCh38
rs773589085 Chr.9: 20770196-20770196 on Build GRCh38
rs759910860 Chr.9: 20770583-20770583 on Build GRCh38
rs955091072 Chr.9: 20770411-20770411 on Build GRCh38
rs540507044 Chr.9: 20770405-20770405 on Build GRCh38
rs533960487 Chr.9: 20770653-20770653 on Build GRCh38
rs748927158 Chr.9: 20770266-20770266 on Build GRCh38
rs372465182 Chr.9: 20770282-20770282 on Build GRCh38
rs186635487 Chr.9: 20770258-20770258 on Build GRCh38
rs560469145 Chr.9: 20770429-20770429 on Build GRCh38
rs766778658 Chr.9: 20770275-20770275 on Build GRCh38
rs770432264 Chr.9: 20770224-20770225 on Build GRCh38
rs201871025 Chr.9: 20770517-20770517 on Build GRCh38
rs1054658202 Chr.9: 20770621-20770621 on Build GRCh38
rs1007454499 Chr.9: 20770380-20770380 on Build GRCh38
rs1010585597 Chr.9: 20770438-20770438 on Build GRCh38
rs556369166 Chr.9: 20770289-20770289 on Build GRCh38
rs567599069 Chr.9: 20770671-20770671 on Build GRCh38
rs930095215 Chr.9: 20770328-20770328 on Build GRCh38
rs966367066 Chr.9: 20770476-20770476 on Build GRCh38
rs934838170 Chr.9: 20770618-20770618 on Build GRCh38
rs762612023 Chr.9: 20770189-20770189 on Build GRCh38
rs1033682378 Chr.9: 20770358-20770358 on Build GRCh38
rs903362314 Chr.9: 20770339-20770339 on Build GRCh38
rs765263021 Chr.9: 20770279-20770279 on Build GRCh38
rs1045796755 Chr.9: 20770592-20770592 on Build GRCh38
rs767047526 Chr.9: 20770204-20770204 on Build GRCh38
rs999092629 Chr.9: 20770345-20770345 on Build GRCh38
rs912607842 Chr.9: 20770699-20770699 on Build GRCh38
rs781174644 Chr.9: 20770253-20770253 on Build GRCh38
rs752859974 Chr.9: 20770288-20770288 on Build GRCh38
rs1045862390 Chr.9: 20770333-20770333 on Build GRCh38
rs975150635 Chr.9: 20770166-20770166 on Build GRCh38
rs551591614 Chr.9: 20770541-20770541 on Build GRCh38
rs534462864 Chr.9: 20770220-20770220 on Build GRCh38
rs916503274 Chr.9: 20770574-20770574 on Build GRCh38
rs372253102 Chr.9: 20770256-20770256 on Build GRCh38
rs950105726 Chr.9: 20770590-20770590 on Build GRCh38
rs533241383 Chr.9: 20770627-20770627 on Build GRCh38
rs75301013 Chr.9: 20770331-20770331 on Build GRCh38
rs556332397 Chr.9: 20770673-20770673 on Build GRCh38
rs368914779 Chr.9: 20770245-20770245 on Build GRCh38
rs769343536 Chr.9: 20770184-20770184 on Build GRCh38
rs776529704 Chr.9: 20770180-20770180 on Build GRCh38
rs995731735 Chr.9: 20770686-20770686 on Build GRCh38
rs897252494 Chr.9: 20770311-20770311 on Build GRCh38
rs755110114 Chr.9: 20770215-20770215 on Build GRCh38
rs191090575 Chr.9: 20770285-20770285 on Build GRCh38
rs758606639 Chr.9: 20770219-20770219 on Build GRCh38
rs942898614 Chr.9: 20770705-20770705 on Build GRCh38
rs745738832 Chr.9: 20770254-20770254 on Build GRCh38
rs893359700 Chr.9: 20770645-20770645 on Build GRCh38
rs571589809 Chr.9: 20770217-20770217 on Build GRCh38
rs754389950 Chr.9: 20770214-20770214 on Build GRCh38
rs961523509 Chr.9: 20770471-20770471 on Build GRCh38
rs554045140 Chr.9: 20770241-20770241 on Build GRCh38
rs544017025 Chr.9: 20770298-20770298 on Build GRCh38
rs536598012 Chr.9: 20770672-20770672 on Build GRCh38
rs768397546 Chr.9: 20770267-20770267 on Build GRCh38
rs571567930 Chr.9: 20770631-20770631 on Build GRCh38
rs756645848 Chr.9: 20770310-20770310 on Build GRCh38
rs773202011 Chr.9: 20770387-20770387 on Build GRCh38
rs752884080 Chr.9: 20770218-20770218 on Build GRCh38
rs57349313 Chr.9: 20770670-20770670 on Build GRCh38
rs1028031825 Chr.9: 20770407-20770407 on Build GRCh38
rs775020053 Chr.9: 20770261-20770261 on Build GRCh38
rs9298809 Chr.9: 20770460-20770460 on Build GRCh38
rs973980021 Chr.9: 20770478-20770478 on Build GRCh38
rs903327854 Chr.9: 20770593-20770593 on Build GRCh38
rs991448426 Chr.9: 20770550-20770550 on Build GRCh38
rs991037472 Chr.9: 20770551-20770551 on Build GRCh38
rs530186508 Chr.9: 20770416-20770416 on Build GRCh38
rs892767891 Chr.9: 20770379-20770379 on Build GRCh38
rs9298810 Chr.9: 20770488-20770488 on Build GRCh38
rs761186476 Chr.9: 20770270-20770270 on Build GRCh38
rs771872098 Chr.9: 20770259-20770259 on Build GRCh38
rs767798738 Chr.9: 20770192-20770192 on Build GRCh38
rs200516182 Chr.9: 20770236-20770236 on Build GRCh38
rs1055753914 Chr.9: 20770308-20770308 on Build GRCh38
rs960585761 Chr.9: 20770543-20770543 on Build GRCh38
rs562718488 Chr.9: 20770501-20770501 on Build GRCh38
rs777287633 Chr.9: 20770276-20770276 on Build GRCh38
rs776445481 Chr.9: 20770288-20770289 on Build GRCh38
rs915948430 Chr.9: 20770564-20770564 on Build GRCh38
rs1027982887 Chr.9: 20770665-20770665 on Build GRCh38
rs760038872 Chr.9: 20770277-20770277 on Build GRCh38
rs1026861148 Chr.9: 20770687-20770687 on Build GRCh38
rs199930049 Chr.9: 20770315-20770315 on Build GRCh38
rs966417554 Chr.9: 20770175-20770175 on Build GRCh38
rs577570348 Chr.9: 20770395-20770395 on Build GRCh38
rs764436536 Chr.9: 20770290-20770290 on Build GRCh38
rs745442705 Chr.9: 20770181-20770181 on Build GRCh38
rs774131179 Chr.9: 20770269-20770269 on Build GRCh38

View Full Product Details