Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 63912583-63913121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
UniGene
Hs.476595
Gene Chromosome Location
Chr.3: 63864557-64003462 on Build GRCh38
Transcripts
NM_000333.3 NM_001177387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408857 c.198C>T p.G66G Chr.3: 63912796-63912796 on Build GRCh38
4119785 c.315T>C p.P105P Chr.3: 63912913-63912913 on Build GRCh38
1131365 c.240G>A p.E80E Chr.3: 63912838-63912838 on Build GRCh38
5517953 c.126T>C p.P42P Chr.3: 63912724-63912724 on Build GRCh38
4119784 c.315T>C p.P105P Chr.3: 63912913-63912913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758571761 Chr.3: 63912786-63912786 on Build GRCh38
rs767066861 Chr.3: 63912699-63912699 on Build GRCh38
rs1002448866 Chr.3: 63912641-63912641 on Build GRCh38
rs879074992 Chr.3: 63912687-63912687 on Build GRCh38
rs770246503 Chr.3: 63912957-63912957 on Build GRCh38
rs564106533 Chr.3: 63912920-63912920 on Build GRCh38
rs747271054 Chr.3: 63912789-63912789 on Build GRCh38
rs1011041837 Chr.3: 63912662-63912662 on Build GRCh38
rs773822597 Chr.3: 63912645-63912645 on Build GRCh38
rs752810032 Chr.3: 63912785-63912785 on Build GRCh38
rs897546151 Chr.3: 63912630-63912630 on Build GRCh38
rs767487520 Chr.3: 63912644-63912646 on Build GRCh38
rs538503525 Chr.3: 63912696-63912696 on Build GRCh38
rs542050695 Chr.3: 63912851-63912851 on Build GRCh38
rs1024213914 Chr.3: 63912747-63912747 on Build GRCh38
rs777627983 Chr.3: 63912949-63912949 on Build GRCh38
rs774055087 Chr.3: 63912749-63912757 on Build GRCh38
rs757472878 Chr.3: 63912838-63912838 on Build GRCh38
rs746524497 Chr.3: 63912953-63912953 on Build GRCh38
rs760500769 Chr.3: 63912682-63912683 on Build GRCh38
rs561644071 Chr.3: 63912961-63912961 on Build GRCh38
rs764551639 Chr.3: 63912912-63912912 on Build GRCh38
rs775462169 Chr.3: 63912943-63912944 on Build GRCh38
rs938304643 Chr.3: 63912754-63912754 on Build GRCh38
rs759914630 Chr.3: 63912899-63912899 on Build GRCh38
rs981883891 Chr.3: 63913051-63913055 on Build GRCh38
rs531605953 Chr.3: 63912954-63912954 on Build GRCh38
rs894084261 Chr.3: 63913098-63913098 on Build GRCh38
rs751693748 Chr.3: 63912835-63912835 on Build GRCh38
rs562754099 Chr.3: 63912701-63912702 on Build GRCh38
rs199563441 Chr.3: 63912944-63912944 on Build GRCh38
rs572429085 Chr.3: 63912886-63912886 on Build GRCh38
rs747587822 Chr.3: 63912803-63912803 on Build GRCh38
rs755508121 Chr.3: 63912777-63912777 on Build GRCh38
rs781445074 Chr.3: 63912839-63912839 on Build GRCh38
rs922612813 Chr.3: 63912685-63912690 on Build GRCh38
rs1051696903 Chr.3: 63913043-63913043 on Build GRCh38
rs539552087 Chr.3: 63912612-63912612 on Build GRCh38
rs777111926 Chr.3: 63912817-63912817 on Build GRCh38
rs781716445 Chr.3: 63912685-63912693 on Build GRCh38
rs776092702 Chr.3: 63912908-63912908 on Build GRCh38
rs910378260 Chr.3: 63912610-63912610 on Build GRCh38
rs770295927 Chr.3: 63912904-63912904 on Build GRCh38
rs762884569 Chr.3: 63912833-63912833 on Build GRCh38
rs1046517241 Chr.3: 63912719-63912719 on Build GRCh38
rs766786119 Chr.3: 63912767-63912767 on Build GRCh38
rs201152899 Chr.3: 63912877-63912877 on Build GRCh38
rs749124142 Chr.3: 63912796-63912796 on Build GRCh38
rs889065976 Chr.3: 63913052-63913052 on Build GRCh38
rs1021114622 Chr.3: 63912804-63912804 on Build GRCh38
rs199663915 Chr.3: 63912821-63912821 on Build GRCh38
rs529226400 Chr.3: 63912988-63912988 on Build GRCh38
rs374352159 Chr.3: 63912955-63912955 on Build GRCh38
rs760493800 Chr.3: 63912944-63912944 on Build GRCh38
rs376084779 Chr.3: 63912911-63912911 on Build GRCh38
rs919026295 Chr.3: 63912708-63912708 on Build GRCh38
rs748653222 Chr.3: 63912697-63912698 on Build GRCh38
rs575260447 Chr.3: 63912829-63912829 on Build GRCh38
rs779859093 Chr.3: 63912793-63912793 on Build GRCh38
rs1055385554 Chr.3: 63912757-63912757 on Build GRCh38
rs1053660143 Chr.3: 63913075-63913075 on Build GRCh38
rs998506326 Chr.3: 63912959-63912959 on Build GRCh38
rs777791690 Chr.3: 63912788-63912788 on Build GRCh38
rs780440474 Chr.3: 63912685-63912687 on Build GRCh38
rs966759720 Chr.3: 63912680-63912680 on Build GRCh38
rs1037414995 Chr.3: 63912628-63912628 on Build GRCh38
rs546300041 Chr.3: 63912888-63912888 on Build GRCh38
rs781167034 Chr.3: 63913057-63913057 on Build GRCh38
rs755818890 Chr.3: 63912791-63912791 on Build GRCh38
rs754838412 Chr.3: 63912848-63912848 on Build GRCh38
rs770527876 Chr.3: 63912813-63912813 on Build GRCh38
rs1036739776 Chr.3: 63912923-63912923 on Build GRCh38
rs532856549 Chr.3: 63913079-63913079 on Build GRCh38
rs576125874 Chr.3: 63912640-63912640 on Build GRCh38
rs771619986 Chr.3: 63912808-63912808 on Build GRCh38
rs751638330 Chr.3: 63912915-63912915 on Build GRCh38
rs770364745 Chr.3: 63912714-63912715 on Build GRCh38
rs565893504 Chr.3: 63913017-63913017 on Build GRCh38
rs752638522 Chr.3: 63912650-63912673 on Build GRCh38
rs954823047 Chr.3: 63912685-63912705 on Build GRCh38
rs774791610 Chr.3: 63912974-63912974 on Build GRCh38
rs772918244 Chr.3: 63912702-63912702 on Build GRCh38
rs752472931 Chr.3: 63912948-63912948 on Build GRCh38
rs749666427 Chr.3: 63912967-63912967 on Build GRCh38
rs774334964 Chr.3: 63912801-63912801 on Build GRCh38
rs547258246 Chr.3: 63913016-63913016 on Build GRCh38
rs745537994 Chr.3: 63912956-63912956 on Build GRCh38
rs976770853 Chr.3: 63912681-63912681 on Build GRCh38
rs370299464 Chr.3: 63912855-63912855 on Build GRCh38
rs201561286 Chr.3: 63912809-63912809 on Build GRCh38
rs993137587 Chr.3: 63912632-63912632 on Build GRCh38
rs557445610 Chr.3: 63912635-63912635 on Build GRCh38
rs745367934 Chr.3: 63912939-63912975 on Build GRCh38
rs866637540 Chr.3: 63912690-63912690 on Build GRCh38
rs754714055 Chr.3: 63912946-63912946 on Build GRCh38
rs193922929 Chr.3: 63912686-63912715 on Build GRCh38
rs958875926 Chr.3: 63912863-63912863 on Build GRCh38
rs771527403 Chr.3: 63912942-63912943 on Build GRCh38
rs1053336 Chr.3: 63912895-63912895 on Build GRCh38
rs765894375 Chr.3: 63912781-63912781 on Build GRCh38
rs933212191 Chr.3: 63913037-63913037 on Build GRCh38
rs986062347 Chr.3: 63912693-63912693 on Build GRCh38
rs767725471 Chr.3: 63912940-63912940 on Build GRCh38
rs768562639 Chr.3: 63912798-63912798 on Build GRCh38
rs909280730 Chr.3: 63912887-63912887 on Build GRCh38
rs753261682 Chr.3: 63912830-63912830 on Build GRCh38
rs1034299413 Chr.3: 63912642-63912642 on Build GRCh38
rs764153847 Chr.3: 63912971-63912972 on Build GRCh38
rs576518931 Chr.3: 63912684-63912685 on Build GRCh38
rs543266864 Chr.3: 63912958-63912958 on Build GRCh38
rs201796752 Chr.3: 63912818-63912818 on Build GRCh38
rs771439534 Chr.3: 63912864-63912864 on Build GRCh38
rs958191401 Chr.3: 63912648-63912648 on Build GRCh38
rs1021062254 Chr.3: 63912675-63912675 on Build GRCh38
rs769196344 Chr.3: 63912968-63912968 on Build GRCh38
rs760887474 Chr.3: 63912722-63912722 on Build GRCh38
rs893539558 Chr.3: 63913076-63913076 on Build GRCh38
rs754355306 Chr.3: 63912776-63912776 on Build GRCh38
rs919269181 Chr.3: 63912926-63912926 on Build GRCh38
rs919078725 Chr.3: 63912991-63912991 on Build GRCh38
rs753621707 Chr.3: 63912840-63912840 on Build GRCh38
rs201334618 Chr.3: 63912871-63912871 on Build GRCh38
rs929040913 Chr.3: 63912709-63912709 on Build GRCh38
rs868854400 Chr.3: 63912714-63912714 on Build GRCh38
rs910421275 Chr.3: 63912694-63912695 on Build GRCh38

View Full Product Details