Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105339218-105339783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf49
Entrez Gene ID
79074
Gene Name
chromosome 2 open reading frame 49
Gene Aliases
asw
UniGene
Hs.549577
Gene Chromosome Location
Chr.2: 105337257-105359940 on Build GRCh38
Transcripts
NM_001286537.1 NM_024093.2 XM_017004892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6466167 c.253G>A p.D85N Chr.2: 105339622-105339622 on Build GRCh38
3836472 c.176C>G p.P59R Chr.2: 105339659-105339659 on Build GRCh38
1004769 c.174A>G p.I58M Chr.2: 105339657-105339657 on Build GRCh38
5736165 c.188G>T p.R63M Chr.2: 105339671-105339671 on Build GRCh38
3564674 c.265G>A p.D89N Chr.2: 105339634-105339634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760084452 Chr.2: 105339682-105339682 on Build GRCh38
rs924003093 Chr.2: 105339510-105339510 on Build GRCh38
rs774117581 Chr.2: 105339668-105339668 on Build GRCh38
rs755559070 Chr.2: 105339594-105339594 on Build GRCh38
rs147396516 Chr.2: 105339605-105339605 on Build GRCh38
rs369225704 Chr.2: 105339396-105339396 on Build GRCh38
rs192026146 Chr.2: 105339477-105339477 on Build GRCh38
rs766877216 Chr.2: 105339539-105339539 on Build GRCh38
rs751744611 Chr.2: 105339727-105339727 on Build GRCh38
rs984823057 Chr.2: 105339622-105339622 on Build GRCh38
rs898293384 Chr.2: 105339260-105339260 on Build GRCh38
rs1007653525 Chr.2: 105339412-105339412 on Build GRCh38
rs529270510 Chr.2: 105339508-105339508 on Build GRCh38
rs552115885 Chr.2: 105339451-105339451 on Build GRCh38
rs566813931 Chr.2: 105339282-105339282 on Build GRCh38
rs181746891 Chr.2: 105339267-105339267 on Build GRCh38
rs747048708 Chr.2: 105339624-105339624 on Build GRCh38
rs781144157 Chr.2: 105339634-105339634 on Build GRCh38
rs768281690 Chr.2: 105339655-105339655 on Build GRCh38
rs183157251 Chr.2: 105339636-105339636 on Build GRCh38
rs769508063 Chr.2: 105339277-105339277 on Build GRCh38
rs761423521 Chr.2: 105339321-105339321 on Build GRCh38
rs552618417 Chr.2: 105339707-105339707 on Build GRCh38
rs369458082 Chr.2: 105339579-105339579 on Build GRCh38
rs1038223297 Chr.2: 105339250-105339250 on Build GRCh38
rs779375738 Chr.2: 105339596-105339596 on Build GRCh38
rs866603582 Chr.2: 105339636-105339637 on Build GRCh38
rs771620917 Chr.2: 105339414-105339414 on Build GRCh38
rs746323773 Chr.2: 105339749-105339758 on Build GRCh38
rs373759867 Chr.2: 105339454-105339454 on Build GRCh38
rs763693517 Chr.2: 105339719-105339721 on Build GRCh38
rs765632108 Chr.2: 105339740-105339740 on Build GRCh38
rs745987093 Chr.2: 105339746-105339746 on Build GRCh38
rs761380102 Chr.2: 105339673-105339673 on Build GRCh38
rs1049738709 Chr.2: 105339313-105339313 on Build GRCh38
rs766998627 Chr.2: 105339680-105339680 on Build GRCh38
rs758663370 Chr.2: 105339610-105339610 on Build GRCh38
rs111947412 Chr.2: 105339432-105339432 on Build GRCh38
rs575436900 Chr.2: 105339328-105339328 on Build GRCh38
rs369657994 Chr.2: 105339418-105339418 on Build GRCh38
rs755760937 Chr.2: 105339563-105339563 on Build GRCh38
rs79232208 Chr.2: 105339516-105339516 on Build GRCh38
rs200946111 Chr.2: 105339554-105339554 on Build GRCh38
rs769754399 Chr.2: 105339645-105339645 on Build GRCh38
rs530740042 Chr.2: 105339618-105339618 on Build GRCh38
rs748981596 Chr.2: 105339618-105339618 on Build GRCh38
rs368739873 Chr.2: 105339468-105339468 on Build GRCh38
rs759880474 Chr.2: 105339702-105339702 on Build GRCh38
rs1008366461 Chr.2: 105339657-105339657 on Build GRCh38
rs756653775 Chr.2: 105339637-105339638 on Build GRCh38
rs777346544 Chr.2: 105339568-105339568 on Build GRCh38
rs899458252 Chr.2: 105339696-105339696 on Build GRCh38
rs960566118 Chr.2: 105339517-105339517 on Build GRCh38
rs968174642 Chr.2: 105339497-105339497 on Build GRCh38
rs765886202 Chr.2: 105339697-105339697 on Build GRCh38
rs762781213 Chr.2: 105339654-105339654 on Build GRCh38
rs752529468 Chr.2: 105339559-105339562 on Build GRCh38
rs1010572100 Chr.2: 105339456-105339456 on Build GRCh38
rs919246640 Chr.2: 105339257-105339257 on Build GRCh38
rs145082926 Chr.2: 105339681-105339681 on Build GRCh38
rs751007963 Chr.2: 105339538-105339538 on Build GRCh38
rs762756032 Chr.2: 105339504-105339504 on Build GRCh38
rs766793992 Chr.2: 105339577-105339577 on Build GRCh38
rs1022608744 Chr.2: 105339462-105339462 on Build GRCh38
rs767277979 Chr.2: 105339533-105339538 on Build GRCh38
rs76512432 Chr.2: 105339633-105339633 on Build GRCh38
rs202080360 Chr.2: 105339722-105339722 on Build GRCh38
rs141950651 Chr.2: 105339717-105339717 on Build GRCh38
rs889394568 Chr.2: 105339393-105339393 on Build GRCh38
rs781291440 Chr.2: 105339745-105339745 on Build GRCh38
rs753090039 Chr.2: 105339706-105339706 on Build GRCh38
rs186831577 Chr.2: 105339453-105339453 on Build GRCh38
rs775230034 Chr.2: 105339651-105339651 on Build GRCh38

View Full Product Details