Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53941426-53941984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
UniGene
Hs.631597
Gene Chromosome Location
Chr.19: 53909425-53943944 on Build GRCh38
Transcripts
NM_031896.4 XM_005259124.3 XM_017027093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550743 c.507G>T p.Q169H Chr.19: 53941552-53941552 on Build GRCh38
5528064 c.432C>T p.S144S Chr.19: 53941477-53941477 on Build GRCh38
4448399 c.570+8_570+9CC>TT p.? Chr.19: 53941623-53941624 on Build GRCh38
7302878 c.448G>T p.V150F Chr.19: 53941493-53941493 on Build GRCh38
7359259 c.451C>G p.L151V Chr.19: 53941496-53941496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530496577 Chr.19: 53941882-53941882 on Build GRCh38
rs759629903 Chr.19: 53941531-53941531 on Build GRCh38
rs185901155 Chr.19: 53941856-53941856 on Build GRCh38
rs746925804 Chr.19: 53941633-53941633 on Build GRCh38
rs529135262 Chr.19: 53941753-53941753 on Build GRCh38
rs768161757 Chr.19: 53941589-53941589 on Build GRCh38
rs778373164 Chr.19: 53941621-53941621 on Build GRCh38
rs751982783 Chr.19: 53941550-53941550 on Build GRCh38
rs762462283 Chr.19: 53941560-53941560 on Build GRCh38
rs543348527 Chr.19: 53941691-53941691 on Build GRCh38
rs903324459 Chr.19: 53941788-53941788 on Build GRCh38
rs752695327 Chr.19: 53941924-53941924 on Build GRCh38
rs959901796 Chr.19: 53941447-53941447 on Build GRCh38
rs773952208 Chr.19: 53941585-53941592 on Build GRCh38
rs540825647 Chr.19: 53941754-53941754 on Build GRCh38
rs898408434 Chr.19: 53941946-53941946 on Build GRCh38
rs775782783 Chr.19: 53941538-53941538 on Build GRCh38
rs776738193 Chr.19: 53941494-53941494 on Build GRCh38
rs754541309 Chr.19: 53941603-53941603 on Build GRCh38
rs1042286969 Chr.19: 53941720-53941720 on Build GRCh38
rs984542565 Chr.19: 53941627-53941627 on Build GRCh38
rs7260411 Chr.19: 53941718-53941718 on Build GRCh38
rs557702350 Chr.19: 53941670-53941670 on Build GRCh38
rs575949633 Chr.19: 53941671-53941671 on Build GRCh38
rs1025434271 Chr.19: 53941641-53941641 on Build GRCh38
rs1050826540 Chr.19: 53941822-53941822 on Build GRCh38
rs777753507 Chr.19: 53941813-53941813 on Build GRCh38
rs752451034 Chr.19: 53941624-53941624 on Build GRCh38
rs202206501 Chr.19: 53941620-53941620 on Build GRCh38
rs571950700 Chr.19: 53941813-53941813 on Build GRCh38
rs999005201 Chr.19: 53941802-53941802 on Build GRCh38
rs911493872 Chr.19: 53941707-53941707 on Build GRCh38
rs1014043641 Chr.19: 53941837-53941837 on Build GRCh38
rs745879482 Chr.19: 53941669-53941669 on Build GRCh38
rs781135024 Chr.19: 53941647-53941647 on Build GRCh38
rs532856660 Chr.19: 53941816-53941816 on Build GRCh38
rs761350470 Chr.19: 53941565-53941565 on Build GRCh38
rs1043101545 Chr.19: 53941742-53941742 on Build GRCh38
rs1022809323 Chr.19: 53941838-53941838 on Build GRCh38
rs540884013 Chr.19: 53941825-53941825 on Build GRCh38
rs747015317 Chr.19: 53941453-53941453 on Build GRCh38
rs765401386 Chr.19: 53941535-53941535 on Build GRCh38
rs961434770 Chr.19: 53941962-53941962 on Build GRCh38
rs559399924 Chr.19: 53941771-53941771 on Build GRCh38
rs374671812 Chr.19: 53941591-53941591 on Build GRCh38
rs768884421 Chr.19: 53941678-53941678 on Build GRCh38
rs1017348389 Chr.19: 53941883-53941883 on Build GRCh38
rs1049966197 Chr.19: 53941694-53941694 on Build GRCh38
rs563306927 Chr.19: 53941867-53941867 on Build GRCh38
rs974350162 Chr.19: 53941635-53941635 on Build GRCh38
rs764589712 Chr.19: 53941549-53941549 on Build GRCh38
rs759565578 Chr.19: 53941522-53941522 on Build GRCh38
rs371164760 Chr.19: 53941643-53941643 on Build GRCh38
rs111313486 Chr.19: 53941712-53941712 on Build GRCh38
rs370188701 Chr.19: 53941472-53941472 on Build GRCh38
rs763278689 Chr.19: 53941674-53941674 on Build GRCh38
rs904212614 Chr.19: 53941756-53941756 on Build GRCh38
rs750129348 Chr.19: 53941688-53941688 on Build GRCh38
rs367730313 Chr.19: 53941625-53941625 on Build GRCh38
rs970368416 Chr.19: 53941878-53941878 on Build GRCh38
rs763217669 Chr.19: 53941547-53941547 on Build GRCh38

View Full Product Details