Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28695145-28695688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK2
Entrez Gene ID
11200
Gene Name
checkpoint kinase 2
Gene Aliases
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
UniGene
Hs.291363
Gene Chromosome Location
Chr.22: 28687743-28741866 on Build GRCh38
Transcripts
NM_001005735.1 NM_001257387.1 NM_007194.3 NM_145862.2 XM_006724114.3 XM_006724116.2 XM_011529839.2 XM_011529840.2 XM_011529841.1 XM_011529842.2 XM_011529843.1 XM_011529845.2 XM_017028560.1 XM_017028561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361160 c.1343T>G p.I448S Chr.22: 28695159-28695159 on Build GRCh38
6980423 c.1406C>T p.P469L Chr.22: 28695225-28695225 on Build GRCh38
7361159 c.1472T>G p.I491S Chr.22: 28695159-28695159 on Build GRCh38
6980424 c.1277C>T p.P426L Chr.22: 28695225-28695225 on Build GRCh38
6264429 c.1348G>A p.E450K Chr.22: 28695154-28695154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754520926 Chr.22: 28695284-28695284 on Build GRCh38
rs876660046 Chr.22: 28695181-28695181 on Build GRCh38
rs34016108 Chr.22: 28695285-28695286 on Build GRCh38
rs760037989 Chr.22: 28695255-28695258 on Build GRCh38
rs573757563 Chr.22: 28695656-28695656 on Build GRCh38
rs730881706 Chr.22: 28695252-28695252 on Build GRCh38
rs587780174 Chr.22: 28695239-28695239 on Build GRCh38
rs769933461 Chr.22: 28695225-28695225 on Build GRCh38
rs928628514 Chr.22: 28695430-28695430 on Build GRCh38
rs1043967686 Chr.22: 28695520-28695520 on Build GRCh38
rs78674844 Chr.22: 28695450-28695450 on Build GRCh38
rs17887238 Chr.22: 28695368-28695368 on Build GRCh38
rs553959467 Chr.22: 28695399-28695399 on Build GRCh38
rs769803979 Chr.22: 28695524-28695524 on Build GRCh38
rs77670999 Chr.22: 28695455-28695455 on Build GRCh38
rs139366548 Chr.22: 28695232-28695232 on Build GRCh38
rs78789077 Chr.22: 28695423-28695423 on Build GRCh38
rs571578266 Chr.22: 28695274-28695274 on Build GRCh38
rs781514295 Chr.22: 28695218-28695218 on Build GRCh38
rs28601856 Chr.22: 28695227-28695227 on Build GRCh38
rs375668299 Chr.22: 28695454-28695454 on Build GRCh38
rs863224747 Chr.22: 28695250-28695250 on Build GRCh38
rs749368980 Chr.22: 28695233-28695233 on Build GRCh38
rs541772829 Chr.22: 28695546-28695546 on Build GRCh38
rs1053216992 Chr.22: 28695477-28695477 on Build GRCh38
rs878854915 Chr.22: 28695185-28695185 on Build GRCh38
rs1029278610 Chr.22: 28695539-28695539 on Build GRCh38
rs76797961 Chr.22: 28695427-28695427 on Build GRCh38
rs779383555 Chr.22: 28695177-28695177 on Build GRCh38
rs771772104 Chr.22: 28695211-28695212 on Build GRCh38
rs876660323 Chr.22: 28695202-28695202 on Build GRCh38
rs949005966 Chr.22: 28695379-28695379 on Build GRCh38
rs916136968 Chr.22: 28695376-28695376 on Build GRCh38
rs538526081 Chr.22: 28695333-28695333 on Build GRCh38
rs587780175 Chr.22: 28695209-28695209 on Build GRCh38
rs778989252 Chr.22: 28695187-28695187 on Build GRCh38
rs587778194 Chr.22: 28695169-28695169 on Build GRCh38
rs758102180 Chr.22: 28695215-28695215 on Build GRCh38
rs375832920 Chr.22: 28695292-28695292 on Build GRCh38
rs373864492 Chr.22: 28695212-28695212 on Build GRCh38
rs1003902645 Chr.22: 28695649-28695649 on Build GRCh38
rs765926901 Chr.22: 28695278-28695289 on Build GRCh38
rs878854914 Chr.22: 28695195-28695195 on Build GRCh38
rs762205611 Chr.22: 28695242-28695242 on Build GRCh38
rs974719594 Chr.22: 28695372-28695372 on Build GRCh38
rs786201906 Chr.22: 28695243-28695243 on Build GRCh38
rs756997911 Chr.22: 28695419-28695419 on Build GRCh38
rs745384799 Chr.22: 28695483-28695483 on Build GRCh38
rs908703196 Chr.22: 28695301-28695301 on Build GRCh38
rs1025801565 Chr.22: 28695321-28695321 on Build GRCh38
rs753839004 Chr.22: 28695179-28695179 on Build GRCh38
rs730881691 Chr.22: 28695235-28695235 on Build GRCh38
rs17882922 Chr.22: 28695196-28695196 on Build GRCh38
rs878854912 Chr.22: 28695248-28695248 on Build GRCh38
rs587782299 Chr.22: 28695216-28695216 on Build GRCh38
rs549755590 Chr.22: 28695237-28695237 on Build GRCh38
rs892280999 Chr.22: 28695493-28695494 on Build GRCh38
rs137853011 Chr.22: 28695219-28695219 on Build GRCh38
rs200050883 Chr.22: 28695190-28695190 on Build GRCh38
rs962936394 Chr.22: 28695360-28695360 on Build GRCh38
rs765916432 Chr.22: 28695273-28695273 on Build GRCh38
rs767735261 Chr.22: 28695248-28695248 on Build GRCh38
rs75913383 Chr.22: 28695467-28695467 on Build GRCh38
rs534342752 Chr.22: 28695510-28695510 on Build GRCh38
rs946539491 Chr.22: 28695494-28695494 on Build GRCh38
rs76780431 Chr.22: 28695472-28695472 on Build GRCh38
rs587780176 Chr.22: 28695184-28695184 on Build GRCh38
rs555437811 Chr.22: 28695602-28695602 on Build GRCh38

View Full Product Details