Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97854374-97854900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXE1
Entrez Gene ID
2304
Gene Name
forkhead box E1
Gene Aliases
FKHL15, FOXE2, HFKH4, HFKL5, NMTC4, TITF2, TTF-2, TTF2
UniGene
Hs.159234
Gene Chromosome Location
Chr.9: 97853255-97856715 on Build GRCh38
Transcripts
NM_004473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5485027 c.528C>T p.A176A Chr.9: 97854442-97854442 on Build GRCh38
1207410 c.491C>T p.A164V Chr.9: 97854405-97854405 on Build GRCh38
3779466 c.955G>C p.E319Q Chr.9: 97854869-97854869 on Build GRCh38
5037354 c.777C>T p.T259T Chr.9: 97854691-97854691 on Build GRCh38
4985777 c.743C>G p.A248G Chr.9: 97854657-97854657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759780973 Chr.9: 97854587-97854587 on Build GRCh38
rs896035611 Chr.9: 97854703-97854703 on Build GRCh38
rs1036653031 Chr.9: 97854740-97854740 on Build GRCh38
rs770096351 Chr.9: 97854409-97854410 on Build GRCh38
rs142235457 Chr.9: 97854760-97854760 on Build GRCh38
rs764574001 Chr.9: 97854643-97854643 on Build GRCh38
rs774373703 Chr.9: 97854530-97854530 on Build GRCh38
rs3021524 Chr.9: 97854418-97854418 on Build GRCh38
rs768756124 Chr.9: 97854437-97854437 on Build GRCh38
rs538912281 Chr.9: 97854657-97854657 on Build GRCh38
rs187232918 Chr.9: 97854806-97854806 on Build GRCh38
rs1006530083 Chr.9: 97854799-97854799 on Build GRCh38
rs537210706 Chr.9: 97854499-97854499 on Build GRCh38
rs1027187148 Chr.9: 97854537-97854537 on Build GRCh38
rs749994728 Chr.9: 97854874-97854874 on Build GRCh38
rs764631482 Chr.9: 97854869-97854869 on Build GRCh38
rs965279677 Chr.9: 97854591-97854591 on Build GRCh38
rs556903865 Chr.9: 97854596-97854596 on Build GRCh38
rs751660227 Chr.9: 97854733-97854733 on Build GRCh38
rs756579198 Chr.9: 97854629-97854629 on Build GRCh38
rs3021525 Chr.9: 97854628-97854628 on Build GRCh38
rs565664344 Chr.9: 97854421-97854421 on Build GRCh38
rs775187654 Chr.9: 97854412-97854413 on Build GRCh38
rs562189970 Chr.9: 97854418-97854419 on Build GRCh38
rs763695737 Chr.9: 97854413-97854418 on Build GRCh38
rs953725958 Chr.9: 97854554-97854554 on Build GRCh38
rs764532591 Chr.9: 97854847-97854847 on Build GRCh38
rs866903665 Chr.9: 97854573-97854573 on Build GRCh38
rs988624138 Chr.9: 97854461-97854461 on Build GRCh38
rs758175291 Chr.9: 97854729-97854729 on Build GRCh38
rs377692338 Chr.9: 97854795-97854795 on Build GRCh38
rs767588934 Chr.9: 97854416-97854418 on Build GRCh38
rs753158939 Chr.9: 97854401-97854401 on Build GRCh38
rs751942900 Chr.9: 97854396-97854396 on Build GRCh38
rs767764330 Chr.9: 97854808-97854808 on Build GRCh38
rs754231919 Chr.9: 97854861-97854861 on Build GRCh38
rs766497891 Chr.9: 97854522-97854522 on Build GRCh38
rs913070188 Chr.9: 97854611-97854611 on Build GRCh38
rs940947787 Chr.9: 97854781-97854781 on Build GRCh38
rs777312882 Chr.9: 97854412-97854412 on Build GRCh38
rs574188791 Chr.9: 97854793-97854793 on Build GRCh38
rs71369530 Chr.9: 97854446-97854451 on Build GRCh38
rs1015105511 Chr.9: 97854516-97854516 on Build GRCh38
rs1057444505 Chr.9: 97854673-97854673 on Build GRCh38
rs867468354 Chr.9: 97854851-97854851 on Build GRCh38
rs755282859 Chr.9: 97854397-97854397 on Build GRCh38
rs757806215 Chr.9: 97854865-97854865 on Build GRCh38
rs756663689 Chr.9: 97854668-97854668 on Build GRCh38
rs542731474 Chr.9: 97854791-97854791 on Build GRCh38
rs755911172 Chr.9: 97854419-97854430 on Build GRCh38
rs913291080 Chr.9: 97854481-97854481 on Build GRCh38
rs111863093 Chr.9: 97854715-97854715 on Build GRCh38
rs758622111 Chr.9: 97854419-97854427 on Build GRCh38
rs746922579 Chr.9: 97854433-97854433 on Build GRCh38
rs755194188 Chr.9: 97854424-97854425 on Build GRCh38
rs778487508 Chr.9: 97854422-97854422 on Build GRCh38
rs1057363769 Chr.9: 97854486-97854486 on Build GRCh38
rs779966006 Chr.9: 97854733-97854733 on Build GRCh38
rs766143203 Chr.9: 97854419-97854433 on Build GRCh38
rs774004748 Chr.9: 97854413-97854427 on Build GRCh38
rs1043573644 Chr.9: 97854482-97854482 on Build GRCh38
rs775827100 Chr.9: 97854825-97854825 on Build GRCh38
rs778490079 Chr.9: 97854671-97854671 on Build GRCh38
rs745468926 Chr.9: 97854714-97854714 on Build GRCh38
rs547566398 Chr.9: 97854415-97854415 on Build GRCh38
rs748786457 Chr.9: 97854413-97854413 on Build GRCh38
rs756633940 Chr.9: 97854410-97854410 on Build GRCh38
rs371516340 Chr.9: 97854424-97854424 on Build GRCh38
rs770871331 Chr.9: 97854772-97854772 on Build GRCh38
rs947241980 Chr.9: 97854626-97854626 on Build GRCh38
rs776377853 Chr.9: 97854758-97854758 on Build GRCh38
rs536995636 Chr.9: 97854532-97854532 on Build GRCh38
rs182535331 Chr.9: 97854483-97854483 on Build GRCh38
rs896086448 Chr.9: 97854492-97854492 on Build GRCh38
rs1045902549 Chr.9: 97854786-97854786 on Build GRCh38
rs771953408 Chr.9: 97854427-97854427 on Build GRCh38
rs937334255 Chr.9: 97854665-97854665 on Build GRCh38
rs775448711 Chr.9: 97854430-97854430 on Build GRCh38
rs761084888 Chr.9: 97854826-97854826 on Build GRCh38
rs762571620 Chr.9: 97854413-97854424 on Build GRCh38
rs985554868 Chr.9: 97854579-97854579 on Build GRCh38
rs768628336 Chr.9: 97854749-97854749 on Build GRCh38
rs761789400 Chr.9: 97854413-97854439 on Build GRCh38
rs3021526 Chr.9: 97854739-97854739 on Build GRCh38
rs532218005 Chr.9: 97854400-97854400 on Build GRCh38

View Full Product Details