Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67431175-67431709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD11B2
Entrez Gene ID
3291
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
AME, AME1, HSD11K, HSD2, SDR9C3
UniGene
Hs.1376
Gene Chromosome Location
Chr.16: 67431133-67437553 on Build GRCh38
Transcripts
NM_000196.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863091 c.174C>T p.L58L Chr.16: 67431422-67431422 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs55681036 Chr.16: 67431632-67431632 on Build GRCh38
rs957367492 Chr.16: 67431202-67431202 on Build GRCh38
rs964433254 Chr.16: 67431220-67431220 on Build GRCh38
rs1011724863 Chr.16: 67431566-67431566 on Build GRCh38
rs775832824 Chr.16: 67431423-67431423 on Build GRCh38
rs370549443 Chr.16: 67431457-67431457 on Build GRCh38
rs765574993 Chr.16: 67431372-67431372 on Build GRCh38
rs999673404 Chr.16: 67431526-67431526 on Build GRCh38
rs983251408 Chr.16: 67431291-67431291 on Build GRCh38
rs72649092 Chr.16: 67431563-67431563 on Build GRCh38
rs932698676 Chr.16: 67431528-67431528 on Build GRCh38
rs965322780 Chr.16: 67431470-67431470 on Build GRCh38
rs1030165558 Chr.16: 67431653-67431653 on Build GRCh38
rs56106309 Chr.16: 67431550-67431550 on Build GRCh38
rs761552080 Chr.16: 67431439-67431439 on Build GRCh38
rs1053970729 Chr.16: 67431541-67431541 on Build GRCh38
rs978854602 Chr.16: 67431536-67431536 on Build GRCh38
rs975967480 Chr.16: 67431276-67431276 on Build GRCh38
rs781039840 Chr.16: 67431336-67431382 on Build GRCh38
rs921110870 Chr.16: 67431486-67431486 on Build GRCh38
rs906047343 Chr.16: 67431672-67431672 on Build GRCh38
rs759646288 Chr.16: 67431317-67431317 on Build GRCh38
rs776852403 Chr.16: 67431290-67431290 on Build GRCh38
rs756093283 Chr.16: 67431527-67431527 on Build GRCh38
rs45562134 Chr.16: 67431425-67431425 on Build GRCh38
rs766238938 Chr.16: 67431542-67431542 on Build GRCh38
rs761242831 Chr.16: 67431237-67431237 on Build GRCh38
rs1017675653 Chr.16: 67431213-67431214 on Build GRCh38
rs934659194 Chr.16: 67431562-67431562 on Build GRCh38
rs1044936846 Chr.16: 67431650-67431650 on Build GRCh38
rs995445465 Chr.16: 67431677-67431677 on Build GRCh38
rs553925830 Chr.16: 67431622-67431622 on Build GRCh38
rs771282097 Chr.16: 67431275-67431275 on Build GRCh38
rs989804432 Chr.16: 67431204-67431204 on Build GRCh38
rs794726684 Chr.16: 67431325-67431326 on Build GRCh38
rs1048127921 Chr.16: 67431681-67431681 on Build GRCh38
rs1053081160 Chr.16: 67431594-67431594 on Build GRCh38
rs895118361 Chr.16: 67431381-67431381 on Build GRCh38
rs896036700 Chr.16: 67431623-67431623 on Build GRCh38
rs72649093 Chr.16: 67431619-67431619 on Build GRCh38
rs902772869 Chr.16: 67431485-67431485 on Build GRCh38
rs1017393828 Chr.16: 67431606-67431606 on Build GRCh38
rs753378107 Chr.16: 67431548-67431548 on Build GRCh38
rs568066529 Chr.16: 67431374-67431374 on Build GRCh38
rs1046652532 Chr.16: 67431449-67431449 on Build GRCh38
rs556122396 Chr.16: 67431300-67431300 on Build GRCh38
rs767338144 Chr.16: 67431484-67431484 on Build GRCh38

View Full Product Details