Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50015463-50015974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU2
Entrez Gene ID
8847
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
Gene Aliases
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
UniGene
Hs.547964
Gene Chromosome Location
Chr.13: 49982552-50125541 on Build GRCh38
Transcripts
NR_002612.1 NM_173605.1 NM_199464.2 NM_001007278.2 NM_005798.4 NM_052811.3 NM_213590.2

Gene Symbol
KCNRG
Entrez Gene ID
283518
Gene Name
potassium channel regulator
Gene Aliases
DLTET
UniGene
Hs.660161
Gene Chromosome Location
Chr.13: 50015254-50020922 on Build GRCh38
Transcripts
NR_002612.1 NM_173605.1 NM_199464.2 NM_001007278.2 NM_005798.4 NM_052811.3 NM_213590.2

Gene Symbol
TRIM13
Entrez Gene ID
10206
Gene Name
tripartite motif containing 13
Gene Aliases
CAR, DLEU5, LEU5, RFP2, RNF77
UniGene
Hs.436922
Gene Chromosome Location
Chr.13: 49997007-50018467 on Build GRCh38
Transcripts
NR_002612.1 NM_173605.1 NM_199464.2 NM_001007278.2 NM_005798.4 NM_052811.3 NM_213590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317504 c.65C>G p.T22R Chr.13: 50015558-50015558 on Build GRCh38
303884 c.113G>A p.R38K Chr.13: 50015606-50015606 on Build GRCh38
5780309 c.331G>A p.E111K Chr.13: 50015824-50015824 on Build GRCh38
2151593 c.33G>C p.V11V Chr.13: 50015526-50015526 on Build GRCh38
3469218 c.285T>A p.V95V Chr.13: 50015778-50015778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956426040 Chr.13: 50015813-50015813 on Build GRCh38
rs773920365 Chr.13: 50015932-50015932 on Build GRCh38
rs201321734 Chr.13: 50015701-50015701 on Build GRCh38
rs370829504 Chr.13: 50015497-50015497 on Build GRCh38
rs762253211 Chr.13: 50015539-50015539 on Build GRCh38
rs775187503 Chr.13: 50015782-50015782 on Build GRCh38
rs12583744 Chr.13: 50015735-50015735 on Build GRCh38
rs1040510271 Chr.13: 50015637-50015637 on Build GRCh38
rs779637484 Chr.13: 50015486-50015486 on Build GRCh38
rs752992853 Chr.13: 50015830-50015830 on Build GRCh38
rs374886526 Chr.13: 50015611-50015611 on Build GRCh38
rs780297114 Chr.13: 50015593-50015593 on Build GRCh38
rs191718855 Chr.13: 50015940-50015940 on Build GRCh38
rs779090101 Chr.13: 50015765-50015765 on Build GRCh38
rs927563777 Chr.13: 50015903-50015903 on Build GRCh38
rs749546546 Chr.13: 50015606-50015606 on Build GRCh38
rs774378163 Chr.13: 50015538-50015538 on Build GRCh38
rs775679320 Chr.13: 50015912-50015912 on Build GRCh38
rs771505850 Chr.13: 50015774-50015774 on Build GRCh38
rs367947576 Chr.13: 50015680-50015680 on Build GRCh38
rs768695076 Chr.13: 50015525-50015525 on Build GRCh38
rs765481496 Chr.13: 50015829-50015829 on Build GRCh38
rs138037488 Chr.13: 50015842-50015842 on Build GRCh38
rs761085252 Chr.13: 50015558-50015558 on Build GRCh38
rs751370278 Chr.13: 50015590-50015590 on Build GRCh38
rs371471440 Chr.13: 50015522-50015522 on Build GRCh38
rs781761422 Chr.13: 50015871-50015871 on Build GRCh38
rs868162881 Chr.13: 50015904-50015905 on Build GRCh38
rs762412533 Chr.13: 50015792-50015792 on Build GRCh38
rs765080074 Chr.13: 50015497-50015503 on Build GRCh38
rs757905746 Chr.13: 50015862-50015862 on Build GRCh38
rs199969189 Chr.13: 50015942-50015942 on Build GRCh38
rs748222155 Chr.13: 50015766-50015766 on Build GRCh38
rs777085665 Chr.13: 50015657-50015657 on Build GRCh38
rs760983832 Chr.13: 50015634-50015634 on Build GRCh38
rs142559163 Chr.13: 50015939-50015939 on Build GRCh38
rs757054444 Chr.13: 50015591-50015591 on Build GRCh38
rs146042869 Chr.13: 50015943-50015943 on Build GRCh38
rs928034303 Chr.13: 50015924-50015924 on Build GRCh38
rs1016862593 Chr.13: 50015885-50015885 on Build GRCh38
rs754704579 Chr.13: 50015578-50015578 on Build GRCh38
rs750987719 Chr.13: 50015797-50015797 on Build GRCh38
rs200474686 Chr.13: 50015768-50015768 on Build GRCh38
rs746284967 Chr.13: 50015874-50015874 on Build GRCh38
rs764861264 Chr.13: 50015571-50015571 on Build GRCh38
rs201807354 Chr.13: 50015759-50015759 on Build GRCh38
rs767380049 Chr.13: 50015811-50015811 on Build GRCh38
rs182933979 Chr.13: 50015543-50015543 on Build GRCh38
rs765814274 Chr.13: 50015726-50015727 on Build GRCh38
rs750272280 Chr.13: 50015815-50015815 on Build GRCh38
rs953086075 Chr.13: 50015496-50015496 on Build GRCh38
rs751486313 Chr.13: 50015726-50015726 on Build GRCh38
rs143306362 Chr.13: 50015582-50015582 on Build GRCh38
rs748933939 Chr.13: 50015548-50015548 on Build GRCh38
rs765993786 Chr.13: 50015559-50015559 on Build GRCh38
rs768138543 Chr.13: 50015919-50015919 on Build GRCh38
rs567668233 Chr.13: 50015803-50015803 on Build GRCh38
rs750165798 Chr.13: 50015733-50015733 on Build GRCh38
rs754635280 Chr.13: 50015568-50015568 on Build GRCh38
rs757105415 Chr.13: 50015729-50015729 on Build GRCh38
rs778981728 Chr.13: 50015624-50015624 on Build GRCh38
rs759974304 Chr.13: 50015776-50015776 on Build GRCh38
rs758301144 Chr.13: 50015681-50015682 on Build GRCh38
rs780991807 Chr.13: 50015730-50015730 on Build GRCh38
rs763989101 Chr.13: 50015714-50015714 on Build GRCh38
rs763692832 Chr.13: 50015796-50015796 on Build GRCh38
rs748251688 Chr.13: 50015630-50015630 on Build GRCh38
rs980901366 Chr.13: 50015920-50015920 on Build GRCh38
rs140212682 Chr.13: 50015626-50015626 on Build GRCh38
rs750353649 Chr.13: 50015650-50015651 on Build GRCh38
rs758499261 Chr.13: 50015832-50015832 on Build GRCh38
rs773100412 Chr.13: 50015941-50015941 on Build GRCh38
rs771452564 Chr.13: 50015650-50015650 on Build GRCh38
rs769979405 Chr.13: 50015502-50015502 on Build GRCh38
rs901382700 Chr.13: 50015669-50015669 on Build GRCh38
rs770254671 Chr.13: 50015881-50015881 on Build GRCh38
rs1036584400 Chr.13: 50015672-50015672 on Build GRCh38
rs778207725 Chr.13: 50015937-50015937 on Build GRCh38
rs748995629 Chr.13: 50015918-50015918 on Build GRCh38
rs780873281 Chr.13: 50015883-50015884 on Build GRCh38
rs143015161 Chr.13: 50015494-50015494 on Build GRCh38
rs773512879 Chr.13: 50015545-50015545 on Build GRCh38
rs764916467 Chr.13: 50015690-50015690 on Build GRCh38
rs753517654 Chr.13: 50015946-50015946 on Build GRCh38
rs576707522 Chr.13: 50015793-50015793 on Build GRCh38
rs775677947 Chr.13: 50015518-50015518 on Build GRCh38
rs754472352 Chr.13: 50015843-50015843 on Build GRCh38
rs780921066 Chr.13: 50015594-50015594 on Build GRCh38
rs1052401170 Chr.13: 50015550-50015550 on Build GRCh38
rs150313045 Chr.13: 50015632-50015632 on Build GRCh38
rs200397876 Chr.13: 50015816-50015816 on Build GRCh38
rs746993779 Chr.13: 50015843-50015843 on Build GRCh38
rs200994762 Chr.13: 50015707-50015707 on Build GRCh38
rs34905525 Chr.13: 50015904-50015905 on Build GRCh38
rs773744562 Chr.13: 50015633-50015633 on Build GRCh38
rs769629288 Chr.13: 50015781-50015781 on Build GRCh38
rs375169401 Chr.13: 50015767-50015767 on Build GRCh38
rs758273779 Chr.13: 50015581-50015581 on Build GRCh38
rs866665695 Chr.13: 50015749-50015749 on Build GRCh38
rs777921805 Chr.13: 50015836-50015836 on Build GRCh38
rs375378114 Chr.13: 50015615-50015615 on Build GRCh38
rs777138806 Chr.13: 50015775-50015775 on Build GRCh38

View Full Product Details