Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45641592-45642119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR1-IT1
Entrez Gene ID
147081
Gene Name
CRHR1 intronic transcript 1
Gene Aliases
C17orf69
UniGene
Hs.128813
Gene Chromosome Location
Chr.17: 45638975-45646229 on Build GRCh38
Transcripts
NR_026905.2 NR_026906.2 NM_001256299.2 NM_001303016.1

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 45620329-45835828 on Build GRCh38
Transcripts
NR_026905.2 NR_026906.2 NM_001256299.2 NM_001303016.1

SNP Detail

SNP ID Chromosome Location
rs902836012 Chr.17: 45641989-45641989 on Build GRCh38
rs393152 Chr.17: 45641777-45641777 on Build GRCh38
rs1012736581 Chr.17: 45642036-45642036 on Build GRCh38
rs534788748 Chr.17: 45641642-45641642 on Build GRCh38
rs1040444139 Chr.17: 45641945-45641945 on Build GRCh38
rs753751212 Chr.17: 45641886-45641886 on Build GRCh38
rs981841890 Chr.17: 45641784-45641784 on Build GRCh38
rs1040151540 Chr.17: 45641616-45641616 on Build GRCh38
rs750968607 Chr.17: 45641849-45641849 on Build GRCh38
rs537746446 Chr.17: 45641920-45641920 on Build GRCh38
rs767832743 Chr.17: 45641781-45641781 on Build GRCh38
rs752006670 Chr.17: 45641768-45641768 on Build GRCh38
rs1022865101 Chr.17: 45641751-45641751 on Build GRCh38
rs944424372 Chr.17: 45641928-45641928 on Build GRCh38
rs546397187 Chr.17: 45641644-45641644 on Build GRCh38
rs1048815141 Chr.17: 45641990-45641990 on Build GRCh38
rs763378482 Chr.17: 45641713-45641713 on Build GRCh38
rs915834418 Chr.17: 45641956-45641956 on Build GRCh38
rs1045773068 Chr.17: 45641691-45641691 on Build GRCh38
rs997539422 Chr.17: 45641787-45641787 on Build GRCh38
rs930579827 Chr.17: 45641639-45641639 on Build GRCh38
rs760217002 Chr.17: 45641673-45641673 on Build GRCh38
rs912804274 Chr.17: 45641898-45641898 on Build GRCh38
rs559538184 Chr.17: 45641865-45641865 on Build GRCh38
rs1054123795 Chr.17: 45642054-45642054 on Build GRCh38
rs775014490 Chr.17: 45641675-45641675 on Build GRCh38
rs559155277 Chr.17: 45642058-45642058 on Build GRCh38
rs934437191 Chr.17: 45642035-45642035 on Build GRCh38
rs535472890 Chr.17: 45641831-45641831 on Build GRCh38
rs1028874481 Chr.17: 45642095-45642095 on Build GRCh38
rs764501316 Chr.17: 45641754-45641754 on Build GRCh38
rs529906392 Chr.17: 45642034-45642034 on Build GRCh38
rs1032095787 Chr.17: 45642088-45642088 on Build GRCh38
rs775995655 Chr.17: 45641682-45641682 on Build GRCh38
rs374320721 Chr.17: 45641994-45641994 on Build GRCh38
rs774653321 Chr.17: 45641974-45641974 on Build GRCh38
rs1045388017 Chr.17: 45641971-45641971 on Build GRCh38
rs929057722 Chr.17: 45641978-45641978 on Build GRCh38
rs907111697 Chr.17: 45642075-45642075 on Build GRCh38
rs747750991 Chr.17: 45641622-45641622 on Build GRCh38
rs949975676 Chr.17: 45641969-45641969 on Build GRCh38
rs756216031 Chr.17: 45641838-45641838 on Build GRCh38
rs1044367976 Chr.17: 45642069-45642069 on Build GRCh38
rs894167439 Chr.17: 45641674-45641674 on Build GRCh38
rs920455697 Chr.17: 45641907-45641907 on Build GRCh38
rs990160544 Chr.17: 45641828-45641828 on Build GRCh38
rs556991329 Chr.17: 45641858-45641858 on Build GRCh38
rs971903587 Chr.17: 45641614-45641614 on Build GRCh38
rs762146361 Chr.17: 45642086-45642086 on Build GRCh38
rs1031795674 Chr.17: 45641793-45641793 on Build GRCh38
rs771681919 Chr.17: 45641634-45641634 on Build GRCh38
rs974135672 Chr.17: 45641877-45641878 on Build GRCh38
rs769036750 Chr.17: 45641824-45641824 on Build GRCh38
rs1012851628 Chr.17: 45641741-45641741 on Build GRCh38
rs914757465 Chr.17: 45641834-45641834 on Build GRCh38
rs760893774 Chr.17: 45641795-45641796 on Build GRCh38
rs372204179 Chr.17: 45641819-45641819 on Build GRCh38
rs887628699 Chr.17: 45642024-45642024 on Build GRCh38
rs954190024 Chr.17: 45641863-45641863 on Build GRCh38
rs986825554 Chr.17: 45641847-45641847 on Build GRCh38
rs930486728 Chr.17: 45641915-45641915 on Build GRCh38

View Full Product Details