Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74151890-74152409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLA3-AS1
Entrez Gene ID
100507171
Gene Name
BOLA3 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.503348
Gene Chromosome Location
Chr.2: 74147981-74152389 on Build GRCh38
Transcripts
NR_045634.1 NR_045636.1

SNP Detail

SNP ID Chromosome Location
rs572811259 Chr.2: 74152149-74152149 on Build GRCh38
rs750868000 Chr.2: 74152048-74152048 on Build GRCh38
rs1028672779 Chr.2: 74152359-74152359 on Build GRCh38
rs1003875990 Chr.2: 74152303-74152303 on Build GRCh38
rs917699115 Chr.2: 74152069-74152082 on Build GRCh38
rs1055993630 Chr.2: 74151929-74151929 on Build GRCh38
rs758086831 Chr.2: 74152042-74152043 on Build GRCh38
rs372205686 Chr.2: 74152152-74152152 on Build GRCh38
rs987491395 Chr.2: 74152316-74152316 on Build GRCh38
rs928868293 Chr.2: 74152141-74152141 on Build GRCh38
rs533825726 Chr.2: 74152177-74152177 on Build GRCh38
rs1026746061 Chr.2: 74151975-74151975 on Build GRCh38
rs145470459 Chr.2: 74152375-74152376 on Build GRCh38
rs187089479 Chr.2: 74152376-74152376 on Build GRCh38
rs1051757355 Chr.2: 74151994-74151994 on Build GRCh38
rs778959544 Chr.2: 74152332-74152332 on Build GRCh38
rs996753737 Chr.2: 74152358-74152358 on Build GRCh38
rs1054056322 Chr.2: 74152035-74152035 on Build GRCh38
rs922043411 Chr.2: 74152291-74152291 on Build GRCh38
rs935258135 Chr.2: 74152380-74152380 on Build GRCh38
rs938967572 Chr.2: 74152169-74152169 on Build GRCh38
rs74591230 Chr.2: 74152119-74152119 on Build GRCh38
rs140210622 Chr.2: 74152334-74152334 on Build GRCh38
rs990165523 Chr.2: 74152092-74152092 on Build GRCh38
rs902466320 Chr.2: 74152022-74152022 on Build GRCh38
rs1056466321 Chr.2: 74152187-74152187 on Build GRCh38
rs1048136424 Chr.2: 74152281-74152281 on Build GRCh38
rs143034661 Chr.2: 74151938-74151938 on Build GRCh38
rs1053685321 Chr.2: 74152383-74152383 on Build GRCh38
rs974828766 Chr.2: 74152272-74152272 on Build GRCh38
rs767498539 Chr.2: 74152083-74152083 on Build GRCh38
rs894613463 Chr.2: 74152193-74152193 on Build GRCh38
rs983710055 Chr.2: 74152199-74152199 on Build GRCh38
rs906217161 Chr.2: 74152143-74152143 on Build GRCh38
rs535845459 Chr.2: 74152378-74152378 on Build GRCh38
rs933386792 Chr.2: 74152304-74152304 on Build GRCh38
rs752374741 Chr.2: 74152093-74152093 on Build GRCh38
rs371399933 Chr.2: 74152063-74152063 on Build GRCh38
rs182581625 Chr.2: 74152064-74152064 on Build GRCh38
rs914659606 Chr.2: 74152098-74152098 on Build GRCh38
rs951058895 Chr.2: 74152192-74152192 on Build GRCh38
rs950995443 Chr.2: 74152024-74152024 on Build GRCh38
rs753963812 Chr.2: 74152085-74152085 on Build GRCh38
rs933270725 Chr.2: 74151983-74151983 on Build GRCh38
rs779086697 Chr.2: 74152137-74152137 on Build GRCh38
rs756450534 Chr.2: 74151925-74151925 on Build GRCh38
rs1019305542 Chr.2: 74152309-74152309 on Build GRCh38
rs574306610 Chr.2: 74152288-74152288 on Build GRCh38
rs963518532 Chr.2: 74152234-74152234 on Build GRCh38
rs80035870 Chr.2: 74152203-74152203 on Build GRCh38
rs550829186 Chr.2: 74151923-74151923 on Build GRCh38
rs935242380 Chr.2: 74152031-74152031 on Build GRCh38
rs755937911 Chr.2: 74152270-74152271 on Build GRCh38
rs552061991 Chr.2: 74151971-74151971 on Build GRCh38

View Full Product Details