Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29010591-29011133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
UniGene
Hs.233389
Gene Chromosome Location
Chr.7: 28995630-29146537 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1 NR_038965.1

Gene Symbol
LOC100506497
Entrez Gene ID
100506497
Gene Name
uncharacterized LOC100506497
Gene Aliases
-
UniGene
Hs.586328
Gene Chromosome Location
Chr.7: 28979967-29013369 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1 NR_038965.1

SNP Detail

SNP ID Chromosome Location
rs565076560 Chr.7: 29010894-29010894 on Build GRCh38
rs773786337 Chr.7: 29011012-29011012 on Build GRCh38
rs562687280 Chr.7: 29011078-29011078 on Build GRCh38
rs1045631340 Chr.7: 29010674-29010674 on Build GRCh38
rs975330464 Chr.7: 29010951-29010951 on Build GRCh38
rs16874739 Chr.7: 29010630-29010630 on Build GRCh38
rs907089980 Chr.7: 29010682-29010682 on Build GRCh38
rs951021331 Chr.7: 29010851-29010851 on Build GRCh38
rs946053163 Chr.7: 29011109-29011109 on Build GRCh38
rs962804383 Chr.7: 29010778-29010778 on Build GRCh38
rs966793011 Chr.7: 29010640-29010640 on Build GRCh38
rs183131854 Chr.7: 29010864-29010864 on Build GRCh38
rs912397160 Chr.7: 29010763-29010763 on Build GRCh38
rs2108382 Chr.7: 29011051-29011051 on Build GRCh38
rs995250030 Chr.7: 29010786-29010786 on Build GRCh38
rs572586353 Chr.7: 29010806-29010806 on Build GRCh38
rs986433823 Chr.7: 29010881-29010881 on Build GRCh38
rs913284518 Chr.7: 29011085-29011085 on Build GRCh38
rs966261574 Chr.7: 29010949-29010949 on Build GRCh38
rs931286323 Chr.7: 29010947-29010947 on Build GRCh38
rs954021195 Chr.7: 29010724-29010724 on Build GRCh38
rs532267022 Chr.7: 29010964-29010964 on Build GRCh38
rs1010142366 Chr.7: 29010667-29010667 on Build GRCh38
rs554291311 Chr.7: 29010727-29010727 on Build GRCh38
rs574771547 Chr.7: 29010636-29010636 on Build GRCh38
rs912152079 Chr.7: 29010905-29010905 on Build GRCh38
rs1034156110 Chr.7: 29010718-29010718 on Build GRCh38
rs972803046 Chr.7: 29010832-29010832 on Build GRCh38
rs745464582 Chr.7: 29010651-29010651 on Build GRCh38
rs658474 Chr.7: 29010706-29010706 on Build GRCh38
rs763271268 Chr.7: 29010915-29010915 on Build GRCh38

View Full Product Details