Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 12145441-12145952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66D
Entrez Gene ID
100132923
Gene Name
family with sequence similarity 66 member D
Gene Aliases
-
UniGene
Hs.546711
Gene Chromosome Location
Chr.8: 12115782-12151189 on Build GRCh38
Transcripts
NR_027425.1

SNP Detail

SNP ID Chromosome Location
rs571044152 Chr.8: 12145867-12145867 on Build GRCh38
rs10101320 Chr.8: 12145730-12145730 on Build GRCh38
rs527634811 Chr.8: 12145762-12145762 on Build GRCh38
rs539214726 Chr.8: 12145926-12145926 on Build GRCh38
rs892231752 Chr.8: 12145714-12145714 on Build GRCh38
rs1045001303 Chr.8: 12145618-12145618 on Build GRCh38
rs3882689 Chr.8: 12145882-12145882 on Build GRCh38
rs533137261 Chr.8: 12145681-12145681 on Build GRCh38
rs952910173 Chr.8: 12145862-12145862 on Build GRCh38
rs537513221 Chr.8: 12145820-12145820 on Build GRCh38
rs1036607197 Chr.8: 12145534-12145534 on Build GRCh38
rs566931691 Chr.8: 12145796-12145796 on Build GRCh38
rs4118267 Chr.8: 12145648-12145648 on Build GRCh38
rs543952747 Chr.8: 12145676-12145676 on Build GRCh38
rs4118265 Chr.8: 12145738-12145738 on Build GRCh38
rs180750357 Chr.8: 12145718-12145718 on Build GRCh38
rs1053355880 Chr.8: 12145693-12145693 on Build GRCh38
rs1028607509 Chr.8: 12145856-12145856 on Build GRCh38
rs543006151 Chr.8: 12145535-12145535 on Build GRCh38
rs965610461 Chr.8: 12145758-12145758 on Build GRCh38
rs949233344 Chr.8: 12145576-12145576 on Build GRCh38
rs996998032 Chr.8: 12145825-12145825 on Build GRCh38
rs984137275 Chr.8: 12145886-12145886 on Build GRCh38
rs1020212020 Chr.8: 12145749-12145749 on Build GRCh38
rs1000516337 Chr.8: 12145675-12145675 on Build GRCh38
rs562642544 Chr.8: 12145679-12145679 on Build GRCh38
rs561130779 Chr.8: 12145540-12145540 on Build GRCh38
rs572691521 Chr.8: 12145479-12145479 on Build GRCh38
rs779430776 Chr.8: 12145776-12145780 on Build GRCh38
rs60099293 Chr.8: 12145861-12145862 on Build GRCh38
rs773150436 Chr.8: 12145819-12145819 on Build GRCh38
rs908638789 Chr.8: 12145515-12145515 on Build GRCh38
rs554431986 Chr.8: 12145471-12145471 on Build GRCh38
rs548916192 Chr.8: 12145769-12145769 on Build GRCh38
rs905101360 Chr.8: 12145661-12145661 on Build GRCh38
rs940095729 Chr.8: 12145529-12145529 on Build GRCh38
rs772121004 Chr.8: 12145517-12145517 on Build GRCh38
rs200785343 Chr.8: 12145859-12145860 on Build GRCh38
rs4118266 Chr.8: 12145696-12145696 on Build GRCh38
rs549444402 Chr.8: 12145848-12145848 on Build GRCh38

View Full Product Details