Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39402691-39403223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
UniGene
Hs.369765
Gene Chromosome Location
Chr.8: 39314663-39417226 on Build GRCh38
Transcripts
NR_001448.1

SNP Detail

SNP ID Chromosome Location
rs549844226 Chr.8: 39403065-39403065 on Build GRCh38
rs577451683 Chr.8: 39403111-39403112 on Build GRCh38
rs899528105 Chr.8: 39402775-39402775 on Build GRCh38
rs991016388 Chr.8: 39403147-39403147 on Build GRCh38
rs7001393 Chr.8: 39403008-39403008 on Build GRCh38
rs189325995 Chr.8: 39403041-39403041 on Build GRCh38
rs7000399 Chr.8: 39403134-39403134 on Build GRCh38
rs565765455 Chr.8: 39402980-39402980 on Build GRCh38
rs778216063 Chr.8: 39403091-39403091 on Build GRCh38
rs1008412126 Chr.8: 39402937-39402937 on Build GRCh38
rs372682261 Chr.8: 39403073-39403073 on Build GRCh38
rs943974181 Chr.8: 39402746-39402746 on Build GRCh38
rs75920401 Chr.8: 39402722-39402722 on Build GRCh38
rs748080023 Chr.8: 39403131-39403131 on Build GRCh38
rs868192741 Chr.8: 39403151-39403151 on Build GRCh38
rs529968009 Chr.8: 39403050-39403050 on Build GRCh38
rs138674965 Chr.8: 39402971-39402971 on Build GRCh38
rs11448944 Chr.8: 39403146-39403147 on Build GRCh38
rs79925196 Chr.8: 39402898-39402898 on Build GRCh38
rs932277229 Chr.8: 39402810-39402810 on Build GRCh38
rs78933172 Chr.8: 39402896-39402896 on Build GRCh38
rs888155451 Chr.8: 39402912-39402912 on Build GRCh38
rs570013192 Chr.8: 39403127-39403127 on Build GRCh38
rs1006398199 Chr.8: 39402961-39402961 on Build GRCh38
rs375896242 Chr.8: 39403133-39403134 on Build GRCh38
rs375600973 Chr.8: 39403125-39403125 on Build GRCh38
rs547554855 Chr.8: 39402872-39402872 on Build GRCh38
rs569721676 Chr.8: 39402865-39402865 on Build GRCh38
rs538940186 Chr.8: 39403151-39403152 on Build GRCh38
rs10652191 Chr.8: 39403162-39403163 on Build GRCh38
rs376664137 Chr.8: 39403044-39403044 on Build GRCh38
rs891331547 Chr.8: 39402894-39402894 on Build GRCh38
rs79462908 Chr.8: 39403133-39403133 on Build GRCh38
rs1037576587 Chr.8: 39402981-39402981 on Build GRCh38
rs1040964817 Chr.8: 39402763-39402763 on Build GRCh38

View Full Product Details