Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16619796-16620344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCCP2
Entrez Gene ID
84809
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 2
Gene Aliases
CROCCL1
UniGene
Hs.631865
Gene Chromosome Location
Chr.1: 16618256-16630906 on Build GRCh38
Transcripts
NR_026752.1

SNP Detail

SNP ID Chromosome Location
rs1759234 Chr.1: 16620167-16620167 on Build GRCh38
rs399721 Chr.1: 16620245-16620245 on Build GRCh38
rs776009683 Chr.1: 16620026-16620026 on Build GRCh38
rs550499577 Chr.1: 16620324-16620324 on Build GRCh38
rs868019397 Chr.1: 16620226-16620226 on Build GRCh38
rs879101891 Chr.1: 16619857-16619857 on Build GRCh38
rs367013 Chr.1: 16619901-16619901 on Build GRCh38
rs11586784 Chr.1: 16619841-16619841 on Build GRCh38
rs201265716 Chr.1: 16620257-16620257 on Build GRCh38
rs564001968 Chr.1: 16619966-16619966 on Build GRCh38
rs10796418 Chr.1: 16619912-16619912 on Build GRCh38
rs1011876795 Chr.1: 16619929-16619929 on Build GRCh38
rs572479688 Chr.1: 16619922-16619922 on Build GRCh38
rs60132265 Chr.1: 16620182-16620182 on Build GRCh38
rs373008872 Chr.1: 16620250-16620250 on Build GRCh38
rs12086176 Chr.1: 16619881-16619881 on Build GRCh38
rs2262202 Chr.1: 16619942-16619942 on Build GRCh38
rs879032110 Chr.1: 16620033-16620033 on Build GRCh38
rs583343 Chr.1: 16620135-16620135 on Build GRCh38
rs59452670 Chr.1: 16620017-16620017 on Build GRCh38
rs375097 Chr.1: 16620228-16620228 on Build GRCh38
rs11260836 Chr.1: 16620299-16620299 on Build GRCh38
rs546357136 Chr.1: 16620078-16620078 on Build GRCh38
rs367060 Chr.1: 16619939-16619939 on Build GRCh38
rs878952169 Chr.1: 16619998-16619998 on Build GRCh38
rs535051637 Chr.1: 16619935-16619935 on Build GRCh38
rs569514556 Chr.1: 16620076-16620077 on Build GRCh38
rs577412340 Chr.1: 16620044-16620044 on Build GRCh38
rs425424 Chr.1: 16620254-16620254 on Build GRCh38
rs1762938 Chr.1: 16620067-16620067 on Build GRCh38
rs437905 Chr.1: 16620040-16620040 on Build GRCh38
rs28392876 Chr.1: 16619943-16619943 on Build GRCh38

View Full Product Details