Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100170424-100170969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2P5
Entrez Gene ID
55592
Gene Name
golgin A2 pseudogene 5
Gene Aliases
GOLGA2B, GOLGA2L1
UniGene
Hs.524660
Gene Chromosome Location
Chr.12: 100156391-100173343 on Build GRCh38
Transcripts
NR_024261.2 NR_036632.1

SNP Detail

SNP ID Chromosome Location
rs186991123 Chr.12: 100170900-100170900 on Build GRCh38
rs918570721 Chr.12: 100170588-100170588 on Build GRCh38
rs17041314 Chr.12: 100170847-100170847 on Build GRCh38
rs1026552333 Chr.12: 100170643-100170643 on Build GRCh38
rs922494924 Chr.12: 100170661-100170661 on Build GRCh38
rs976091819 Chr.12: 100170599-100170599 on Build GRCh38
rs113138819 Chr.12: 100170487-100170487 on Build GRCh38
rs144510970 Chr.12: 100170794-100170794 on Build GRCh38
rs903121908 Chr.12: 100170812-100170812 on Build GRCh38
rs138569083 Chr.12: 100170646-100170646 on Build GRCh38
rs970946058 Chr.12: 100170853-100170853 on Build GRCh38
rs545215493 Chr.12: 100170592-100170592 on Build GRCh38
rs984007799 Chr.12: 100170614-100170614 on Build GRCh38
rs951325292 Chr.12: 100170920-100170920 on Build GRCh38
rs763465719 Chr.12: 100170559-100170559 on Build GRCh38
rs951377265 Chr.12: 100170601-100170601 on Build GRCh38
rs769137303 Chr.12: 100170486-100170486 on Build GRCh38
rs574048731 Chr.12: 100170925-100170925 on Build GRCh38
rs878954385 Chr.12: 100170705-100170705 on Build GRCh38
rs535053558 Chr.12: 100170939-100170939 on Build GRCh38
rs915659400 Chr.12: 100170761-100170761 on Build GRCh38
rs539179732 Chr.12: 100170915-100170915 on Build GRCh38
rs933722192 Chr.12: 100170679-100170679 on Build GRCh38
rs941125717 Chr.12: 100170717-100170717 on Build GRCh38
rs1054301259 Chr.12: 100170822-100170822 on Build GRCh38
rs942507860 Chr.12: 100170653-100170653 on Build GRCh38
rs547671022 Chr.12: 100170790-100170790 on Build GRCh38
rs1008023391 Chr.12: 100170499-100170499 on Build GRCh38
rs570165141 Chr.12: 100170818-100170818 on Build GRCh38
rs982223702 Chr.12: 100170554-100170554 on Build GRCh38
rs764225962 Chr.12: 100170782-100170782 on Build GRCh38
rs551993732 Chr.12: 100170809-100170809 on Build GRCh38
rs529528457 Chr.12: 100170552-100170552 on Build GRCh38
rs560498441 Chr.12: 100170728-100170728 on Build GRCh38
rs770028590 Chr.12: 100170744-100170744 on Build GRCh38
rs114400049 Chr.12: 100170760-100170760 on Build GRCh38
rs958200151 Chr.12: 100170505-100170505 on Build GRCh38
rs149294106 Chr.12: 100170657-100170657 on Build GRCh38
rs563232351 Chr.12: 100170645-100170645 on Build GRCh38
rs750947920 Chr.12: 100170942-100170942 on Build GRCh38
rs776997014 Chr.12: 100170553-100170553 on Build GRCh38
rs566061940 Chr.12: 100170791-100170791 on Build GRCh38
rs577746764 Chr.12: 100170541-100170541 on Build GRCh38
rs1012536433 Chr.12: 100170837-100170837 on Build GRCh38
rs867846905 Chr.12: 100170513-100170513 on Build GRCh38
rs901914728 Chr.12: 100170919-100170919 on Build GRCh38
rs999709724 Chr.12: 100170457-100170457 on Build GRCh38
rs775754065 Chr.12: 100170454-100170454 on Build GRCh38
rs1023846084 Chr.12: 100170514-100170514 on Build GRCh38

View Full Product Details