Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44538961-44539521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM22P
Entrez Gene ID
388550
Gene Name
carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
Gene Aliases
-
UniGene
Hs.446909
Gene Chromosome Location
Chr.19: 44537058-44556917 on Build GRCh38
Transcripts
NR_027754.2

SNP Detail

SNP ID Chromosome Location
rs907159763 Chr.19: 44539378-44539378 on Build GRCh38
rs996909456 Chr.19: 44539424-44539424 on Build GRCh38
rs986975607 Chr.19: 44539235-44539235 on Build GRCh38
rs543829400 Chr.19: 44539459-44539459 on Build GRCh38
rs937334678 Chr.19: 44539381-44539381 on Build GRCh38
rs907211334 Chr.19: 44539074-44539074 on Build GRCh38
rs191564592 Chr.19: 44539490-44539490 on Build GRCh38
rs975415650 Chr.19: 44538993-44538993 on Build GRCh38
rs191923553 Chr.19: 44539060-44539060 on Build GRCh38
rs189252373 Chr.19: 44539313-44539313 on Build GRCh38
rs148456954 Chr.19: 44539413-44539413 on Build GRCh38
rs1007791770 Chr.19: 44539117-44539117 on Build GRCh38
rs1039255520 Chr.19: 44539456-44539456 on Build GRCh38
rs766741974 Chr.19: 44539181-44539183 on Build GRCh38
rs898403100 Chr.19: 44539384-44539384 on Build GRCh38
rs766320728 Chr.19: 44539479-44539479 on Build GRCh38
rs547003083 Chr.19: 44539123-44539131 on Build GRCh38
rs116586024 Chr.19: 44539236-44539236 on Build GRCh38
rs945453409 Chr.19: 44539394-44539394 on Build GRCh38
rs926585164 Chr.19: 44539000-44539000 on Build GRCh38
rs111835475 Chr.19: 44539344-44539344 on Build GRCh38
rs144524749 Chr.19: 44539318-44539318 on Build GRCh38
rs964057951 Chr.19: 44539242-44539242 on Build GRCh38
rs899467056 Chr.19: 44539169-44539169 on Build GRCh38
rs113745414 Chr.19: 44539351-44539351 on Build GRCh38
rs563750822 Chr.19: 44539474-44539474 on Build GRCh38
rs930461 Chr.19: 44539272-44539272 on Build GRCh38
rs528956173 Chr.19: 44539485-44539485 on Build GRCh38
rs183746873 Chr.19: 44539478-44539478 on Build GRCh38
rs117638070 Chr.19: 44539498-44539498 on Build GRCh38
rs1017874661 Chr.19: 44539135-44539135 on Build GRCh38
rs780708663 Chr.19: 44539371-44539371 on Build GRCh38
rs576653661 Chr.19: 44539315-44539315 on Build GRCh38
rs756693792 Chr.19: 44538985-44538985 on Build GRCh38
rs891879246 Chr.19: 44539058-44539058 on Build GRCh38
rs142758410 Chr.19: 44538994-44538994 on Build GRCh38
rs1056902117 Chr.19: 44539053-44539053 on Build GRCh38
rs561372810 Chr.19: 44539412-44539412 on Build GRCh38
rs1054669949 Chr.19: 44539023-44539023 on Build GRCh38
rs1046859163 Chr.19: 44539386-44539386 on Build GRCh38
rs930462 Chr.19: 44539082-44539082 on Build GRCh38
rs774460387 Chr.19: 44539187-44539187 on Build GRCh38
rs537692861 Chr.19: 44539387-44539387 on Build GRCh38
rs1046240375 Chr.19: 44539362-44539362 on Build GRCh38
rs780649102 Chr.19: 44539063-44539063 on Build GRCh38
rs898455341 Chr.19: 44539140-44539140 on Build GRCh38
rs533626984 Chr.19: 44539094-44539094 on Build GRCh38
rs547106078 Chr.19: 44539198-44539198 on Build GRCh38
rs768815160 Chr.19: 44539102-44539102 on Build GRCh38
rs1016698705 Chr.19: 44539278-44539278 on Build GRCh38
rs1055832262 Chr.19: 44539383-44539383 on Build GRCh38
rs1034680233 Chr.19: 44539133-44539133 on Build GRCh38
rs936157259 Chr.19: 44539004-44539004 on Build GRCh38
rs115933723 Chr.19: 44539475-44539475 on Build GRCh38
rs963868180 Chr.19: 44539415-44539415 on Build GRCh38
rs992154732 Chr.19: 44539345-44539345 on Build GRCh38
rs186705285 Chr.19: 44539483-44539483 on Build GRCh38
rs541512360 Chr.19: 44539408-44539408 on Build GRCh38
rs951118002 Chr.19: 44539207-44539207 on Build GRCh38
rs995408979 Chr.19: 44539385-44539385 on Build GRCh38
rs939373684 Chr.19: 44539028-44539028 on Build GRCh38

View Full Product Details