Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130103728-130104286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86HP
Entrez Gene ID
729375
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.123633
Gene Chromosome Location
Chr.3: 130097782-130111433 on Build GRCh38
Transcripts
NR_024252.1

SNP Detail

SNP ID Chromosome Location
rs752414776 Chr.3: 130103849-130103849 on Build GRCh38
rs146467554 Chr.3: 130104042-130104042 on Build GRCh38
rs887782785 Chr.3: 130103778-130103778 on Build GRCh38
rs937489981 Chr.3: 130103871-130103871 on Build GRCh38
rs906315429 Chr.3: 130104226-130104226 on Build GRCh38
rs199587050 Chr.3: 130103962-130103962 on Build GRCh38
rs2624639 Chr.3: 130103796-130103796 on Build GRCh38
rs199758595 Chr.3: 130104258-130104258 on Build GRCh38
rs543006192 Chr.3: 130103830-130103833 on Build GRCh38
rs1050475637 Chr.3: 130104161-130104161 on Build GRCh38
rs909358812 Chr.3: 130104013-130104013 on Build GRCh38
rs768044547 Chr.3: 130104168-130104168 on Build GRCh38
rs527672453 Chr.3: 130103887-130103887 on Build GRCh38
rs1005359602 Chr.3: 130103894-130103894 on Build GRCh38
rs62282625 Chr.3: 130104060-130104060 on Build GRCh38
rs542150637 Chr.3: 130104061-130104061 on Build GRCh38
rs925025500 Chr.3: 130103823-130103823 on Build GRCh38
rs1004918294 Chr.3: 130103787-130103787 on Build GRCh38
rs571678762 Chr.3: 130104096-130104096 on Build GRCh38
rs1024759827 Chr.3: 130104215-130104215 on Build GRCh38
rs906009196 Chr.3: 130103755-130103755 on Build GRCh38
rs970442515 Chr.3: 130104070-130104070 on Build GRCh38
rs554850155 Chr.3: 130104178-130104178 on Build GRCh38
rs1001174736 Chr.3: 130104053-130104053 on Build GRCh38
rs965975887 Chr.3: 130103790-130103790 on Build GRCh38
rs559407136 Chr.3: 130103838-130103838 on Build GRCh38
rs9713346 Chr.3: 130103791-130103791 on Build GRCh38
rs923386139 Chr.3: 130104154-130104154 on Build GRCh38
rs185149635 Chr.3: 130103795-130103795 on Build GRCh38
rs550016046 Chr.3: 130103928-130103928 on Build GRCh38
rs567762674 Chr.3: 130103900-130103900 on Build GRCh38
rs186880350 Chr.3: 130104077-130104077 on Build GRCh38
rs1045903795 Chr.3: 130103753-130103753 on Build GRCh38
rs141535736 Chr.3: 130104141-130104141 on Build GRCh38
rs538791425 Chr.3: 130104081-130104081 on Build GRCh38
rs73866096 Chr.3: 130104157-130104157 on Build GRCh38
rs151186314 Chr.3: 130103981-130103981 on Build GRCh38
rs1032589732 Chr.3: 130103958-130103958 on Build GRCh38
rs927601384 Chr.3: 130104008-130104008 on Build GRCh38
rs7610964 Chr.3: 130104165-130104165 on Build GRCh38
rs113156717 Chr.3: 130104214-130104214 on Build GRCh38
rs889080882 Chr.3: 130104043-130104043 on Build GRCh38
rs887906832 Chr.3: 130103890-130103890 on Build GRCh38
rs185312246 Chr.3: 130104011-130104011 on Build GRCh38
rs1036392722 Chr.3: 130103908-130103908 on Build GRCh38
rs777361123 Chr.3: 130104039-130104039 on Build GRCh38
rs901847281 Chr.3: 130104032-130104032 on Build GRCh38
rs906041776 Chr.3: 130103869-130103869 on Build GRCh38
rs572683689 Chr.3: 130103967-130103967 on Build GRCh38
rs181218687 Chr.3: 130103949-130103949 on Build GRCh38
rs1036458364 Chr.3: 130104128-130104128 on Build GRCh38
rs576908198 Chr.3: 130104248-130104248 on Build GRCh38
rs183628134 Chr.3: 130104062-130104062 on Build GRCh38
rs558381538 Chr.3: 130104012-130104012 on Build GRCh38
rs1000742453 Chr.3: 130103957-130103957 on Build GRCh38
rs892306143 Chr.3: 130104181-130104181 on Build GRCh38
rs940800962 Chr.3: 130104126-130104126 on Build GRCh38
rs769205957 Chr.3: 130104224-130104224 on Build GRCh38
rs536668337 Chr.3: 130104234-130104234 on Build GRCh38
rs997453386 Chr.3: 130103940-130103940 on Build GRCh38
rs571778638 Chr.3: 130103939-130103939 on Build GRCh38
rs558596559 Chr.3: 130104238-130104238 on Build GRCh38
rs953344501 Chr.3: 130103950-130103950 on Build GRCh38
rs549441465 Chr.3: 130103895-130103895 on Build GRCh38
rs575771935 Chr.3: 130103788-130103788 on Build GRCh38
rs189612957 Chr.3: 130103921-130103921 on Build GRCh38
rs959015639 Chr.3: 130104084-130104084 on Build GRCh38
rs535834825 Chr.3: 130103978-130103978 on Build GRCh38
rs576939346 Chr.3: 130104027-130104027 on Build GRCh38
rs540905144 Chr.3: 130104036-130104036 on Build GRCh38
rs563318644 Chr.3: 130103833-130103833 on Build GRCh38
rs200605484 Chr.3: 130104016-130104016 on Build GRCh38
rs909402152 Chr.3: 130104119-130104119 on Build GRCh38
rs78806708 Chr.3: 130103773-130103773 on Build GRCh38
rs940768371 Chr.3: 130104021-130104021 on Build GRCh38
rs1002042103 Chr.3: 130103756-130103756 on Build GRCh38
rs866160256 Chr.3: 130104080-130104080 on Build GRCh38
rs140995046 Chr.3: 130103982-130103982 on Build GRCh38
rs917599110 Chr.3: 130103979-130103979 on Build GRCh38
rs1045936648 Chr.3: 130103855-130103855 on Build GRCh38
rs12495527 Chr.3: 130104107-130104107 on Build GRCh38
rs542242532 Chr.3: 130103758-130103758 on Build GRCh38
rs547634082 Chr.3: 130103824-130103824 on Build GRCh38

View Full Product Details