Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23586151-23586694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK2P1
Entrez Gene ID
1197
Gene Name
CDC like kinase 2, pseudogene 1
Gene Aliases
CLK2P
UniGene
Hs.632635
Gene Chromosome Location
Chr.7: 23584716-23586527 on Build GRCh38
Transcripts
NR_002711.3

SNP Detail

SNP ID Chromosome Location
rs868689514 Chr.7: 23586386-23586386 on Build GRCh38
rs961658184 Chr.7: 23586319-23586319 on Build GRCh38
rs1005748111 Chr.7: 23586447-23586447 on Build GRCh38
rs190182259 Chr.7: 23586636-23586636 on Build GRCh38
rs548357185 Chr.7: 23586324-23586324 on Build GRCh38
rs915348317 Chr.7: 23586502-23586502 on Build GRCh38
rs987450693 Chr.7: 23586640-23586640 on Build GRCh38
rs60425605 Chr.7: 23586591-23586592 on Build GRCh38
rs1023141682 Chr.7: 23586243-23586243 on Build GRCh38
rs955998458 Chr.7: 23586622-23586622 on Build GRCh38
rs562131828 Chr.7: 23586365-23586365 on Build GRCh38
rs769810179 Chr.7: 23586582-23586582 on Build GRCh38
rs559887608 Chr.7: 23586402-23586402 on Build GRCh38
rs1029668248 Chr.7: 23586489-23586489 on Build GRCh38
rs886766769 Chr.7: 23586512-23586512 on Build GRCh38
rs924695652 Chr.7: 23586615-23586615 on Build GRCh38
rs902833844 Chr.7: 23586178-23586178 on Build GRCh38
rs991737455 Chr.7: 23586470-23586470 on Build GRCh38
rs115991129 Chr.7: 23586198-23586198 on Build GRCh38
rs551477364 Chr.7: 23586524-23586524 on Build GRCh38
rs185040632 Chr.7: 23586525-23586525 on Build GRCh38
rs56204884 Chr.7: 23586583-23586583 on Build GRCh38
rs934693175 Chr.7: 23586436-23586436 on Build GRCh38
rs34576444 Chr.7: 23586374-23586374 on Build GRCh38
rs951146210 Chr.7: 23586340-23586340 on Build GRCh38
rs899599572 Chr.7: 23586302-23586302 on Build GRCh38
rs35821834 Chr.7: 23586418-23586418 on Build GRCh38
rs867836837 Chr.7: 23586627-23586627 on Build GRCh38
rs969330254 Chr.7: 23586599-23586599 on Build GRCh38
rs560001802 Chr.7: 23586616-23586616 on Build GRCh38
rs1050179194 Chr.7: 23586366-23586366 on Build GRCh38
rs1013055812 Chr.7: 23586237-23586237 on Build GRCh38
rs113649464 Chr.7: 23586258-23586258 on Build GRCh38
rs386711260 Chr.7: 23586592-23586597 on Build GRCh38
rs924631292 Chr.7: 23586432-23586432 on Build GRCh38
rs554320697 Chr.7: 23586503-23586503 on Build GRCh38
rs910405837 Chr.7: 23586370-23586370 on Build GRCh38
rs985524026 Chr.7: 23586496-23586496 on Build GRCh38
rs762113620 Chr.7: 23586495-23586495 on Build GRCh38
rs77759499 Chr.7: 23586236-23586236 on Build GRCh38
rs534739472 Chr.7: 23586419-23586419 on Build GRCh38
rs771440527 Chr.7: 23586313-23586313 on Build GRCh38
rs983029528 Chr.7: 23586348-23586348 on Build GRCh38
rs564943967 Chr.7: 23586535-23586535 on Build GRCh38
rs916181918 Chr.7: 23586485-23586485 on Build GRCh38
rs888604286 Chr.7: 23586441-23586441 on Build GRCh38
rs1043346296 Chr.7: 23586379-23586379 on Build GRCh38
rs1035842909 Chr.7: 23586228-23586228 on Build GRCh38
rs747190390 Chr.7: 23586320-23586320 on Build GRCh38
rs529544926 Chr.7: 23586261-23586261 on Build GRCh38
rs58498239 Chr.7: 23586597-23586598 on Build GRCh38
rs778991793 Chr.7: 23586175-23586175 on Build GRCh38
rs150787450 Chr.7: 23586508-23586508 on Build GRCh38
rs960299352 Chr.7: 23586556-23586556 on Build GRCh38
rs530763689 Chr.7: 23586371-23586371 on Build GRCh38
rs903741716 Chr.7: 23586184-23586184 on Build GRCh38
rs577638105 Chr.7: 23586558-23586558 on Build GRCh38
rs544100295 Chr.7: 23586171-23586171 on Build GRCh38
rs895515456 Chr.7: 23586410-23586410 on Build GRCh38
rs556993336 Chr.7: 23586550-23586550 on Build GRCh38
rs752725296 Chr.7: 23586200-23586200 on Build GRCh38
rs1052268582 Chr.7: 23586461-23586461 on Build GRCh38
rs935229535 Chr.7: 23586194-23586194 on Build GRCh38
rs868178021 Chr.7: 23586196-23586196 on Build GRCh38
rs546503168 Chr.7: 23586603-23586603 on Build GRCh38
rs932748739 Chr.7: 23586361-23586361 on Build GRCh38
rs551990165 Chr.7: 23586407-23586407 on Build GRCh38
rs59957695 Chr.7: 23586593-23586594 on Build GRCh38
rs961685622 Chr.7: 23586465-23586465 on Build GRCh38
rs1016258504 Chr.7: 23586448-23586448 on Build GRCh38
rs780819390 Chr.7: 23586435-23586435 on Build GRCh38
rs1035907686 Chr.7: 23586552-23586552 on Build GRCh38
rs912481261 Chr.7: 23586462-23586462 on Build GRCh38
rs569664819 Chr.7: 23586375-23586375 on Build GRCh38
rs977127024 Chr.7: 23586471-23586471 on Build GRCh38
rs1013062795 Chr.7: 23586595-23586595 on Build GRCh38
rs548421021 Chr.7: 23586528-23586528 on Build GRCh38
rs10258020 Chr.7: 23586221-23586221 on Build GRCh38
rs991660747 Chr.7: 23586382-23586382 on Build GRCh38
rs1023122821 Chr.7: 23586598-23586600 on Build GRCh38
rs530635234 Chr.7: 23586608-23586608 on Build GRCh38
rs879030094 Chr.7: 23586460-23586460 on Build GRCh38
rs903137538 Chr.7: 23586400-23586400 on Build GRCh38
rs912449553 Chr.7: 23586663-23586663 on Build GRCh38
rs528618666 Chr.7: 23586273-23586273 on Build GRCh38
rs976819560 Chr.7: 23586331-23586331 on Build GRCh38
rs922671287 Chr.7: 23586356-23586356 on Build GRCh38
rs61253820 Chr.7: 23586592-23586593 on Build GRCh38

View Full Product Details