Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44889890-44890062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB2
Entrez Gene ID
3689
Gene Name
integrin subunit beta 2
Gene Aliases
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7
UniGene
Hs.375957
Gene Chromosome Location
Chr.21: 44885949-44928873 on Build GRCh38
Transcripts
NM_000211.4 NM_001127491.2 NM_001303238.1 XM_006724001.1 XM_017028341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6688188 c.1650C>T p.G550G Chr.21: 44889985-44889985 on Build GRCh38
5734967 c.1605G>C p.E535D Chr.21: 44890030-44890030 on Build GRCh38
7338280 c.1657G>A p.G553R Chr.21: 44889978-44889978 on Build GRCh38
725842 c.1586T>C p.I529T Chr.21: 44890049-44890049 on Build GRCh38
4650563 c.1597T>C p.Y533H Chr.21: 44890038-44890038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900964697 Chr.21: 44889984-44889984 on Build GRCh38
rs148306562 Chr.21: 44890018-44890018 on Build GRCh38
rs901307822 Chr.21: 44889921-44889921 on Build GRCh38
rs376438520 Chr.21: 44889945-44889945 on Build GRCh38
rs759039359 Chr.21: 44889979-44889979 on Build GRCh38
rs767772567 Chr.21: 44889994-44889994 on Build GRCh38
rs751424754 Chr.21: 44889987-44889987 on Build GRCh38
rs1040161674 Chr.21: 44889916-44889916 on Build GRCh38
rs752599780 Chr.21: 44889933-44889933 on Build GRCh38
rs202215012 Chr.21: 44889938-44889938 on Build GRCh38
rs546243410 Chr.21: 44889931-44889931 on Build GRCh38
rs554317807 Chr.21: 44889913-44889913 on Build GRCh38
rs940347605 Chr.21: 44890024-44890024 on Build GRCh38
rs543193419 Chr.21: 44889927-44889927 on Build GRCh38
rs1032950586 Chr.21: 44889949-44889949 on Build GRCh38
rs371454881 Chr.21: 44889946-44889946 on Build GRCh38
rs770099996 Chr.21: 44889973-44889973 on Build GRCh38
rs201453026 Chr.21: 44889950-44889950 on Build GRCh38
rs748756345 Chr.21: 44889936-44889938 on Build GRCh38
rs201836589 Chr.21: 44889926-44889926 on Build GRCh38
rs200926893 Chr.21: 44889970-44889970 on Build GRCh38
rs757436659 Chr.21: 44889988-44889988 on Build GRCh38
rs1052752055 Chr.21: 44889999-44889999 on Build GRCh38
rs191487739 Chr.21: 44890016-44890016 on Build GRCh38
rs748574145 Chr.21: 44890033-44890033 on Build GRCh38
rs774867906 Chr.21: 44889964-44889964 on Build GRCh38
rs779414090 Chr.21: 44890027-44890027 on Build GRCh38
rs775983819 Chr.21: 44889972-44889972 on Build GRCh38
rs201703437 Chr.21: 44890005-44890005 on Build GRCh38
rs772587844 Chr.21: 44890043-44890043 on Build GRCh38
rs886057116 Chr.21: 44889993-44889993 on Build GRCh38
rs757536938 Chr.21: 44889941-44889941 on Build GRCh38
rs61747606 Chr.21: 44890000-44890000 on Build GRCh38
rs112883974 Chr.21: 44889912-44889912 on Build GRCh38
rs777553794 Chr.21: 44889939-44889939 on Build GRCh38
rs770417369 Chr.21: 44890008-44890008 on Build GRCh38
rs114479227 Chr.21: 44889969-44889969 on Build GRCh38
rs201000019 Chr.21: 44889980-44889980 on Build GRCh38
rs762895748 Chr.21: 44889982-44889982 on Build GRCh38
rs142476743 Chr.21: 44889985-44889985 on Build GRCh38
rs749744549 Chr.21: 44890010-44890010 on Build GRCh38

View Full Product Details