Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138812104-138812272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA1
Entrez Gene ID
1495
Gene Name
catenin alpha 1
Gene Aliases
CAP102, MDPT2
UniGene
Hs.445981
Gene Chromosome Location
Chr.5: 138753386-138935034 on Build GRCh38
Transcripts
NM_001290307.2 NM_001290309.2 NM_001290310.2 NM_001323982.1 NM_001323983.1 NM_001323984.1 NM_001323985.1 NM_001323986.1 NM_001903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019319 c.540A>C p.L180L Chr.5: 138812254-138812254 on Build GRCh38
1061436 c.520G>T p.G174* Chr.5: 138812234-138812234 on Build GRCh38
5723782 c.483C>T p.I161I Chr.5: 138812197-138812197 on Build GRCh38
482012 c.471G>T p.V157V Chr.5: 138812185-138812185 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754001909 Chr.5: 138812166-138812169 on Build GRCh38
rs200960267 Chr.5: 138812169-138812169 on Build GRCh38
rs778667305 Chr.5: 138812219-138812219 on Build GRCh38
rs200342369 Chr.5: 138812239-138812239 on Build GRCh38
rs771582713 Chr.5: 138812175-138812175 on Build GRCh38
rs533573408 Chr.5: 138812190-138812190 on Build GRCh38
rs777054384 Chr.5: 138812178-138812178 on Build GRCh38
rs779824402 Chr.5: 138812150-138812150 on Build GRCh38
rs778580048 Chr.5: 138812165-138812165 on Build GRCh38
rs749020625 Chr.5: 138812164-138812164 on Build GRCh38
rs375617895 Chr.5: 138812224-138812224 on Build GRCh38
rs746371282 Chr.5: 138812179-138812179 on Build GRCh38
rs767383317 Chr.5: 138812137-138812137 on Build GRCh38
rs755965555 Chr.5: 138812142-138812142 on Build GRCh38
rs761836161 Chr.5: 138812234-138812234 on Build GRCh38
rs955023956 Chr.5: 138812197-138812197 on Build GRCh38
rs774590663 Chr.5: 138812225-138812225 on Build GRCh38
rs370088383 Chr.5: 138812167-138812167 on Build GRCh38
rs754710691 Chr.5: 138812153-138812153 on Build GRCh38
rs150912388 Chr.5: 138812199-138812199 on Build GRCh38
rs766465706 Chr.5: 138812133-138812134 on Build GRCh38
rs139770883 Chr.5: 138812217-138812217 on Build GRCh38

View Full Product Details