Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43665601-43665844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4A
Entrez Gene ID
9682
Gene Name
lysine demethylase 4A
Gene Aliases
JHDM3A, JMJD2, JMJD2A, TDRD14A
UniGene
Hs.155983
Gene Chromosome Location
Chr.1: 43650118-43705518 on Build GRCh38
Transcripts
NM_014663.2 XM_005271354.3 XM_005271355.3 XM_005271356.3 XM_017002912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053520 c.639T>C p.P213P Chr.1: 43665711-43665711 on Build GRCh38
7089425 c.635C>A p.P212Q Chr.1: 43665707-43665707 on Build GRCh38
3689709 c.653G>A p.R218Q Chr.1: 43665725-43665725 on Build GRCh38
6692400 c.630T>A p.S210S Chr.1: 43665702-43665702 on Build GRCh38
4008296 c.666C>T p.L222L Chr.1: 43665738-43665738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758455774 Chr.1: 43665723-43665723 on Build GRCh38
rs760024094 Chr.1: 43665756-43665756 on Build GRCh38
rs753901104 Chr.1: 43665681-43665681 on Build GRCh38
rs775768000 Chr.1: 43665777-43665777 on Build GRCh38
rs561082409 Chr.1: 43665633-43665633 on Build GRCh38
rs752011798 Chr.1: 43665750-43665751 on Build GRCh38
rs772827417 Chr.1: 43665754-43665754 on Build GRCh38
rs767846483 Chr.1: 43665659-43665659 on Build GRCh38
rs188850156 Chr.1: 43665794-43665794 on Build GRCh38
rs778874325 Chr.1: 43665706-43665706 on Build GRCh38
rs779978360 Chr.1: 43665727-43665727 on Build GRCh38
rs562958939 Chr.1: 43665781-43665781 on Build GRCh38
rs201943297 Chr.1: 43665750-43665750 on Build GRCh38
rs964749317 Chr.1: 43665725-43665725 on Build GRCh38
rs139088184 Chr.1: 43665738-43665738 on Build GRCh38
rs757309292 Chr.1: 43665683-43665683 on Build GRCh38
rs968525621 Chr.1: 43665815-43665815 on Build GRCh38
rs772635487 Chr.1: 43665775-43665775 on Build GRCh38
rs995781690 Chr.1: 43665700-43665700 on Build GRCh38
rs201847175 Chr.1: 43665666-43665666 on Build GRCh38
rs746865217 Chr.1: 43665730-43665730 on Build GRCh38
rs760884692 Chr.1: 43665667-43665667 on Build GRCh38
rs1004320277 Chr.1: 43665802-43665802 on Build GRCh38
rs750292249 Chr.1: 43665709-43665709 on Build GRCh38
rs1019691337 Chr.1: 43665819-43665819 on Build GRCh38
rs769217786 Chr.1: 43665752-43665752 on Build GRCh38
rs915368889 Chr.1: 43665801-43665801 on Build GRCh38
rs764228845 Chr.1: 43665670-43665670 on Build GRCh38
rs765509007 Chr.1: 43665675-43665675 on Build GRCh38
rs768354168 Chr.1: 43665733-43665733 on Build GRCh38
rs764332320 Chr.1: 43665783-43665783 on Build GRCh38

View Full Product Details