Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29053807-29054078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA2
Entrez Gene ID
321
Gene Name
amyloid beta precursor protein binding family A member 2
Gene Aliases
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
UniGene
Hs.618112
Gene Chromosome Location
Chr.15: 28885479-29118315 on Build GRCh38
Transcripts
NM_001130414.1 NM_005503.3 XM_011521488.2 XM_011521489.2 XM_011521490.2 XM_011521491.1 XM_011521492.2 XM_011521493.2 XM_011521494.2 XM_017022101.1 XM_017022102.1 XM_017022103.1 XM_017022104.1 XM_017022105.1 XM_017022106.1 XM_017022107.1 XM_017022108.1 XM_017022109.1 XM_017022110.1 XM_017022111.1 XM_017022112.1 XM_017022113.1 XM_017022114.1 XM_017022115.1 XM_017022116.1 XM_017022117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4833466 c.88G>C p.E30Q Chr.15: 29053972-29053972 on Build GRCh38
5689401 c.130G>A p.E44K Chr.15: 29054014-29054014 on Build GRCh38
4053879 c.72T>A p.P24P Chr.15: 29053956-29053956 on Build GRCh38
5666446 c.157C>A p.P53T Chr.15: 29054041-29054041 on Build GRCh38
4820889 c.192C>G p.H64Q Chr.15: 29054076-29054076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117098828 Chr.15: 29054039-29054039 on Build GRCh38
rs372072184 Chr.15: 29053855-29053855 on Build GRCh38
rs139506148 Chr.15: 29054014-29054014 on Build GRCh38
rs750653807 Chr.15: 29053932-29053932 on Build GRCh38
rs376706582 Chr.15: 29053878-29053878 on Build GRCh38
rs765271124 Chr.15: 29053931-29053931 on Build GRCh38
rs116449254 Chr.15: 29053856-29053856 on Build GRCh38
rs183585376 Chr.15: 29054004-29054004 on Build GRCh38
rs149792305 Chr.15: 29053918-29053918 on Build GRCh38
rs775406200 Chr.15: 29053851-29053851 on Build GRCh38
rs776598599 Chr.15: 29054006-29054006 on Build GRCh38
rs746831479 Chr.15: 29053994-29053994 on Build GRCh38
rs187302261 Chr.15: 29054018-29054018 on Build GRCh38
rs778348823 Chr.15: 29053976-29053976 on Build GRCh38
rs777078546 Chr.15: 29053895-29053895 on Build GRCh38
rs374330797 Chr.15: 29053961-29053961 on Build GRCh38
rs774288821 Chr.15: 29053850-29053850 on Build GRCh38
rs774025744 Chr.15: 29053909-29053909 on Build GRCh38
rs116221298 Chr.15: 29053917-29053917 on Build GRCh38
rs56312364 Chr.15: 29053941-29053941 on Build GRCh38
rs982836014 Chr.15: 29053935-29053935 on Build GRCh38
rs755550991 Chr.15: 29053894-29053894 on Build GRCh38
rs371759617 Chr.15: 29054053-29054053 on Build GRCh38
rs755500155 Chr.15: 29053958-29053958 on Build GRCh38
rs761723488 Chr.15: 29054013-29054013 on Build GRCh38
rs575379564 Chr.15: 29054038-29054038 on Build GRCh38
rs571580177 Chr.15: 29053923-29053923 on Build GRCh38
rs776526267 Chr.15: 29053927-29053927 on Build GRCh38
rs764052686 Chr.15: 29053926-29053926 on Build GRCh38
rs371009893 Chr.15: 29053928-29053928 on Build GRCh38
rs747501250 Chr.15: 29053886-29053886 on Build GRCh38
rs367887200 Chr.15: 29054052-29054052 on Build GRCh38
rs757998938 Chr.15: 29054057-29054057 on Build GRCh38
rs145726347 Chr.15: 29053948-29053948 on Build GRCh38
rs142678624 Chr.15: 29054048-29054048 on Build GRCh38
rs763197580 Chr.15: 29054033-29054033 on Build GRCh38
rs141358568 Chr.15: 29054054-29054054 on Build GRCh38
rs778993688 Chr.15: 29053984-29053984 on Build GRCh38
rs770399185 Chr.15: 29053908-29053908 on Build GRCh38
rs545698147 Chr.15: 29053971-29053971 on Build GRCh38
rs753958933 Chr.15: 29053862-29053862 on Build GRCh38
rs529677959 Chr.15: 29053831-29053831 on Build GRCh38
rs767343117 Chr.15: 29053840-29053840 on Build GRCh38
rs750153512 Chr.15: 29053983-29053983 on Build GRCh38
rs753188362 Chr.15: 29053970-29053970 on Build GRCh38
rs771845389 Chr.15: 29053912-29053912 on Build GRCh38
rs766735777 Chr.15: 29054036-29054036 on Build GRCh38
rs557200678 Chr.15: 29053977-29053977 on Build GRCh38
rs145094576 Chr.15: 29054055-29054055 on Build GRCh38
rs112707956 Chr.15: 29053871-29053871 on Build GRCh38
rs771718714 Chr.15: 29053978-29053978 on Build GRCh38
rs138680751 Chr.15: 29053884-29053884 on Build GRCh38
rs1009977625 Chr.15: 29053866-29053866 on Build GRCh38
rs758751173 Chr.15: 29053942-29053942 on Build GRCh38
rs748858665 Chr.15: 29053899-29053899 on Build GRCh38
rs765516950 Chr.15: 29053877-29053877 on Build GRCh38

View Full Product Details