Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98822953-98823188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
UniGene
Hs.469398
Gene Chromosome Location
Chr.2: 98793846-98936221 on Build GRCh38
Transcripts
NM_207362.2 XM_005263923.3 XM_005263924.3 XM_005263926.3 XM_011511093.1 XM_011511094.1 XM_011511095.2 XM_011511096.1 XM_017003971.1 XM_017003972.1 XM_017003973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5745931 c.1288G>T p.G430W Chr.2: 98822985-98822985 on Build GRCh38
6401164 c.1234A>G p.T412A Chr.2: 98823039-98823039 on Build GRCh38
5354449 c.1284C>G p.R428R Chr.2: 98822989-98822989 on Build GRCh38
5183339 c.1242C>T p.P414P Chr.2: 98823031-98823031 on Build GRCh38
4644476 c.1203G>A p.P401P Chr.2: 98823070-98823070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779624709 Chr.2: 98823006-98823006 on Build GRCh38
rs769046322 Chr.2: 98823059-98823059 on Build GRCh38
rs764688697 Chr.2: 98823110-98823110 on Build GRCh38
rs753438324 Chr.2: 98823103-98823103 on Build GRCh38
rs755950652 Chr.2: 98822998-98822998 on Build GRCh38
rs752367709 Chr.2: 98823122-98823122 on Build GRCh38
rs769055823 Chr.2: 98823016-98823016 on Build GRCh38
rs774518111 Chr.2: 98823146-98823146 on Build GRCh38
rs748390102 Chr.2: 98823149-98823149 on Build GRCh38
rs1052486408 Chr.2: 98823055-98823055 on Build GRCh38
rs975613469 Chr.2: 98823092-98823092 on Build GRCh38
rs550777088 Chr.2: 98823161-98823161 on Build GRCh38
rs991872810 Chr.2: 98823037-98823037 on Build GRCh38
rs559619381 Chr.2: 98823023-98823023 on Build GRCh38
rs768881020 Chr.2: 98823144-98823144 on Build GRCh38
rs753284324 Chr.2: 98823008-98823008 on Build GRCh38
rs746399760 Chr.2: 98823036-98823036 on Build GRCh38
rs1044083074 Chr.2: 98823005-98823005 on Build GRCh38
rs867812183 Chr.2: 98823060-98823060 on Build GRCh38
rs1028353193 Chr.2: 98823102-98823102 on Build GRCh38
rs149622421 Chr.2: 98823140-98823140 on Build GRCh38
rs767360786 Chr.2: 98823070-98823070 on Build GRCh38
rs766214701 Chr.2: 98823088-98823088 on Build GRCh38
rs749417186 Chr.2: 98823142-98823142 on Build GRCh38
rs760736948 Chr.2: 98823081-98823081 on Build GRCh38
rs368429942 Chr.2: 98823134-98823134 on Build GRCh38
rs767494728 Chr.2: 98823142-98823142 on Build GRCh38
rs537525119 Chr.2: 98823151-98823151 on Build GRCh38
rs955537794 Chr.2: 98823127-98823127 on Build GRCh38
rs551912252 Chr.2: 98823139-98823139 on Build GRCh38
rs774681646 Chr.2: 98823061-98823061 on Build GRCh38
rs372545695 Chr.2: 98822997-98822997 on Build GRCh38
rs754455638 Chr.2: 98823109-98823109 on Build GRCh38
rs778515833 Chr.2: 98823018-98823018 on Build GRCh38
rs750122514 Chr.2: 98823076-98823076 on Build GRCh38
rs766381405 Chr.2: 98823162-98823162 on Build GRCh38
rs751454818 Chr.2: 98822985-98822985 on Build GRCh38
rs772183469 Chr.2: 98823150-98823150 on Build GRCh38
rs771314260 Chr.2: 98823029-98823029 on Build GRCh38
rs1038329063 Chr.2: 98822971-98822971 on Build GRCh38
rs764403462 Chr.2: 98822976-98822976 on Build GRCh38
rs905422583 Chr.2: 98823133-98823133 on Build GRCh38
rs747415781 Chr.2: 98823021-98823021 on Build GRCh38
rs763107732 Chr.2: 98823056-98823056 on Build GRCh38
rs186269747 Chr.2: 98823049-98823049 on Build GRCh38
rs368628945 Chr.2: 98822989-98822989 on Build GRCh38
rs776583129 Chr.2: 98823168-98823168 on Build GRCh38
rs759418878 Chr.2: 98823171-98823171 on Build GRCh38
rs781361854 Chr.2: 98823031-98823031 on Build GRCh38
rs1022386435 Chr.2: 98823075-98823075 on Build GRCh38
rs1031823100 Chr.2: 98823032-98823032 on Build GRCh38
rs761675428 Chr.2: 98822986-98822986 on Build GRCh38
rs746210600 Chr.2: 98823135-98823135 on Build GRCh38
rs567697961 Chr.2: 98823172-98823172 on Build GRCh38
rs375696418 Chr.2: 98823129-98823129 on Build GRCh38

View Full Product Details