Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10433354-10433581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_006720954.3 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023708.1 XM_017023710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641984 c.887G>A p.R296H Chr.16: 10433576-10433576 on Build GRCh38
4939014 c.873G>A p.E291E Chr.16: 10433562-10433562 on Build GRCh38
6446064 c.891G>A p.M297I Chr.16: 10433580-10433580 on Build GRCh38
6420314 c.879T>A p.N293K Chr.16: 10433568-10433568 on Build GRCh38
3505699 c.874G>A p.E292K Chr.16: 10433563-10433563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048328939 Chr.16: 10433381-10433381 on Build GRCh38
rs373909615 Chr.16: 10433405-10433405 on Build GRCh38
rs59469386 Chr.16: 10433408-10433408 on Build GRCh38
rs753280215 Chr.16: 10433498-10433498 on Build GRCh38
rs942460587 Chr.16: 10433427-10433427 on Build GRCh38
rs768459665 Chr.16: 10433543-10433543 on Build GRCh38
rs151108987 Chr.16: 10433545-10433545 on Build GRCh38
rs149006305 Chr.16: 10433412-10433412 on Build GRCh38
rs756635952 Chr.16: 10433509-10433509 on Build GRCh38
rs755402494 Chr.16: 10433491-10433491 on Build GRCh38
rs748682973 Chr.16: 10433494-10433497 on Build GRCh38
rs373265497 Chr.16: 10433538-10433538 on Build GRCh38
rs749646325 Chr.16: 10433516-10433518 on Build GRCh38
rs778140845 Chr.16: 10433513-10433513 on Build GRCh38
rs890911465 Chr.16: 10433417-10433417 on Build GRCh38
rs374785518 Chr.16: 10433476-10433476 on Build GRCh38
rs769926353 Chr.16: 10433432-10433432 on Build GRCh38
rs746766010 Chr.16: 10433541-10433541 on Build GRCh38
rs756197596 Chr.16: 10433499-10433499 on Build GRCh38
rs973984687 Chr.16: 10433389-10433389 on Build GRCh38
rs749512324 Chr.16: 10433516-10433516 on Build GRCh38
rs751918426 Chr.16: 10433479-10433479 on Build GRCh38
rs141089693 Chr.16: 10433547-10433547 on Build GRCh38
rs550934018 Chr.16: 10433392-10433392 on Build GRCh38
rs769345893 Chr.16: 10433554-10433554 on Build GRCh38
rs768098696 Chr.16: 10433493-10433493 on Build GRCh38
rs777756775 Chr.16: 10433503-10433503 on Build GRCh38
rs902754189 Chr.16: 10433511-10433511 on Build GRCh38
rs1022478994 Chr.16: 10433494-10433494 on Build GRCh38
rs73497840 Chr.16: 10433526-10433526 on Build GRCh38
rs1033048803 Chr.16: 10433536-10433536 on Build GRCh38
rs757363855 Chr.16: 10433522-10433522 on Build GRCh38
rs181508868 Chr.16: 10433395-10433395 on Build GRCh38

View Full Product Details