Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12650988-12651261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL4
Entrez Gene ID
343066
Gene Name
arylacetamide deacetylase like 4
Gene Aliases
-
UniGene
Hs.209954
Gene Chromosome Location
Chr.1: 12644547-12667086 on Build GRCh38
Transcripts
NM_001013630.1 XM_017001152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3417970 c.176T>C p.I59T Chr.1: 12651130-12651130 on Build GRCh38
6589225 c.174T>C p.N58N Chr.1: 12651128-12651128 on Build GRCh38
3984149 c.264G>T p.V88V Chr.1: 12651218-12651218 on Build GRCh38
1472494 c.172A>T p.N58Y Chr.1: 12651126-12651126 on Build GRCh38
6589226 c.283G>A p.G95R Chr.1: 12651237-12651237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924220943 Chr.1: 12651159-12651159 on Build GRCh38
rs539452254 Chr.1: 12651242-12651242 on Build GRCh38
rs117928838 Chr.1: 12651093-12651093 on Build GRCh38
rs144096970 Chr.1: 12651186-12651186 on Build GRCh38
rs1012107787 Chr.1: 12651137-12651137 on Build GRCh38
rs756287230 Chr.1: 12651193-12651193 on Build GRCh38
rs775815654 Chr.1: 12651147-12651147 on Build GRCh38
rs1036746421 Chr.1: 12651223-12651223 on Build GRCh38
rs749533019 Chr.1: 12651202-12651202 on Build GRCh38
rs986248510 Chr.1: 12651204-12651204 on Build GRCh38
rs772785801 Chr.1: 12651233-12651233 on Build GRCh38
rs749930578 Chr.1: 12651086-12651086 on Build GRCh38
rs369351522 Chr.1: 12651099-12651099 on Build GRCh38
rs970454738 Chr.1: 12651152-12651152 on Build GRCh38
rs768757535 Chr.1: 12651203-12651203 on Build GRCh38
rs764770045 Chr.1: 12651231-12651231 on Build GRCh38
rs768513905 Chr.1: 12651154-12651154 on Build GRCh38
rs187441277 Chr.1: 12651206-12651206 on Build GRCh38
rs528198918 Chr.1: 12651163-12651163 on Build GRCh38
rs146047731 Chr.1: 12651064-12651064 on Build GRCh38
rs367808836 Chr.1: 12651241-12651241 on Build GRCh38
rs777941473 Chr.1: 12651108-12651108 on Build GRCh38
rs200551069 Chr.1: 12651207-12651207 on Build GRCh38
rs541552858 Chr.1: 12651172-12651172 on Build GRCh38
rs753540436 Chr.1: 12651168-12651168 on Build GRCh38
rs772039677 Chr.1: 12651213-12651213 on Build GRCh38
rs747554816 Chr.1: 12651113-12651113 on Build GRCh38
rs543973930 Chr.1: 12651225-12651225 on Build GRCh38
rs139448447 Chr.1: 12651156-12651156 on Build GRCh38
rs148090511 Chr.1: 12651230-12651230 on Build GRCh38
rs74546579 Chr.1: 12651208-12651208 on Build GRCh38
rs575745701 Chr.1: 12651008-12651008 on Build GRCh38
rs770177277 Chr.1: 12651146-12651146 on Build GRCh38
rs753390937 Chr.1: 12651098-12651098 on Build GRCh38
rs767429067 Chr.1: 12651157-12651157 on Build GRCh38
rs932080704 Chr.1: 12651176-12651176 on Build GRCh38
rs544917975 Chr.1: 12651039-12651039 on Build GRCh38
rs779944375 Chr.1: 12651195-12651195 on Build GRCh38
rs770479268 Chr.1: 12651227-12651227 on Build GRCh38
rs745918050 Chr.1: 12651132-12651132 on Build GRCh38
rs199989547 Chr.1: 12651178-12651178 on Build GRCh38
rs772760183 Chr.1: 12651160-12651160 on Build GRCh38
rs751853145 Chr.1: 12651181-12651181 on Build GRCh38
rs575065583 Chr.1: 12651129-12651129 on Build GRCh38
rs775999994 Chr.1: 12651229-12651229 on Build GRCh38
rs374661453 Chr.1: 12651236-12651236 on Build GRCh38
rs1023497807 Chr.1: 12651143-12651143 on Build GRCh38
rs140616597 Chr.1: 12651224-12651224 on Build GRCh38
rs368891966 Chr.1: 12651054-12651054 on Build GRCh38
rs781754032 Chr.1: 12651188-12651188 on Build GRCh38
rs558877298 Chr.1: 12651121-12651121 on Build GRCh38
rs201507886 Chr.1: 12651232-12651232 on Build GRCh38
rs375635678 Chr.1: 12651171-12651171 on Build GRCh38
rs767020147 Chr.1: 12651079-12651079 on Build GRCh38
rs781475069 Chr.1: 12651125-12651125 on Build GRCh38
rs138804535 Chr.1: 12651094-12651094 on Build GRCh38
rs752009904 Chr.1: 12651237-12651237 on Build GRCh38
rs370456693 Chr.1: 12651189-12651189 on Build GRCh38
rs997908128 Chr.1: 12651232-12651233 on Build GRCh38

View Full Product Details