Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197512061-197512306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
UniGene
Hs.274539
Gene Chromosome Location
Chr.3: 197509783-197573323 on Build GRCh38
Transcripts
NM_004051.4 NM_203314.2 NM_203315.2 XM_005269352.3 XM_005269355.4 XM_011513067.2 XM_017007007.1 XM_017007008.1 XM_017007009.1 XM_017007010.1 XM_017007011.1 XM_017007012.1 XM_017007014.1 XM_017007015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164166 c.641G>T p.G214V Chr.3: 197512286-197512286 on Build GRCh38
4450720 c.797_798delAG p.E266fs*12 Chr.3: 197512129-197512130 on Build GRCh38
2947856 c.802G>A p.V268M Chr.3: 197512125-197512125 on Build GRCh38
6498084 c.709G>C p.E237Q Chr.3: 197512218-197512218 on Build GRCh38
4584215 c.816C>T p.Y272Y Chr.3: 197512111-197512111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768975972 Chr.3: 197512245-197512245 on Build GRCh38
rs928975303 Chr.3: 197512209-197512209 on Build GRCh38
rs1000703569 Chr.3: 197512243-197512243 on Build GRCh38
rs773113109 Chr.3: 197512260-197512260 on Build GRCh38
rs887386685 Chr.3: 197512215-197512215 on Build GRCh38
rs747867608 Chr.3: 197512251-197512251 on Build GRCh38
rs202110030 Chr.3: 197512179-197512179 on Build GRCh38
rs752919802 Chr.3: 197512230-197512230 on Build GRCh38
rs746397441 Chr.3: 197512232-197512232 on Build GRCh38
rs1038980493 Chr.3: 197512161-197512161 on Build GRCh38
rs987355205 Chr.3: 197512141-197512141 on Build GRCh38
rs368212467 Chr.3: 197512212-197512212 on Build GRCh38
rs773135340 Chr.3: 197512198-197512198 on Build GRCh38
rs370655515 Chr.3: 197512137-197512137 on Build GRCh38
rs767131775 Chr.3: 197512271-197512271 on Build GRCh38
rs559038515 Chr.3: 197512273-197512273 on Build GRCh38
rs145598252 Chr.3: 197512083-197512083 on Build GRCh38
rs890810529 Chr.3: 197512214-197512214 on Build GRCh38
rs762920612 Chr.3: 197512262-197512262 on Build GRCh38
rs183131131 Chr.3: 197512126-197512126 on Build GRCh38
rs145104101 Chr.3: 197512267-197512267 on Build GRCh38
rs779504270 Chr.3: 197512109-197512109 on Build GRCh38
rs757649832 Chr.3: 197512277-197512277 on Build GRCh38
rs140457761 Chr.3: 197512200-197512200 on Build GRCh38
rs745877266 Chr.3: 197512155-197512155 on Build GRCh38
rs1037863518 Chr.3: 197512174-197512174 on Build GRCh38
rs757702931 Chr.3: 197512224-197512224 on Build GRCh38
rs371927738 Chr.3: 197512270-197512270 on Build GRCh38
rs147271608 Chr.3: 197512084-197512084 on Build GRCh38
rs140846840 Chr.3: 197512153-197512153 on Build GRCh38
rs61745694 Chr.3: 197512183-197512183 on Build GRCh38
rs748674418 Chr.3: 197512241-197512241 on Build GRCh38
rs754729827 Chr.3: 197512213-197512213 on Build GRCh38
rs772311219 Chr.3: 197512159-197512159 on Build GRCh38
rs779524014 Chr.3: 197512225-197512225 on Build GRCh38
rs114600793 Chr.3: 197512284-197512284 on Build GRCh38
rs751249430 Chr.3: 197512121-197512121 on Build GRCh38
rs144830970 Chr.3: 197512286-197512286 on Build GRCh38
rs913842521 Chr.3: 197512111-197512111 on Build GRCh38
rs765097046 Chr.3: 197512197-197512197 on Build GRCh38
rs771624244 Chr.3: 197512136-197512136 on Build GRCh38
rs545192310 Chr.3: 197512158-197512158 on Build GRCh38
rs765743905 Chr.3: 197512278-197512278 on Build GRCh38
rs747191828 Chr.3: 197512237-197512237 on Build GRCh38
rs758384765 Chr.3: 197512236-197512236 on Build GRCh38
rs367827020 Chr.3: 197512122-197512122 on Build GRCh38
rs769605817 Chr.3: 197512258-197512258 on Build GRCh38
rs1002260866 Chr.3: 197512242-197512242 on Build GRCh38
rs144603437 Chr.3: 197512171-197512171 on Build GRCh38
rs761005595 Chr.3: 197512163-197512163 on Build GRCh38
rs781744588 Chr.3: 197512135-197512136 on Build GRCh38
rs149945578 Chr.3: 197512216-197512216 on Build GRCh38
rs374039230 Chr.3: 197512182-197512182 on Build GRCh38
rs776728556 Chr.3: 197512249-197512249 on Build GRCh38
rs1044673041 Chr.3: 197512142-197512142 on Build GRCh38

View Full Product Details