Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89963146-89963395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEF8
Entrez Gene ID
54849
Gene Name
differentially expressed in FDCP 8 homolog (mouse)
Gene Aliases
-
UniGene
Hs.62771
Gene Chromosome Location
Chr.16: 89948697-89968060 on Build GRCh38
Transcripts
NM_001242816.1 NM_001242817.1 NM_001242818.1 NM_001242819.1 NM_001242820.1 NM_207514.2 XM_005256318.2 XM_011523156.2 XM_011523157.2 XM_011523158.1 XM_011523159.2 XM_011523160.2 XM_011523161.2 XM_011523162.2 XM_017023358.1 XM_017023359.1 XM_017023360.1 XM_017023361.1 XM_017023362.1 XM_017023363.1 XM_017023364.1 XM_017023365.1 XM_017023366.1 XM_017023367.1 XM_017023368.1 XM_017023369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063703 c.1113C>A p.R371R Chr.16: 89963371-89963371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs930388745 Chr.16: 89963204-89963204 on Build GRCh38
rs1052406314 Chr.16: 89963260-89963260 on Build GRCh38
rs370905978 Chr.16: 89963317-89963317 on Build GRCh38
rs971725803 Chr.16: 89963183-89963183 on Build GRCh38
rs372723862 Chr.16: 89963374-89963374 on Build GRCh38
rs985837438 Chr.16: 89963350-89963350 on Build GRCh38
rs201332289 Chr.16: 89963313-89963313 on Build GRCh38
rs948899703 Chr.16: 89963299-89963299 on Build GRCh38
rs777982333 Chr.16: 89963352-89963352 on Build GRCh38
rs916026439 Chr.16: 89963292-89963292 on Build GRCh38
rs748931782 Chr.16: 89963365-89963365 on Build GRCh38
rs144445236 Chr.16: 89963237-89963237 on Build GRCh38
rs115841388 Chr.16: 89963252-89963252 on Build GRCh38
rs750442807 Chr.16: 89963314-89963314 on Build GRCh38
rs569973947 Chr.16: 89963279-89963279 on Build GRCh38
rs903094101 Chr.16: 89963244-89963244 on Build GRCh38
rs200532672 Chr.16: 89963316-89963316 on Build GRCh38
rs567935042 Chr.16: 89963170-89963170 on Build GRCh38
rs1050614188 Chr.16: 89963205-89963205 on Build GRCh38
rs944504271 Chr.16: 89963222-89963222 on Build GRCh38
rs977402752 Chr.16: 89963261-89963261 on Build GRCh38
rs555796873 Chr.16: 89963305-89963305 on Build GRCh38
rs537419617 Chr.16: 89963281-89963281 on Build GRCh38
rs1003660854 Chr.16: 89963306-89963306 on Build GRCh38
rs199577324 Chr.16: 89963342-89963342 on Build GRCh38
rs143207147 Chr.16: 89963172-89963172 on Build GRCh38
rs761612204 Chr.16: 89963312-89963312 on Build GRCh38
rs924665616 Chr.16: 89963265-89963265 on Build GRCh38
rs968677215 Chr.16: 89963254-89963254 on Build GRCh38
rs369419867 Chr.16: 89963366-89963366 on Build GRCh38
rs897717267 Chr.16: 89963173-89963173 on Build GRCh38
rs910301882 Chr.16: 89963369-89963369 on Build GRCh38
rs778533326 Chr.16: 89963289-89963289 on Build GRCh38
rs920245719 Chr.16: 89963321-89963321 on Build GRCh38
rs755618328 Chr.16: 89963310-89963310 on Build GRCh38
rs377483048 Chr.16: 89963349-89963349 on Build GRCh38
rs748310678 Chr.16: 89963348-89963348 on Build GRCh38
rs910333160 Chr.16: 89963212-89963212 on Build GRCh38
rs749858482 Chr.16: 89963358-89963358 on Build GRCh38
rs1010961263 Chr.16: 89963286-89963286 on Build GRCh38
rs920259366 Chr.16: 89963193-89963193 on Build GRCh38
rs1051688123 Chr.16: 89963357-89963357 on Build GRCh38
rs865847334 Chr.16: 89963290-89963290 on Build GRCh38

View Full Product Details