Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113001027-113001278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640977 c.497G>A p.R166Q Chr.5: 113001148-113001148 on Build GRCh38
256742 c.442G>T p.E148* Chr.5: 113001093-113001093 on Build GRCh38
1619328 c.460A>G p.K154E Chr.5: 113001111-113001111 on Build GRCh38
4552927 c.575G>A p.R192K Chr.5: 113001226-113001226 on Build GRCh38
6529578 c.577G>C p.E193Q Chr.5: 113001228-113001228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746565006 Chr.5: 113001161-113001161 on Build GRCh38
rs145664285 Chr.5: 113001159-113001159 on Build GRCh38
rs369278042 Chr.5: 113001168-113001168 on Build GRCh38
rs761656672 Chr.5: 113001084-113001084 on Build GRCh38
rs143369361 Chr.5: 113001101-113001101 on Build GRCh38
rs754141957 Chr.5: 113001130-113001130 on Build GRCh38
rs767770193 Chr.5: 113001210-113001210 on Build GRCh38
rs371708969 Chr.5: 113001152-113001152 on Build GRCh38
rs780660617 Chr.5: 113001125-113001125 on Build GRCh38
rs367934885 Chr.5: 113001126-113001126 on Build GRCh38
rs761986554 Chr.5: 113001209-113001209 on Build GRCh38
rs984252692 Chr.5: 113001115-113001115 on Build GRCh38
rs755437872 Chr.5: 113001131-113001131 on Build GRCh38
rs555000060 Chr.5: 113001230-113001230 on Build GRCh38
rs961346675 Chr.5: 113001132-113001132 on Build GRCh38
rs767539577 Chr.5: 113001095-113001095 on Build GRCh38
rs557055649 Chr.5: 113001169-113001169 on Build GRCh38
rs374882349 Chr.5: 113001154-113001154 on Build GRCh38
rs765964956 Chr.5: 113001059-113001060 on Build GRCh38
rs142459154 Chr.5: 113001206-113001206 on Build GRCh38
rs745794074 Chr.5: 113001180-113001180 on Build GRCh38
rs918212455 Chr.5: 113001078-113001078 on Build GRCh38
rs753606346 Chr.5: 113001076-113001077 on Build GRCh38
rs143056869 Chr.5: 113001233-113001233 on Build GRCh38
rs909858090 Chr.5: 113001119-113001119 on Build GRCh38
rs779150526 Chr.5: 113001138-113001138 on Build GRCh38
rs769570611 Chr.5: 113001185-113001185 on Build GRCh38
rs756585237 Chr.5: 113001118-113001118 on Build GRCh38
rs770976241 Chr.5: 113001167-113001167 on Build GRCh38
rs746974670 Chr.5: 113001107-113001107 on Build GRCh38
rs928067250 Chr.5: 113001156-113001156 on Build GRCh38
rs754818941 Chr.5: 113001246-113001247 on Build GRCh38

View Full Product Details