Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137306869-137307104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD3
Entrez Gene ID
54932
Gene Name
exonuclease 3'-5' domain containing 3
Gene Aliases
mut-7
UniGene
Hs.495553
Gene Chromosome Location
Chr.9: 137306894-137423262 on Build GRCh38
Transcripts
NM_017820.4 XM_011518810.1 XM_011518814.1 XM_011518815.1 XM_017014861.1 XM_017014862.1 XM_017014863.1 XM_017014864.1 XM_017014865.1 XM_017014866.1 XM_017014867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426576 c.2598C>T p.S866S Chr.9: 137306983-137306983 on Build GRCh38
4926656 c.2569G>T p.E857* Chr.9: 137307012-137307012 on Build GRCh38
2153928 c.2613G>A p.P871P Chr.9: 137306968-137306968 on Build GRCh38
1284789 c.2493C>T p.C831C Chr.9: 137307088-137307088 on Build GRCh38
5590836 c.2616C>T p.A872A Chr.9: 137306965-137306965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756213606 Chr.9: 137307076-137307076 on Build GRCh38
rs898058049 Chr.9: 137307006-137307006 on Build GRCh38
rs531794122 Chr.9: 137306899-137306899 on Build GRCh38
rs374732855 Chr.9: 137307007-137307007 on Build GRCh38
rs904629748 Chr.9: 137306958-137306958 on Build GRCh38
rs1020109243 Chr.9: 137306979-137306979 on Build GRCh38
rs879717411 Chr.9: 137307084-137307084 on Build GRCh38
rs750524262 Chr.9: 137306993-137306993 on Build GRCh38
rs760247028 Chr.9: 137306977-137306977 on Build GRCh38
rs368309115 Chr.9: 137306908-137306908 on Build GRCh38
rs1033062433 Chr.9: 137306987-137306987 on Build GRCh38
rs143164347 Chr.9: 137307024-137307024 on Build GRCh38
rs576576838 Chr.9: 137307046-137307046 on Build GRCh38
rs745995647 Chr.9: 137306954-137306954 on Build GRCh38
rs1018433766 Chr.9: 137307049-137307049 on Build GRCh38
rs113731401 Chr.9: 137306959-137306959 on Build GRCh38
rs771749432 Chr.9: 137306974-137306974 on Build GRCh38
rs754811360 Chr.9: 137306920-137306920 on Build GRCh38
rs757371794 Chr.9: 137306932-137306932 on Build GRCh38
rs765138117 Chr.9: 137307048-137307048 on Build GRCh38
rs548375974 Chr.9: 137306914-137306914 on Build GRCh38
rs767032443 Chr.9: 137306910-137306910 on Build GRCh38
rs761481299 Chr.9: 137306969-137306969 on Build GRCh38
rs910850696 Chr.9: 137306904-137306904 on Build GRCh38
rs751830278 Chr.9: 137306926-137306926 on Build GRCh38
rs752477082 Chr.9: 137306991-137306991 on Build GRCh38
rs753573360 Chr.9: 137306918-137306918 on Build GRCh38
rs770410177 Chr.9: 137306957-137306957 on Build GRCh38
rs748558804 Chr.9: 137307014-137307014 on Build GRCh38
rs568976884 Chr.9: 137306898-137306898 on Build GRCh38
rs779302555 Chr.9: 137307008-137307008 on Build GRCh38
rs373543265 Chr.9: 137307082-137307082 on Build GRCh38
rs771710735 Chr.9: 137307023-137307023 on Build GRCh38
rs118011827 Chr.9: 137306981-137306981 on Build GRCh38
rs749820023 Chr.9: 137306998-137306998 on Build GRCh38
rs775413905 Chr.9: 137307057-137307057 on Build GRCh38
rs539746946 Chr.9: 137306992-137306992 on Build GRCh38
rs756266785 Chr.9: 137306996-137306996 on Build GRCh38
rs759203903 Chr.9: 137306986-137306986 on Build GRCh38
rs372867176 Chr.9: 137307035-137307035 on Build GRCh38
rs372339289 Chr.9: 137306917-137306917 on Build GRCh38
rs780367341 Chr.9: 137306997-137306997 on Build GRCh38
rs762905785 Chr.9: 137307058-137307058 on Build GRCh38
rs749549421 Chr.9: 137306962-137306962 on Build GRCh38
rs781392909 Chr.9: 137306937-137306937 on Build GRCh38
rs200899844 Chr.9: 137307042-137307042 on Build GRCh38
rs766166062 Chr.9: 137306978-137306978 on Build GRCh38
rs773823557 Chr.9: 137306968-137306968 on Build GRCh38
rs375831870 Chr.9: 137307041-137307041 on Build GRCh38
rs772698953 Chr.9: 137306976-137306976 on Build GRCh38
rs765112099 Chr.9: 137306989-137306989 on Build GRCh38
rs779358702 Chr.9: 137307083-137307083 on Build GRCh38
rs766507998 Chr.9: 137307077-137307077 on Build GRCh38
rs750685846 Chr.9: 137307065-137307065 on Build GRCh38
rs200221658 Chr.9: 137306963-137306963 on Build GRCh38

View Full Product Details