Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137009302-137009531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL20RA
Entrez Gene ID
53832
Gene Name
interleukin 20 receptor subunit alpha
Gene Aliases
CRF2-8, IL-20R-alpha, IL-20R1, IL-20RA
UniGene
Hs.445868
Gene Chromosome Location
Chr.6: 136999971-137045366 on Build GRCh38
Transcripts
NM_001278722.1 NM_001278723.1 NM_001278724.1 NM_014432.3 XM_006715506.2 XM_011535904.2 XM_017010954.1 XM_017010955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869389 c.439G>A p.D147N Chr.6: 137009457-137009457 on Build GRCh38
1311597 c.575G>A p.R192K Chr.6: 137009321-137009321 on Build GRCh38
5869388 c.439G>A p.D147N Chr.6: 137009457-137009457 on Build GRCh38
4945933 c.514A>G p.M172V Chr.6: 137009382-137009382 on Build GRCh38
5041510 c.539A>C p.K180T Chr.6: 137009357-137009357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200678057 Chr.6: 137009499-137009499 on Build GRCh38
rs765880720 Chr.6: 137009357-137009357 on Build GRCh38
rs562417668 Chr.6: 137009497-137009497 on Build GRCh38
rs779216027 Chr.6: 137009397-137009397 on Build GRCh38
rs374476894 Chr.6: 137009436-137009436 on Build GRCh38
rs764459408 Chr.6: 137009480-137009480 on Build GRCh38
rs764759808 Chr.6: 137009341-137009341 on Build GRCh38
rs765851640 Chr.6: 137009448-137009448 on Build GRCh38
rs143005532 Chr.6: 137009349-137009349 on Build GRCh38
rs754378765 Chr.6: 137009483-137009483 on Build GRCh38
rs750100276 Chr.6: 137009344-137009344 on Build GRCh38
rs746087823 Chr.6: 137009401-137009401 on Build GRCh38
rs988227465 Chr.6: 137009449-137009449 on Build GRCh38
rs759117119 Chr.6: 137009472-137009472 on Build GRCh38
rs780336302 Chr.6: 137009409-137009409 on Build GRCh38
rs776665874 Chr.6: 137009430-137009430 on Build GRCh38
rs754111382 Chr.6: 137009375-137009375 on Build GRCh38
rs150333567 Chr.6: 137009455-137009455 on Build GRCh38
rs761470204 Chr.6: 137009431-137009431 on Build GRCh38
rs747024327 Chr.6: 137009500-137009500 on Build GRCh38
rs545881372 Chr.6: 137009393-137009393 on Build GRCh38
rs371581891 Chr.6: 137009498-137009498 on Build GRCh38
rs753199618 Chr.6: 137009361-137009361 on Build GRCh38
rs536577063 Chr.6: 137009350-137009350 on Build GRCh38
rs772784926 Chr.6: 137009441-137009441 on Build GRCh38
rs140193064 Chr.6: 137009371-137009371 on Build GRCh38
rs534517805 Chr.6: 137009332-137009332 on Build GRCh38
rs201864884 Chr.6: 137009428-137009428 on Build GRCh38
rs541826030 Chr.6: 137009486-137009486 on Build GRCh38
rs772272084 Chr.6: 137009408-137009408 on Build GRCh38
rs746924544 Chr.6: 137009414-137009414 on Build GRCh38
rs762662503 Chr.6: 137009445-137009445 on Build GRCh38
rs368638679 Chr.6: 137009469-137009469 on Build GRCh38
rs146644651 Chr.6: 137009366-137009366 on Build GRCh38
rs578163089 Chr.6: 137009465-137009465 on Build GRCh38

View Full Product Details