Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 96758095-96758354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH2
Entrez Gene ID
1730
Gene Name
diaphanous related formin 2
Gene Aliases
DIA, DIA2, DRF2, POF, POF2
UniGene
Hs.226483
Gene Chromosome Location
Chr.X: 96684663-97600598 on Build GRCh38
Transcripts
NM_006729.4 NM_007309.3

SNP Detail

SNP ID Chromosome Location
rs182507250 Chr.X: 96758273-96758273 on Build GRCh38
rs756902044 Chr.X: 96758204-96758204 on Build GRCh38
rs761985480 Chr.X: 96758277-96758277 on Build GRCh38
rs768494353 Chr.X: 96758249-96758249 on Build GRCh38
rs376703873 Chr.X: 96758130-96758130 on Build GRCh38
rs772300351 Chr.X: 96758283-96758283 on Build GRCh38
rs200925409 Chr.X: 96758128-96758128 on Build GRCh38
rs769628938 Chr.X: 96758212-96758212 on Build GRCh38
rs200157443 Chr.X: 96758171-96758171 on Build GRCh38
rs762012999 Chr.X: 96758159-96758159 on Build GRCh38
rs760751479 Chr.X: 96758295-96758295 on Build GRCh38
rs20363 Chr.X: 96758130-96758130 on Build GRCh38
rs754269506 Chr.X: 96758193-96758193 on Build GRCh38
rs201217520 Chr.X: 96758196-96758196 on Build GRCh38
rs751305324 Chr.X: 96758201-96758201 on Build GRCh38
rs762674879 Chr.X: 96758132-96758132 on Build GRCh38
rs768868812 Chr.X: 96758150-96758150 on Build GRCh38
rs751038280 Chr.X: 96758234-96758234 on Build GRCh38
rs201675918 Chr.X: 96758330-96758330 on Build GRCh38
rs773191386 Chr.X: 96758286-96758286 on Build GRCh38
rs761120702 Chr.X: 96758175-96758175 on Build GRCh38
rs138883597 Chr.X: 96758210-96758210 on Build GRCh38
rs386826554 Chr.X: 96758129-96758130 on Build GRCh38
rs750487048 Chr.X: 96758173-96758173 on Build GRCh38
rs774618818 Chr.X: 96758151-96758151 on Build GRCh38
rs755327169 Chr.X: 96758194-96758194 on Build GRCh38
rs2297827 Chr.X: 96758129-96758129 on Build GRCh38
rs779897805 Chr.X: 96758217-96758217 on Build GRCh38
rs766861897 Chr.X: 96758185-96758185 on Build GRCh38
rs780596473 Chr.X: 96758208-96758208 on Build GRCh38

View Full Product Details