Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143567396-143567645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH6
Entrez Gene ID
642475
Gene Name
maestro heat like repeat family member 6
Gene Aliases
C8orf73
UniGene
Hs.531406
Gene Chromosome Location
Chr.8: 143566189-143572758 on Build GRCh38
Transcripts
NM_001100878.1 XM_011517214.1 XM_011517215.1 XM_011517216.2 XM_011517217.1 XM_011517218.1 XM_011517221.2 XM_011517222.1 XM_011517223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750070 c.1944A>G p.R648R Chr.8: 143567455-143567455 on Build GRCh38
5669219 c.1933G>T p.D645Y Chr.8: 143567611-143567611 on Build GRCh38
4162734 c.1968T>C p.A656A Chr.8: 143567431-143567431 on Build GRCh38
5573275 c.2001G>T p.Q667H Chr.8: 143567398-143567398 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371725467 Chr.8: 143567571-143567571 on Build GRCh38
rs995252111 Chr.8: 143567425-143567425 on Build GRCh38
rs566361981 Chr.8: 143567507-143567507 on Build GRCh38
rs904131255 Chr.8: 143567549-143567549 on Build GRCh38
rs770742977 Chr.8: 143567490-143567490 on Build GRCh38
rs573154588 Chr.8: 143567424-143567424 on Build GRCh38
rs745812293 Chr.8: 143567514-143567514 on Build GRCh38
rs200551481 Chr.8: 143567605-143567605 on Build GRCh38
rs373607024 Chr.8: 143567580-143567580 on Build GRCh38
rs529516791 Chr.8: 143567446-143567446 on Build GRCh38
rs1002517537 Chr.8: 143567478-143567478 on Build GRCh38
rs746842560 Chr.8: 143567489-143567489 on Build GRCh38
rs780311664 Chr.8: 143567479-143567479 on Build GRCh38
rs891016985 Chr.8: 143567412-143567412 on Build GRCh38
rs938101886 Chr.8: 143567505-143567505 on Build GRCh38
rs1042648062 Chr.8: 143567547-143567547 on Build GRCh38
rs762627744 Chr.8: 143567448-143567448 on Build GRCh38
rs898365192 Chr.8: 143567603-143567603 on Build GRCh38
rs912473606 Chr.8: 143567536-143567536 on Build GRCh38
rs1040020498 Chr.8: 143567587-143567587 on Build GRCh38
rs756620565 Chr.8: 143567469-143567469 on Build GRCh38
rs774049920 Chr.8: 143567615-143567615 on Build GRCh38
rs527267049 Chr.8: 143567517-143567517 on Build GRCh38
rs13254352 Chr.8: 143567480-143567480 on Build GRCh38
rs563097702 Chr.8: 143567472-143567472 on Build GRCh38
rs767977026 Chr.8: 143567463-143567463 on Build GRCh38
rs997678326 Chr.8: 143567606-143567606 on Build GRCh38
rs968865375 Chr.8: 143567501-143567501 on Build GRCh38
rs776229349 Chr.8: 143567494-143567494 on Build GRCh38
rs367745141 Chr.8: 143567583-143567583 on Build GRCh38
rs779161406 Chr.8: 143567487-143567487 on Build GRCh38
rs13268196 Chr.8: 143567431-143567431 on Build GRCh38
rs551442265 Chr.8: 143567481-143567481 on Build GRCh38
rs977531746 Chr.8: 143567503-143567503 on Build GRCh38
rs746796492 Chr.8: 143567423-143567423 on Build GRCh38
rs763694216 Chr.8: 143567612-143567612 on Build GRCh38
rs776622845 Chr.8: 143567426-143567426 on Build GRCh38
rs556113134 Chr.8: 143567593-143567593 on Build GRCh38
rs1056596402 Chr.8: 143567516-143567516 on Build GRCh38
rs750930123 Chr.8: 143567468-143567468 on Build GRCh38
rs751919075 Chr.8: 143567454-143567454 on Build GRCh38
rs774917378 Chr.8: 143567434-143567434 on Build GRCh38
rs745451354 Chr.8: 143567572-143567587 on Build GRCh38
rs142293548 Chr.8: 143567470-143567472 on Build GRCh38
rs567183367 Chr.8: 143567543-143567543 on Build GRCh38
rs1007145252 Chr.8: 143567413-143567413 on Build GRCh38
rs772904800 Chr.8: 143567419-143567419 on Build GRCh38
rs763691086 Chr.8: 143567450-143567450 on Build GRCh38
rs540947267 Chr.8: 143567430-143567430 on Build GRCh38
rs371571650 Chr.8: 143567618-143567618 on Build GRCh38
rs201434998 Chr.8: 143567585-143567585 on Build GRCh38
rs10097556 Chr.8: 143567455-143567455 on Build GRCh38
rs13254590 Chr.8: 143567520-143567520 on Build GRCh38
rs1039501236 Chr.8: 143567416-143567416 on Build GRCh38
rs376370027 Chr.8: 143567531-143567531 on Build GRCh38
rs924671678 Chr.8: 143567508-143567508 on Build GRCh38

View Full Product Details